Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 63049965-63050463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTX1
Entrez Gene ID
5013
Gene Name
orthodenticle homeobox 1
Gene Aliases
-
UniGene
Hs.734360
Gene Chromosome Location
Chr.2: 63050057-63057831 on Build GRCh38
Transcripts
NM_001199770.1

SNP Detail

SNP ID Chromosome Location
rs923017362 Chr.2: 63050073-63050073 on Build GRCh38
rs13011372 Chr.2: 63050345-63050345 on Build GRCh38
rs1021118748 Chr.2: 63050180-63050180 on Build GRCh38
rs540155965 Chr.2: 63050346-63050346 on Build GRCh38
rs1016488062 Chr.2: 63050040-63050040 on Build GRCh38
rs565156084 Chr.2: 63050402-63050402 on Build GRCh38
rs918348061 Chr.2: 63050412-63050412 on Build GRCh38
rs1035232466 Chr.2: 63050371-63050371 on Build GRCh38
rs534244670 Chr.2: 63050128-63050128 on Build GRCh38
rs572745868 Chr.2: 63050306-63050306 on Build GRCh38
rs188234433 Chr.2: 63050336-63050336 on Build GRCh38
rs982371185 Chr.2: 63050219-63050219 on Build GRCh38
rs897811006 Chr.2: 63050153-63050153 on Build GRCh38
rs941082120 Chr.2: 63050281-63050281 on Build GRCh38
rs986601468 Chr.2: 63050428-63050428 on Build GRCh38
rs867800525 Chr.2: 63050231-63050231 on Build GRCh38
rs897015187 Chr.2: 63050111-63050111 on Build GRCh38
rs979099095 Chr.2: 63050050-63050050 on Build GRCh38
rs997194254 Chr.2: 63050443-63050443 on Build GRCh38
rs962717953 Chr.2: 63050400-63050400 on Build GRCh38
rs1013056563 Chr.2: 63050027-63050027 on Build GRCh38
rs959619066 Chr.2: 63050375-63050375 on Build GRCh38
rs956373672 Chr.2: 63050267-63050267 on Build GRCh38
rs948823502 Chr.2: 63050387-63050387 on Build GRCh38
rs904805933 Chr.2: 63050256-63050256 on Build GRCh38
rs893846483 Chr.2: 63050341-63050341 on Build GRCh38
rs933817993 Chr.2: 63050424-63050424 on Build GRCh38
rs1016608041 Chr.2: 63050399-63050399 on Build GRCh38
rs376672736 Chr.2: 63050051-63050051 on Build GRCh38
rs928214829 Chr.2: 63050223-63050223 on Build GRCh38
rs1050996812 Chr.2: 63050313-63050313 on Build GRCh38
rs975725401 Chr.2: 63050268-63050268 on Build GRCh38
rs575380144 Chr.2: 63050305-63050305 on Build GRCh38
rs894536595 Chr.2: 63050212-63050214 on Build GRCh38
rs889700324 Chr.2: 63050320-63050320 on Build GRCh38
rs998995010 Chr.2: 63050423-63050423 on Build GRCh38
rs571573492 Chr.2: 63050273-63050273 on Build GRCh38
rs768385088 Chr.2: 63050131-63050131 on Build GRCh38
rs562565848 Chr.2: 63050006-63050006 on Build GRCh38
rs890445001 Chr.2: 63050126-63050126 on Build GRCh38
rs780577611 Chr.2: 63050049-63050049 on Build GRCh38
rs747327268 Chr.2: 63050078-63050078 on Build GRCh38
rs1013504496 Chr.2: 63050218-63050218 on Build GRCh38
rs538983198 Chr.2: 63050293-63050293 on Build GRCh38
rs549647672 Chr.2: 63050072-63050072 on Build GRCh38
rs902651881 Chr.2: 63050357-63050357 on Build GRCh38
rs567851745 Chr.2: 63050087-63050087 on Build GRCh38
rs1051478850 Chr.2: 63050114-63050114 on Build GRCh38
rs1011440637 Chr.2: 63050166-63050166 on Build GRCh38
rs909131530 Chr.2: 63050115-63050115 on Build GRCh38
rs776186502 Chr.2: 63050270-63050270 on Build GRCh38
rs547251630 Chr.2: 63050196-63050196 on Build GRCh38
rs904680151 Chr.2: 63050406-63050406 on Build GRCh38
rs968957390 Chr.2: 63050046-63050046 on Build GRCh38
rs1030702355 Chr.2: 63050265-63050265 on Build GRCh38
rs958120694 Chr.2: 63050158-63050158 on Build GRCh38
rs991105493 Chr.2: 63050381-63050381 on Build GRCh38
rs1004137313 Chr.2: 63050364-63050364 on Build GRCh38
rs895024221 Chr.2: 63050301-63050330 on Build GRCh38
rs557177878 Chr.2: 63050302-63050302 on Build GRCh38
rs531134174 Chr.2: 63050070-63050070 on Build GRCh38
rs1017602148 Chr.2: 63050150-63050150 on Build GRCh38
rs959002739 Chr.2: 63050224-63050224 on Build GRCh38
rs938794593 Chr.2: 63050193-63050193 on Build GRCh38
rs943336602 Chr.2: 63050324-63050324 on Build GRCh38
rs998112623 Chr.2: 63050113-63050113 on Build GRCh38
rs990488684 Chr.2: 63050262-63050262 on Build GRCh38
rs1021760428 Chr.2: 63050227-63050227 on Build GRCh38

View Full Product Details