Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 68463883-68464431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO48
Entrez Gene ID
554251
Gene Name
F-box protein 48
Gene Aliases
-
UniGene
Hs.164117
Gene Chromosome Location
Chr.2: 68459419-68467325 on Build GRCh38
Transcripts
NM_001024680.2 XM_005264407.2 XM_017004437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6249882 c.414C>A p.Y138* Chr.2: 68464263-68464263 on Build GRCh38
6327734 c.464G>A p.R155K Chr.2: 68464213-68464213 on Build GRCh38
4141197 c.339A>T p.K113N Chr.2: 68464338-68464338 on Build GRCh38
1615037 c.330G>T p.Q110H Chr.2: 68464347-68464347 on Build GRCh38
5708289 c.344A>G p.K115R Chr.2: 68464333-68464333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114287275 Chr.2: 68464403-68464403 on Build GRCh38
rs901499063 Chr.2: 68464139-68464139 on Build GRCh38
rs36078393 Chr.2: 68464243-68464244 on Build GRCh38
rs755835362 Chr.2: 68464202-68464202 on Build GRCh38
rs764738675 Chr.2: 68464062-68464062 on Build GRCh38
rs946803228 Chr.2: 68464134-68464134 on Build GRCh38
rs775771674 Chr.2: 68464273-68464273 on Build GRCh38
rs945195061 Chr.2: 68464067-68464067 on Build GRCh38
rs766047152 Chr.2: 68464321-68464321 on Build GRCh38
rs778325729 Chr.2: 68464230-68464230 on Build GRCh38
rs1015093567 Chr.2: 68463986-68463986 on Build GRCh38
rs757758296 Chr.2: 68464138-68464138 on Build GRCh38
rs746605408 Chr.2: 68464371-68464371 on Build GRCh38
rs1028718868 Chr.2: 68464344-68464344 on Build GRCh38
rs148116960 Chr.2: 68464277-68464277 on Build GRCh38
rs1018881765 Chr.2: 68464407-68464407 on Build GRCh38
rs570294284 Chr.2: 68464250-68464250 on Build GRCh38
rs950430414 Chr.2: 68464214-68464214 on Build GRCh38
rs987983037 Chr.2: 68464113-68464113 on Build GRCh38
rs76149797 Chr.2: 68464266-68464266 on Build GRCh38
rs771333794 Chr.2: 68464383-68464383 on Build GRCh38
rs770098566 Chr.2: 68464402-68464403 on Build GRCh38
rs981212294 Chr.2: 68463965-68463965 on Build GRCh38
rs781778444 Chr.2: 68464340-68464344 on Build GRCh38
rs939455171 Chr.2: 68463975-68463975 on Build GRCh38
rs757868369 Chr.2: 68464356-68464356 on Build GRCh38
rs752041508 Chr.2: 68464195-68464195 on Build GRCh38
rs777395914 Chr.2: 68464257-68464257 on Build GRCh38
rs535079096 Chr.2: 68464086-68464090 on Build GRCh38
rs777430196 Chr.2: 68464366-68464366 on Build GRCh38
rs374567451 Chr.2: 68464301-68464301 on Build GRCh38
rs751708213 Chr.2: 68464290-68464290 on Build GRCh38
rs201883747 Chr.2: 68464178-68464178 on Build GRCh38
rs747810102 Chr.2: 68464236-68464236 on Build GRCh38
rs963153462 Chr.2: 68464015-68464015 on Build GRCh38
rs377702943 Chr.2: 68464389-68464389 on Build GRCh38
rs778805966 Chr.2: 68464345-68464345 on Build GRCh38
rs748525686 Chr.2: 68464393-68464394 on Build GRCh38
rs987399545 Chr.2: 68464076-68464076 on Build GRCh38
rs1028648345 Chr.2: 68464027-68464027 on Build GRCh38
rs1004343330 Chr.2: 68464239-68464239 on Build GRCh38
rs911788383 Chr.2: 68464079-68464079 on Build GRCh38
rs777229205 Chr.2: 68464268-68464268 on Build GRCh38
rs150621874 Chr.2: 68464328-68464328 on Build GRCh38
rs752267611 Chr.2: 68464349-68464349 on Build GRCh38
rs528078244 Chr.2: 68464145-68464145 on Build GRCh38
rs992544829 Chr.2: 68464340-68464340 on Build GRCh38
rs115907946 Chr.2: 68464135-68464135 on Build GRCh38
rs759896922 Chr.2: 68464270-68464270 on Build GRCh38
rs370342467 Chr.2: 68464390-68464390 on Build GRCh38
rs915349171 Chr.2: 68464125-68464125 on Build GRCh38
rs925090180 Chr.2: 68464393-68464393 on Build GRCh38
rs753536356 Chr.2: 68464218-68464218 on Build GRCh38
rs962969253 Chr.2: 68464267-68464267 on Build GRCh38
rs780088215 Chr.2: 68464204-68464204 on Build GRCh38
rs570526658 Chr.2: 68464044-68464044 on Build GRCh38
rs750299942 Chr.2: 68464193-68464193 on Build GRCh38
rs371079566 Chr.2: 68464263-68464263 on Build GRCh38
rs547368977 Chr.2: 68463970-68463970 on Build GRCh38
rs775813844 Chr.2: 68463963-68463963 on Build GRCh38
rs1056537914 Chr.2: 68464003-68464003 on Build GRCh38
rs749130682 Chr.2: 68464209-68464209 on Build GRCh38
rs764450646 Chr.2: 68464280-68464280 on Build GRCh38
rs146033276 Chr.2: 68464341-68464341 on Build GRCh38
rs776703384 Chr.2: 68464182-68464182 on Build GRCh38
rs770008783 Chr.2: 68464271-68464271 on Build GRCh38
rs527805800 Chr.2: 68463933-68463933 on Build GRCh38
rs572157655 Chr.2: 68464309-68464309 on Build GRCh38
rs956750263 Chr.2: 68464109-68464109 on Build GRCh38
rs755534404 Chr.2: 68464196-68464197 on Build GRCh38
rs994951884 Chr.2: 68464021-68464021 on Build GRCh38
rs1004344026 Chr.2: 68463972-68463972 on Build GRCh38
rs781355942 Chr.2: 68464388-68464388 on Build GRCh38

View Full Product Details