Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73386066-73386574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALMS1
Entrez Gene ID
7840
Gene Name
ALMS1, centrosome and basal body associated protein
Gene Aliases
ALSS
UniGene
Hs.184720
Gene Chromosome Location
Chr.2: 73385758-73609919 on Build GRCh38
Transcripts
NM_015120.4

SNP Detail

SNP ID Chromosome Location
rs760639277 Chr.2: 73386169-73386169 on Build GRCh38
rs956834448 Chr.2: 73386133-73386133 on Build GRCh38
rs899220304 Chr.2: 73386240-73386240 on Build GRCh38
rs541590073 Chr.2: 73386404-73386404 on Build GRCh38
rs886949774 Chr.2: 73386203-73386203 on Build GRCh38
rs771370849 Chr.2: 73386124-73386124 on Build GRCh38
rs764095354 Chr.2: 73386150-73386150 on Build GRCh38
rs1037793096 Chr.2: 73386101-73386101 on Build GRCh38
rs778690416 Chr.2: 73386202-73386202 on Build GRCh38
rs553661392 Chr.2: 73386171-73386171 on Build GRCh38
rs890457403 Chr.2: 73386433-73386433 on Build GRCh38
rs543499940 Chr.2: 73386283-73386283 on Build GRCh38
rs1008828017 Chr.2: 73386438-73386438 on Build GRCh38
rs554126571 Chr.2: 73386255-73386255 on Build GRCh38
rs1052647164 Chr.2: 73386493-73386493 on Build GRCh38
rs927643756 Chr.2: 73386320-73386320 on Build GRCh38
rs938963631 Chr.2: 73386321-73386321 on Build GRCh38
rs1008058925 Chr.2: 73386318-73386318 on Build GRCh38
rs1003701204 Chr.2: 73386214-73386214 on Build GRCh38
rs905373886 Chr.2: 73386087-73386087 on Build GRCh38
rs942157242 Chr.2: 73386485-73386485 on Build GRCh38
rs956562037 Chr.2: 73386228-73386228 on Build GRCh38
rs747750620 Chr.2: 73386113-73386113 on Build GRCh38
rs763023186 Chr.2: 73386149-73386149 on Build GRCh38
rs777177630 Chr.2: 73386140-73386140 on Build GRCh38
rs970384610 Chr.2: 73386426-73386426 on Build GRCh38
rs576081925 Chr.2: 73386256-73386256 on Build GRCh38
rs929370698 Chr.2: 73386528-73386528 on Build GRCh38
rs754611450 Chr.2: 73386183-73386183 on Build GRCh38
rs750326619 Chr.2: 73386151-73386151 on Build GRCh38
rs778271633 Chr.2: 73386120-73386120 on Build GRCh38
rs758707920 Chr.2: 73386440-73386440 on Build GRCh38
rs755847221 Chr.2: 73386096-73386096 on Build GRCh38
rs747490719 Chr.2: 73386223-73386223 on Build GRCh38
rs538598970 Chr.2: 73386146-73386146 on Build GRCh38
rs1032491057 Chr.2: 73386397-73386397 on Build GRCh38
rs941187307 Chr.2: 73386347-73386347 on Build GRCh38
rs1031172088 Chr.2: 73386267-73386267 on Build GRCh38
rs754662930 Chr.2: 73386103-73386103 on Build GRCh38
rs879709360 Chr.2: 73386308-73386308 on Build GRCh38
rs1037776062 Chr.2: 73386293-73386293 on Build GRCh38
rs1046964700 Chr.2: 73386191-73386191 on Build GRCh38
rs577376918 Chr.2: 73386298-73386298 on Build GRCh38
rs775431837 Chr.2: 73386148-73386148 on Build GRCh38
rs1038210821 Chr.2: 73386234-73386234 on Build GRCh38
rs926889872 Chr.2: 73386459-73386459 on Build GRCh38
rs778264668 Chr.2: 73386109-73386109 on Build GRCh38
rs772450464 Chr.2: 73386117-73386117 on Build GRCh38
rs920702326 Chr.2: 73386389-73386389 on Build GRCh38
rs996611487 Chr.2: 73386248-73386248 on Build GRCh38
rs1022489904 Chr.2: 73386297-73386297 on Build GRCh38
rs1051737876 Chr.2: 73386303-73386303 on Build GRCh38
rs372177656 Chr.2: 73386429-73386429 on Build GRCh38
rs551575560 Chr.2: 73386222-73386222 on Build GRCh38
rs919433151 Chr.2: 73386489-73386489 on Build GRCh38
rs879809186 Chr.2: 73386327-73386327 on Build GRCh38
rs770203539 Chr.2: 73386147-73386147 on Build GRCh38
rs1018185421 Chr.2: 73386334-73386334 on Build GRCh38
rs932084431 Chr.2: 73386410-73386410 on Build GRCh38
rs989576553 Chr.2: 73386277-73386277 on Build GRCh38
rs1025964615 Chr.2: 73386119-73386119 on Build GRCh38
rs11126399 Chr.2: 73386213-73386213 on Build GRCh38
rs6706235 Chr.2: 73386430-73386430 on Build GRCh38
rs956462305 Chr.2: 73386398-73386398 on Build GRCh38
rs1037849123 Chr.2: 73386486-73386486 on Build GRCh38
rs890744248 Chr.2: 73386251-73386251 on Build GRCh38
rs747529471 Chr.2: 73386121-73386121 on Build GRCh38
rs868854769 Chr.2: 73386280-73386280 on Build GRCh38
rs187868113 Chr.2: 73386249-73386249 on Build GRCh38
rs988309909 Chr.2: 73386400-73386400 on Build GRCh38
rs908436800 Chr.2: 73386344-73386344 on Build GRCh38
rs1038195171 Chr.2: 73386375-73386375 on Build GRCh38
rs982699513 Chr.2: 73386338-73386338 on Build GRCh38
rs558923755 Chr.2: 73386290-73386290 on Build GRCh38
rs1009097793 Chr.2: 73386252-73386252 on Build GRCh38
rs1018027045 Chr.2: 73386160-73386160 on Build GRCh38
rs989191437 Chr.2: 73386134-73386134 on Build GRCh38
rs866402031 Chr.2: 73386095-73386095 on Build GRCh38
rs35414379 Chr.2: 73386274-73386274 on Build GRCh38
rs368947491 Chr.2: 73386102-73386102 on Build GRCh38
rs752293247 Chr.2: 73386209-73386209 on Build GRCh38
rs750332908 Chr.2: 73386089-73386089 on Build GRCh38
rs956885717 Chr.2: 73386272-73386272 on Build GRCh38
rs765462976 Chr.2: 73386367-73386367 on Build GRCh38
rs753680275 Chr.2: 73386178-73386178 on Build GRCh38

View Full Product Details