Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74534013-74534520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOXL3
Entrez Gene ID
84695
Gene Name
lysyl oxidase like 3
Gene Aliases
LOXL
UniGene
Hs.469045
Gene Chromosome Location
Chr.2: 74532258-74555709 on Build GRCh38
Transcripts
NM_001289164.1 NM_001289165.1 NM_032603.3 XM_011533134.2 XM_017005112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4645475 c.2038G>A p.D680N Chr.2: 74534138-74534138 on Build GRCh38
4980658 c.2023G>C p.D675H Chr.2: 74534153-74534153 on Build GRCh38
7261503 c.1985T>C p.I662T Chr.2: 74534191-74534191 on Build GRCh38
1256610 c.1925C>T p.T642I Chr.2: 74534330-74534330 on Build GRCh38
3714313 c.1957G>T p.E653* Chr.2: 74534219-74534219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759687105 Chr.2: 74534255-74534255 on Build GRCh38
rs943319868 Chr.2: 74534073-74534073 on Build GRCh38
rs779845414 Chr.2: 74534306-74534306 on Build GRCh38
rs747000571 Chr.2: 74534433-74534433 on Build GRCh38
rs1016152170 Chr.2: 74534200-74534200 on Build GRCh38
rs770865200 Chr.2: 74534246-74534246 on Build GRCh38
rs746488976 Chr.2: 74534074-74534074 on Build GRCh38
rs753154543 Chr.2: 74534065-74534065 on Build GRCh38
rs753380842 Chr.2: 74534390-74534390 on Build GRCh38
rs747995192 Chr.2: 74534327-74534327 on Build GRCh38
rs776722155 Chr.2: 74534251-74534251 on Build GRCh38
rs749074923 Chr.2: 74534201-74534201 on Build GRCh38
rs147915651 Chr.2: 74534213-74534213 on Build GRCh38
rs769666456 Chr.2: 74534095-74534095 on Build GRCh38
rs202011389 Chr.2: 74534396-74534396 on Build GRCh38
rs972125257 Chr.2: 74534276-74534276 on Build GRCh38
rs756702540 Chr.2: 74534164-74534164 on Build GRCh38
rs757818416 Chr.2: 74534281-74534281 on Build GRCh38
rs200911515 Chr.2: 74534493-74534493 on Build GRCh38
rs375537672 Chr.2: 74534130-74534130 on Build GRCh38
rs368539606 Chr.2: 74534165-74534165 on Build GRCh38
rs752150504 Chr.2: 74534155-74534155 on Build GRCh38
rs778302814 Chr.2: 74534297-74534298 on Build GRCh38
rs373842923 Chr.2: 74534494-74534494 on Build GRCh38
rs17010021 Chr.2: 74534412-74534412 on Build GRCh38
rs145380329 Chr.2: 74534326-74534326 on Build GRCh38
rs754153697 Chr.2: 74534056-74534056 on Build GRCh38
rs766495239 Chr.2: 74534036-74534036 on Build GRCh38
rs6707302 Chr.2: 74534295-74534295 on Build GRCh38
rs765545235 Chr.2: 74534131-74534131 on Build GRCh38
rs369778040 Chr.2: 74534085-74534085 on Build GRCh38
rs369329249 Chr.2: 74534492-74534492 on Build GRCh38
rs539549422 Chr.2: 74534398-74534398 on Build GRCh38
rs767073805 Chr.2: 74534288-74534288 on Build GRCh38
rs774490993 Chr.2: 74534488-74534488 on Build GRCh38
rs778140028 Chr.2: 74534151-74534151 on Build GRCh38
rs769818115 Chr.2: 74534449-74534449 on Build GRCh38
rs766032709 Chr.2: 74534389-74534389 on Build GRCh38
rs774401306 Chr.2: 74534114-74534114 on Build GRCh38
rs771028309 Chr.2: 74534487-74534494 on Build GRCh38
rs779933189 Chr.2: 74534193-74534193 on Build GRCh38
rs777280011 Chr.2: 74534066-74534066 on Build GRCh38
rs756503932 Chr.2: 74534250-74534250 on Build GRCh38
rs115590529 Chr.2: 74534267-74534267 on Build GRCh38
rs766518527 Chr.2: 74534116-74534116 on Build GRCh38
rs139838276 Chr.2: 74534112-74534112 on Build GRCh38
rs866546439 Chr.2: 74534237-74534237 on Build GRCh38
rs374282063 Chr.2: 74534279-74534279 on Build GRCh38
rs780142553 Chr.2: 74534468-74534468 on Build GRCh38
rs749962269 Chr.2: 74534289-74534289 on Build GRCh38
rs769903746 Chr.2: 74534338-74534338 on Build GRCh38
rs768559100 Chr.2: 74534219-74534219 on Build GRCh38
rs377029343 Chr.2: 74534491-74534491 on Build GRCh38
rs758822068 Chr.2: 74534140-74534140 on Build GRCh38
rs774786906 Chr.2: 74534352-74534352 on Build GRCh38
rs786204838 Chr.2: 74534149-74534149 on Build GRCh38
rs755651920 Chr.2: 74534386-74534386 on Build GRCh38
rs775669859 Chr.2: 74534341-74534341 on Build GRCh38
rs745568434 Chr.2: 74534089-74534089 on Build GRCh38
rs141824904 Chr.2: 74534084-74534084 on Build GRCh38
rs775757800 Chr.2: 74534260-74534260 on Build GRCh38
rs753259295 Chr.2: 74534059-74534060 on Build GRCh38
rs34021958 Chr.2: 74534155-74534156 on Build GRCh38
rs1024141954 Chr.2: 74534224-74534224 on Build GRCh38
rs764328902 Chr.2: 74534343-74534343 on Build GRCh38
rs745944878 Chr.2: 74534337-74534337 on Build GRCh38
rs753074856 Chr.2: 74534136-74534136 on Build GRCh38
rs765432167 Chr.2: 74534258-74534258 on Build GRCh38
rs751033818 Chr.2: 74534380-74534380 on Build GRCh38
rs374003067 Chr.2: 74534107-74534107 on Build GRCh38
rs748986886 Chr.2: 74534309-74534309 on Build GRCh38
rs997290217 Chr.2: 74534490-74534490 on Build GRCh38
rs938803949 Chr.2: 74534282-74534282 on Build GRCh38
rs778505746 Chr.2: 74534407-74534407 on Build GRCh38
rs749406839 Chr.2: 74534470-74534470 on Build GRCh38
rs779261659 Chr.2: 74534495-74534496 on Build GRCh38
rs755303568 Chr.2: 74534060-74534060 on Build GRCh38
rs754705976 Chr.2: 74534315-74534315 on Build GRCh38
rs764571491 Chr.2: 74534270-74534270 on Build GRCh38
rs748163885 Chr.2: 74534238-74534238 on Build GRCh38
rs202168669 Chr.2: 74534064-74534064 on Build GRCh38
rs904517258 Chr.2: 74534411-74534411 on Build GRCh38
rs891628709 Chr.2: 74534158-74534158 on Build GRCh38
rs373974142 Chr.2: 74534035-74534035 on Build GRCh38
rs769578496 Chr.2: 74534189-74534189 on Build GRCh38
rs768940930 Chr.2: 74534484-74534484 on Build GRCh38
rs758256615 Chr.2: 74534415-74534415 on Build GRCh38
rs768120899 Chr.2: 74534379-74534379 on Build GRCh38
rs919424776 Chr.2: 74534057-74534057 on Build GRCh38
rs370095028 Chr.2: 74534124-74534124 on Build GRCh38
rs374270177 Chr.2: 74534374-74534374 on Build GRCh38
rs749716992 Chr.2: 74534331-74534337 on Build GRCh38
rs762913695 Chr.2: 74534108-74534108 on Build GRCh38
rs763262128 Chr.2: 74534342-74534342 on Build GRCh38
rs551992223 Chr.2: 74534227-74534227 on Build GRCh38
rs1043203467 Chr.2: 74534333-74534333 on Build GRCh38
rs929424088 Chr.2: 74534373-74534373 on Build GRCh38
rs916087535 Chr.2: 74534319-74534319 on Build GRCh38
rs777658017 Chr.2: 74534427-74534427 on Build GRCh38
rs760663651 Chr.2: 74534115-74534115 on Build GRCh38
rs773192522 Chr.2: 74534330-74534330 on Build GRCh38

View Full Product Details