Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74854130-74854628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HK2
Entrez Gene ID
3099
Gene Name
hexokinase 2
Gene Aliases
HKII, HXK2
UniGene
Hs.406266
Gene Chromosome Location
Chr.2: 74832655-74893354 on Build GRCh38
Transcripts
NM_000189.4 XM_005264280.1 XM_011532807.1 XM_017003945.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1023102 c.125G>A p.R42Q Chr.2: 74854354-74854354 on Build GRCh38
74921 c.220G>T p.G74W Chr.2: 74854449-74854449 on Build GRCh38
1285380 c.65T>A p.V22D Chr.2: 74854294-74854294 on Build GRCh38
1023101 c.88C>T p.R30C Chr.2: 74854317-74854317 on Build GRCh38
6313208 c.125G>T p.R42L Chr.2: 74854354-74854354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020732705 Chr.2: 74854512-74854512 on Build GRCh38
rs372471328 Chr.2: 74854269-74854269 on Build GRCh38
rs375609405 Chr.2: 74854354-74854354 on Build GRCh38
rs1005328031 Chr.2: 74854419-74854419 on Build GRCh38
rs369629969 Chr.2: 74854424-74854424 on Build GRCh38
rs747711639 Chr.2: 74854328-74854328 on Build GRCh38
rs773421826 Chr.2: 74854425-74854425 on Build GRCh38
rs539959546 Chr.2: 74854244-74854244 on Build GRCh38
rs1055936058 Chr.2: 74854222-74854222 on Build GRCh38
rs760949940 Chr.2: 74854426-74854426 on Build GRCh38
rs566952576 Chr.2: 74854278-74854278 on Build GRCh38
rs201380686 Chr.2: 74854360-74854360 on Build GRCh38
rs770835565 Chr.2: 74854359-74854359 on Build GRCh38
rs765750267 Chr.2: 74854451-74854451 on Build GRCh38
rs778584249 Chr.2: 74854604-74854604 on Build GRCh38
rs745698718 Chr.2: 74854488-74854488 on Build GRCh38
rs751199172 Chr.2: 74854465-74854465 on Build GRCh38
rs991053474 Chr.2: 74854500-74854500 on Build GRCh38
rs200787018 Chr.2: 74854298-74854298 on Build GRCh38
rs777481912 Chr.2: 74854461-74854461 on Build GRCh38
rs532008664 Chr.2: 74854445-74854445 on Build GRCh38
rs780011538 Chr.2: 74854501-74854501 on Build GRCh38
rs142716543 Chr.2: 74854603-74854603 on Build GRCh38
rs756611794 Chr.2: 74854344-74854344 on Build GRCh38
rs764021667 Chr.2: 74854262-74854262 on Build GRCh38
rs555746033 Chr.2: 74854489-74854489 on Build GRCh38
rs547705655 Chr.2: 74854576-74854576 on Build GRCh38
rs35614058 Chr.2: 74854430-74854431 on Build GRCh38
rs1042358446 Chr.2: 74854237-74854237 on Build GRCh38
rs776702857 Chr.2: 74854260-74854260 on Build GRCh38
rs928812476 Chr.2: 74854545-74854545 on Build GRCh38
rs139553168 Chr.2: 74854400-74854400 on Build GRCh38
rs780811091 Chr.2: 74854486-74854486 on Build GRCh38
rs761901487 Chr.2: 74854273-74854273 on Build GRCh38
rs938936096 Chr.2: 74854218-74854218 on Build GRCh38
rs772127874 Chr.2: 74854505-74854505 on Build GRCh38
rs924035274 Chr.2: 74854527-74854527 on Build GRCh38
rs916127608 Chr.2: 74854569-74854569 on Build GRCh38
rs759575503 Chr.2: 74854261-74854261 on Build GRCh38
rs562340218 Chr.2: 74854231-74854231 on Build GRCh38
rs371308214 Chr.2: 74854242-74854242 on Build GRCh38
rs780432817 Chr.2: 74854431-74854431 on Build GRCh38
rs1054971137 Chr.2: 74854224-74854224 on Build GRCh38
rs749293640 Chr.2: 74854502-74854502 on Build GRCh38
rs982003047 Chr.2: 74854185-74854185 on Build GRCh38
rs369793096 Chr.2: 74854256-74854256 on Build GRCh38
rs750561945 Chr.2: 74854280-74854280 on Build GRCh38
rs148486945 Chr.2: 74854254-74854254 on Build GRCh38
rs915309601 Chr.2: 74854230-74854230 on Build GRCh38
rs1028213527 Chr.2: 74854439-74854439 on Build GRCh38
rs766799292 Chr.2: 74854314-74854314 on Build GRCh38
rs372666431 Chr.2: 74854353-74854353 on Build GRCh38
rs773297096 Chr.2: 74854507-74854507 on Build GRCh38
rs781053032 Chr.2: 74854367-74854367 on Build GRCh38
rs200036720 Chr.2: 74854437-74854437 on Build GRCh38
rs577191829 Chr.2: 74854553-74854553 on Build GRCh38
rs1042429540 Chr.2: 74854521-74854521 on Build GRCh38
rs142756990 Chr.2: 74854318-74854318 on Build GRCh38
rs773090783 Chr.2: 74854251-74854251 on Build GRCh38
rs759907165 Chr.2: 74854448-74854448 on Build GRCh38
rs764384651 Chr.2: 74854441-74854441 on Build GRCh38
rs544720864 Chr.2: 74854556-74854556 on Build GRCh38
rs753182125 Chr.2: 74854452-74854452 on Build GRCh38
rs774494528 Chr.2: 74854263-74854263 on Build GRCh38
rs529443537 Chr.2: 74854537-74854537 on Build GRCh38
rs937810942 Chr.2: 74854221-74854221 on Build GRCh38
rs534023973 Chr.2: 74854538-74854539 on Build GRCh38
rs977599499 Chr.2: 74854522-74854522 on Build GRCh38
rs900740871 Chr.2: 74854557-74854557 on Build GRCh38
rs555921699 Chr.2: 74854392-74854392 on Build GRCh38
rs779325514 Chr.2: 74854494-74854495 on Build GRCh38
rs754291294 Chr.2: 74854315-74854315 on Build GRCh38
rs567201785 Chr.2: 74854317-74854317 on Build GRCh38
rs190959382 Chr.2: 74854212-74854212 on Build GRCh38
rs372897379 Chr.2: 74854458-74854458 on Build GRCh38
rs756974043 Chr.2: 74854471-74854471 on Build GRCh38
rs927880840 Chr.2: 74854211-74854211 on Build GRCh38
rs376757872 Chr.2: 74854302-74854302 on Build GRCh38
rs774248814 Chr.2: 74854405-74854405 on Build GRCh38
rs747768103 Chr.2: 74854605-74854605 on Build GRCh38
rs928924960 Chr.2: 74854206-74854206 on Build GRCh38
rs761931853 Chr.2: 74854410-74854410 on Build GRCh38
rs756772477 Chr.2: 74854305-74854313 on Build GRCh38
rs758028308 Chr.2: 74854346-74854346 on Build GRCh38
rs756476705 Chr.2: 74854387-74854387 on Build GRCh38

View Full Product Details