Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86751808-86752322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RMND5A
Entrez Gene ID
64795
Gene Name
required for meiotic nuclear division 5 homolog A
Gene Aliases
CTLH, GID2, GID2A, RMD5, p44CTLH
UniGene
Hs.75277
Gene Chromosome Location
Chr.2: 86720291-86778041 on Build GRCh38
Transcripts
NM_022780.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6767053 c.327C>A p.C109* Chr.2: 86751937-86751937 on Build GRCh38
3583837 c.305G>C p.S102T Chr.2: 86751915-86751915 on Build GRCh38
6371101 c.400G>A p.V134M Chr.2: 86752010-86752010 on Build GRCh38
5021306 c.360G>A p.E120E Chr.2: 86751970-86751970 on Build GRCh38
3910740 c.378C>T p.F126F Chr.2: 86751988-86751988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934796109 Chr.2: 86752156-86752156 on Build GRCh38
rs1030027805 Chr.2: 86752137-86752137 on Build GRCh38
rs778635902 Chr.2: 86751871-86751871 on Build GRCh38
rs746811175 Chr.2: 86751850-86751850 on Build GRCh38
rs765647235 Chr.2: 86751880-86751880 on Build GRCh38
rs113845452 Chr.2: 86752042-86752042 on Build GRCh38
rs749575023 Chr.2: 86752075-86752075 on Build GRCh38
rs572813846 Chr.2: 86752103-86752103 on Build GRCh38
rs533793465 Chr.2: 86752243-86752243 on Build GRCh38
rs753101180 Chr.2: 86751885-86751885 on Build GRCh38
rs751972316 Chr.2: 86751931-86751931 on Build GRCh38
rs201698250 Chr.2: 86751916-86751916 on Build GRCh38
rs779790145 Chr.2: 86751966-86751966 on Build GRCh38
rs137956456 Chr.2: 86752297-86752297 on Build GRCh38
rs2887012 Chr.2: 86752294-86752294 on Build GRCh38
rs769569947 Chr.2: 86751964-86751964 on Build GRCh38
rs367596649 Chr.2: 86752029-86752029 on Build GRCh38
rs914874468 Chr.2: 86751951-86751951 on Build GRCh38
rs888621975 Chr.2: 86751935-86751935 on Build GRCh38
rs1051869837 Chr.2: 86752164-86752164 on Build GRCh38
rs759731685 Chr.2: 86751875-86751875 on Build GRCh38
rs1044958034 Chr.2: 86751900-86751900 on Build GRCh38
rs766404363 Chr.2: 86751849-86751849 on Build GRCh38
rs188891336 Chr.2: 86752090-86752090 on Build GRCh38
rs978221144 Chr.2: 86752043-86752043 on Build GRCh38
rs993145090 Chr.2: 86752120-86752120 on Build GRCh38
rs771008771 Chr.2: 86752159-86752159 on Build GRCh38
rs985936467 Chr.2: 86752229-86752229 on Build GRCh38
rs550283294 Chr.2: 86751955-86751955 on Build GRCh38
rs959268263 Chr.2: 86751908-86751908 on Build GRCh38
rs537969101 Chr.2: 86751937-86751937 on Build GRCh38
rs780829109 Chr.2: 86751942-86751942 on Build GRCh38
rs753598302 Chr.2: 86751861-86751861 on Build GRCh38
rs772466826 Chr.2: 86752296-86752296 on Build GRCh38
rs184216428 Chr.2: 86752085-86752085 on Build GRCh38
rs968530691 Chr.2: 86752276-86752276 on Build GRCh38
rs912182938 Chr.2: 86752262-86752262 on Build GRCh38
rs1017816117 Chr.2: 86752256-86752256 on Build GRCh38
rs954603573 Chr.2: 86752147-86752147 on Build GRCh38
rs768283493 Chr.2: 86752037-86752037 on Build GRCh38
rs201728271 Chr.2: 86751913-86751913 on Build GRCh38
rs895869403 Chr.2: 86751840-86751840 on Build GRCh38
rs554050534 Chr.2: 86752073-86752073 on Build GRCh38
rs774340705 Chr.2: 86752251-86752251 on Build GRCh38
rs756816715 Chr.2: 86751862-86751865 on Build GRCh38
rs754097991 Chr.2: 86751871-86751871 on Build GRCh38
rs778367566 Chr.2: 86751926-86751926 on Build GRCh38
rs10186663 Chr.2: 86751970-86751970 on Build GRCh38

View Full Product Details