Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88737891-88738394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPIA
Entrez Gene ID
22934
Gene Name
ribose 5-phosphate isomerase A
Gene Aliases
RPI, RPIAD
UniGene
Hs.469264
Gene Chromosome Location
Chr.2: 88691658-88750935 on Build GRCh38
Transcripts
NM_144563.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1023621 c.746T>C p.V249A Chr.2: 88737984-88737984 on Build GRCh38
1023622 c.794T>C p.V265A Chr.2: 88738032-88738032 on Build GRCh38
3963210 c.838+9A>C p.? Chr.2: 88738085-88738085 on Build GRCh38
7411648 c.777C>G p.D259E Chr.2: 88738015-88738015 on Build GRCh38
4758598 c.739-1G>A p.? Chr.2: 88737976-88737976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949879541 Chr.2: 88738350-88738350 on Build GRCh38
rs891563941 Chr.2: 88738068-88738068 on Build GRCh38
rs375154564 Chr.2: 88738109-88738109 on Build GRCh38
rs935635327 Chr.2: 88738295-88738295 on Build GRCh38
rs1006607451 Chr.2: 88738320-88738320 on Build GRCh38
rs138136751 Chr.2: 88738164-88738164 on Build GRCh38
rs80106793 Chr.2: 88737988-88737988 on Build GRCh38
rs534079393 Chr.2: 88738254-88738254 on Build GRCh38
rs372040051 Chr.2: 88738075-88738075 on Build GRCh38
rs754120268 Chr.2: 88738037-88738037 on Build GRCh38
rs748446592 Chr.2: 88738029-88738029 on Build GRCh38
rs544458883 Chr.2: 88738333-88738333 on Build GRCh38
rs780587081 Chr.2: 88738247-88738247 on Build GRCh38
rs759029647 Chr.2: 88738105-88738105 on Build GRCh38
rs1014412309 Chr.2: 88738085-88738085 on Build GRCh38
rs972770299 Chr.2: 88738193-88738193 on Build GRCh38
rs6742233 Chr.2: 88738136-88738136 on Build GRCh38
rs1014913222 Chr.2: 88738356-88738356 on Build GRCh38
rs996333602 Chr.2: 88738275-88738275 on Build GRCh38
rs780159716 Chr.2: 88738002-88738002 on Build GRCh38
rs374590507 Chr.2: 88737970-88737970 on Build GRCh38
rs765928346 Chr.2: 88738106-88738107 on Build GRCh38
rs962700656 Chr.2: 88738178-88738178 on Build GRCh38
rs992683861 Chr.2: 88738156-88738156 on Build GRCh38
rs765936895 Chr.2: 88737926-88737926 on Build GRCh38
rs373151536 Chr.2: 88738158-88738158 on Build GRCh38
rs147431137 Chr.2: 88738034-88738034 on Build GRCh38
rs730880316 Chr.2: 88738000-88738000 on Build GRCh38
rs750701985 Chr.2: 88737973-88737973 on Build GRCh38
rs749365715 Chr.2: 88738021-88738021 on Build GRCh38
rs554521901 Chr.2: 88738235-88738235 on Build GRCh38
rs572767402 Chr.2: 88738241-88738241 on Build GRCh38
rs974671398 Chr.2: 88738252-88738252 on Build GRCh38
rs980009794 Chr.2: 88738171-88738171 on Build GRCh38
rs201938367 Chr.2: 88738028-88738028 on Build GRCh38
rs538894128 Chr.2: 88738130-88738130 on Build GRCh38
rs955674781 Chr.2: 88738211-88738211 on Build GRCh38
rs758469360 Chr.2: 88738242-88738242 on Build GRCh38
rs752523900 Chr.2: 88737957-88737957 on Build GRCh38
rs535945161 Chr.2: 88738201-88738201 on Build GRCh38
rs143609946 Chr.2: 88738255-88738255 on Build GRCh38
rs758279517 Chr.2: 88737958-88737958 on Build GRCh38
rs962165895 Chr.2: 88738142-88738142 on Build GRCh38
rs201892603 Chr.2: 88738051-88738051 on Build GRCh38
rs945803168 Chr.2: 88738217-88738217 on Build GRCh38
rs765158489 Chr.2: 88737952-88737952 on Build GRCh38
rs1040550329 Chr.2: 88738232-88738232 on Build GRCh38
rs753341690 Chr.2: 88737929-88737929 on Build GRCh38
rs776203731 Chr.2: 88738104-88738104 on Build GRCh38
rs747463465 Chr.2: 88738371-88738371 on Build GRCh38
rs186877065 Chr.2: 88738368-88738368 on Build GRCh38
rs765480027 Chr.2: 88738154-88738154 on Build GRCh38
rs1185172 Chr.2: 88738268-88738268 on Build GRCh38
rs759489532 Chr.2: 88737933-88737933 on Build GRCh38
rs756213561 Chr.2: 88737987-88737987 on Build GRCh38
rs183264672 Chr.2: 88738289-88738289 on Build GRCh38
rs943319164 Chr.2: 88737944-88737944 on Build GRCh38
rs1052799566 Chr.2: 88738304-88738304 on Build GRCh38
rs1050600194 Chr.2: 88738291-88738291 on Build GRCh38
rs1045860241 Chr.2: 88738351-88738351 on Build GRCh38
rs770533953 Chr.2: 88738082-88738082 on Build GRCh38
rs1029238068 Chr.2: 88738096-88738096 on Build GRCh38
rs757359784 Chr.2: 88738102-88738102 on Build GRCh38
rs906035642 Chr.2: 88738352-88738352 on Build GRCh38
rs952037718 Chr.2: 88738114-88738114 on Build GRCh38
rs1006224841 Chr.2: 88738123-88738123 on Build GRCh38
rs368582524 Chr.2: 88738027-88738027 on Build GRCh38
rs544792088 Chr.2: 88738306-88738306 on Build GRCh38
rs750532834 Chr.2: 88738208-88738208 on Build GRCh38

View Full Product Details