Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 88749900-88750410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPIA
Entrez Gene ID
22934
Gene Name
ribose 5-phosphate isomerase A
Gene Aliases
RPI, RPIAD
UniGene
Hs.469264
Gene Chromosome Location
Chr.2: 88691658-88750935 on Build GRCh38
Transcripts
NM_144563.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3583926 c.930C>T p.F310F Chr.2: 88750072-88750072 on Build GRCh38
722923 c.839-1G>T p.? Chr.2: 88749980-88749980 on Build GRCh38
270069 c.847G>A p.D283N Chr.2: 88749989-88749989 on Build GRCh38
6343710 c.854G>T p.G285V Chr.2: 88749996-88749996 on Build GRCh38
4001783 c.866A>G p.N289S Chr.2: 88750008-88750008 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745308118 Chr.2: 88750109-88750109 on Build GRCh38
rs574766296 Chr.2: 88750334-88750334 on Build GRCh38
rs375899017 Chr.2: 88750093-88750093 on Build GRCh38
rs751285828 Chr.2: 88749983-88749985 on Build GRCh38
rs755019666 Chr.2: 88750006-88750006 on Build GRCh38
rs761383830 Chr.2: 88750077-88750077 on Build GRCh38
rs748677263 Chr.2: 88750062-88750062 on Build GRCh38
rs762792557 Chr.2: 88749953-88749953 on Build GRCh38
rs754581835 Chr.2: 88750026-88750026 on Build GRCh38
rs142235173 Chr.2: 88750148-88750148 on Build GRCh38
rs908562428 Chr.2: 88750067-88750067 on Build GRCh38
rs943767734 Chr.2: 88750213-88750213 on Build GRCh38
rs1043354011 Chr.2: 88749947-88749947 on Build GRCh38
rs183747347 Chr.2: 88750107-88750107 on Build GRCh38
rs888684279 Chr.2: 88750283-88750283 on Build GRCh38
rs1052569032 Chr.2: 88750359-88750359 on Build GRCh38
rs188862738 Chr.2: 88750238-88750238 on Build GRCh38
rs76788057 Chr.2: 88750368-88750368 on Build GRCh38
rs766575101 Chr.2: 88749985-88749985 on Build GRCh38
rs564562388 Chr.2: 88750118-88750118 on Build GRCh38
rs531713655 Chr.2: 88750119-88750119 on Build GRCh38
rs969166434 Chr.2: 88750378-88750378 on Build GRCh38
rs986636562 Chr.2: 88750202-88750202 on Build GRCh38
rs1023025019 Chr.2: 88750373-88750373 on Build GRCh38
rs1041194642 Chr.2: 88750222-88750222 on Build GRCh38
rs936613927 Chr.2: 88750170-88750171 on Build GRCh38
rs369448619 Chr.2: 88749974-88749974 on Build GRCh38
rs113052379 Chr.2: 88750357-88750358 on Build GRCh38
rs550119609 Chr.2: 88750121-88750121 on Build GRCh38
rs200612417 Chr.2: 88750053-88750053 on Build GRCh38
rs548137310 Chr.2: 88750158-88750158 on Build GRCh38
rs559102861 Chr.2: 88750056-88750056 on Build GRCh38
rs923379451 Chr.2: 88750187-88750187 on Build GRCh38
rs980560852 Chr.2: 88750381-88750381 on Build GRCh38
rs778084229 Chr.2: 88750081-88750081 on Build GRCh38
rs942553368 Chr.2: 88750256-88750256 on Build GRCh38
rs1027183196 Chr.2: 88750254-88750254 on Build GRCh38
rs151226092 Chr.2: 88750319-88750319 on Build GRCh38
rs763998933 Chr.2: 88749968-88749968 on Build GRCh38
rs947893107 Chr.2: 88750135-88750135 on Build GRCh38
rs1044937890 Chr.2: 88750146-88750146 on Build GRCh38
rs201414020 Chr.2: 88750047-88750047 on Build GRCh38
rs568376633 Chr.2: 88750128-88750128 on Build GRCh38
rs997029120 Chr.2: 88750251-88750251 on Build GRCh38
rs756328504 Chr.2: 88749975-88749975 on Build GRCh38
rs199784219 Chr.2: 88750078-88750078 on Build GRCh38
rs903787839 Chr.2: 88750302-88750302 on Build GRCh38
rs566268591 Chr.2: 88750190-88750190 on Build GRCh38
rs1030672871 Chr.2: 88750059-88750059 on Build GRCh38
rs1052103435 Chr.2: 88750196-88750197 on Build GRCh38
rs747444505 Chr.2: 88750091-88750091 on Build GRCh38
rs1043594321 Chr.2: 88750301-88750301 on Build GRCh38
rs779231148 Chr.2: 88750358-88750358 on Build GRCh38
rs992887662 Chr.2: 88750340-88750340 on Build GRCh38
rs368918368 Chr.2: 88750117-88750117 on Build GRCh38
rs972063765 Chr.2: 88750166-88750166 on Build GRCh38
rs754444410 Chr.2: 88750057-88750060 on Build GRCh38
rs765128115 Chr.2: 88750126-88750126 on Build GRCh38
rs576062000 Chr.2: 88750252-88750252 on Build GRCh38
rs867796610 Chr.2: 88750192-88750192 on Build GRCh38
rs142341148 Chr.2: 88750000-88750000 on Build GRCh38
rs982787963 Chr.2: 88750066-88750066 on Build GRCh38
rs540964815 Chr.2: 88750008-88750008 on Build GRCh38
rs35508879 Chr.2: 88750271-88750272 on Build GRCh38
rs985018216 Chr.2: 88750094-88750094 on Build GRCh38
rs558073148 Chr.2: 88750248-88750248 on Build GRCh38
rs964215339 Chr.2: 88749938-88749938 on Build GRCh38
rs925217202 Chr.2: 88750169-88750169 on Build GRCh38
rs886056424 Chr.2: 88750369-88750369 on Build GRCh38
rs892159755 Chr.2: 88750212-88750212 on Build GRCh38
rs552241099 Chr.2: 88750116-88750116 on Build GRCh38
rs942646573 Chr.2: 88749926-88749926 on Build GRCh38
rs1013747116 Chr.2: 88750087-88750087 on Build GRCh38
rs775571112 Chr.2: 88749951-88749951 on Build GRCh38

View Full Product Details