Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 101383607-101384116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREG2
Entrez Gene ID
200407
Gene Name
cellular repressor of E1A stimulated genes 2
Gene Aliases
-
UniGene
Hs.732289
Gene Chromosome Location
Chr.2: 101348354-101387970 on Build GRCh38
Transcripts
NM_153836.3 XM_011510777.2 XM_017003565.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3894391 c.511C>T p.P171S Chr.2: 101383633-101383633 on Build GRCh38
3701906 c.464A>G p.N155S Chr.2: 101383680-101383680 on Build GRCh38
1004578 c.516C>A p.F172L Chr.2: 101383628-101383628 on Build GRCh38
5859773 c.448_449GG>AA p.G150K Chr.2: 101383695-101383696 on Build GRCh38
1004579 c.507G>A p.G169G Chr.2: 101383637-101383637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572785251 Chr.2: 101383657-101383657 on Build GRCh38
rs903801225 Chr.2: 101383951-101383951 on Build GRCh38
rs767953794 Chr.2: 101383633-101383633 on Build GRCh38
rs756389827 Chr.2: 101383815-101383815 on Build GRCh38
rs992153700 Chr.2: 101383888-101383888 on Build GRCh38
rs1032223449 Chr.2: 101383814-101383814 on Build GRCh38
rs778883670 Chr.2: 101383681-101383693 on Build GRCh38
rs748123128 Chr.2: 101383670-101383670 on Build GRCh38
rs944709538 Chr.2: 101383805-101383806 on Build GRCh38
rs958826092 Chr.2: 101383885-101383885 on Build GRCh38
rs893711673 Chr.2: 101384074-101384074 on Build GRCh38
rs769349901 Chr.2: 101383705-101383705 on Build GRCh38
rs138973393 Chr.2: 101383669-101383669 on Build GRCh38
rs144554988 Chr.2: 101383865-101383865 on Build GRCh38
rs370080358 Chr.2: 101383722-101383722 on Build GRCh38
rs1009686896 Chr.2: 101383943-101383943 on Build GRCh38
rs771725781 Chr.2: 101383672-101383672 on Build GRCh38
rs1056625484 Chr.2: 101383942-101383942 on Build GRCh38
rs181744759 Chr.2: 101383636-101383636 on Build GRCh38
rs545387104 Chr.2: 101383998-101383998 on Build GRCh38
rs928890144 Chr.2: 101383659-101383659 on Build GRCh38
rs533919125 Chr.2: 101383663-101383664 on Build GRCh38
rs988334525 Chr.2: 101383637-101383637 on Build GRCh38
rs545627932 Chr.2: 101383822-101383822 on Build GRCh38
rs946204628 Chr.2: 101383646-101383646 on Build GRCh38
rs139618261 Chr.2: 101383881-101383881 on Build GRCh38
rs997177204 Chr.2: 101383808-101383808 on Build GRCh38
rs779560622 Chr.2: 101384028-101384028 on Build GRCh38
rs574260505 Chr.2: 101383905-101383905 on Build GRCh38
rs1020746537 Chr.2: 101383861-101383861 on Build GRCh38
rs753278288 Chr.2: 101383642-101383642 on Build GRCh38
rs377504420 Chr.2: 101383687-101383687 on Build GRCh38
rs966066829 Chr.2: 101384070-101384070 on Build GRCh38
rs755044821 Chr.2: 101383750-101383750 on Build GRCh38
rs986179160 Chr.2: 101384025-101384025 on Build GRCh38
rs557550012 Chr.2: 101383884-101383884 on Build GRCh38
rs749378522 Chr.2: 101383660-101383660 on Build GRCh38
rs1009671985 Chr.2: 101384089-101384089 on Build GRCh38
rs770539676 Chr.2: 101383690-101383690 on Build GRCh38
rs920186528 Chr.2: 101383908-101383908 on Build GRCh38
rs930801989 Chr.2: 101383879-101383879 on Build GRCh38
rs777647098 Chr.2: 101383686-101383686 on Build GRCh38
rs1047594546 Chr.2: 101383732-101383732 on Build GRCh38
rs768915670 Chr.2: 101384003-101384003 on Build GRCh38
rs1021325627 Chr.2: 101383640-101383640 on Build GRCh38
rs796278396 Chr.2: 101384090-101384090 on Build GRCh38
rs149316087 Chr.2: 101383778-101383778 on Build GRCh38
rs1019478490 Chr.2: 101384029-101384029 on Build GRCh38
rs776322983 Chr.2: 101383695-101383695 on Build GRCh38
rs774679018 Chr.2: 101383716-101383716 on Build GRCh38
rs766698901 Chr.2: 101383736-101383736 on Build GRCh38
rs59472923 Chr.2: 101383979-101383979 on Build GRCh38
rs769493011 Chr.2: 101383857-101383857 on Build GRCh38
rs761184555 Chr.2: 101383734-101383734 on Build GRCh38
rs543224481 Chr.2: 101383936-101383936 on Build GRCh38
rs1019384834 Chr.2: 101383760-101383760 on Build GRCh38
rs762401521 Chr.2: 101383721-101383721 on Build GRCh38
rs919528198 Chr.2: 101383871-101383871 on Build GRCh38
rs944141830 Chr.2: 101383980-101383980 on Build GRCh38
rs745661161 Chr.2: 101383714-101383715 on Build GRCh38
rs755169821 Chr.2: 101383664-101383664 on Build GRCh38
rs996690302 Chr.2: 101384046-101384046 on Build GRCh38
rs987817428 Chr.2: 101383828-101383828 on Build GRCh38
rs972504461 Chr.2: 101384078-101384078 on Build GRCh38
rs957984638 Chr.2: 101383816-101383816 on Build GRCh38
rs758987909 Chr.2: 101383700-101383700 on Build GRCh38
rs565089913 Chr.2: 101384011-101384011 on Build GRCh38
rs560125801 Chr.2: 101383975-101383975 on Build GRCh38
rs1054147754 Chr.2: 101384049-101384049 on Build GRCh38
rs11694978 Chr.2: 101383748-101383748 on Build GRCh38

View Full Product Details