Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130157090-130157588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMPD4
Entrez Gene ID
55627
Gene Name
sphingomyelin phosphodiesterase 4
Gene Aliases
NET13, NSMASE-3, NSMASE3
UniGene
Hs.516450
Gene Chromosome Location
Chr.2: 130151392-130182750 on Build GRCh38
Transcripts
NM_001171083.2 NM_017751.4 NM_017951.4 NR_033230.2 NR_033231.2 NR_033232.2 XM_011511443.2 XM_011511444.2 XM_011511445.2 XM_017004448.1 XM_017004449.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1006468 c.1138G>A p.A380T Chr.2: 130157327-130157327 on Build GRCh38
7391132 c.1214G>T p.R405L Chr.2: 130157251-130157251 on Build GRCh38
715834 c.1092G>T p.E364D Chr.2: 130157373-130157373 on Build GRCh38
715835 c.1093C>A p.H365N Chr.2: 130157372-130157372 on Build GRCh38
715836 c.1132G>T p.A378S Chr.2: 130157333-130157333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35660918 Chr.2: 130157198-130157198 on Build GRCh38
rs576639239 Chr.2: 130157141-130157141 on Build GRCh38
rs35560314 Chr.2: 130157149-130157149 on Build GRCh38
rs760411195 Chr.2: 130157426-130157426 on Build GRCh38
rs116702261 Chr.2: 130157453-130157453 on Build GRCh38
rs190205752 Chr.2: 130157479-130157479 on Build GRCh38
rs367891171 Chr.2: 130157292-130157292 on Build GRCh38
rs777027595 Chr.2: 130157261-130157261 on Build GRCh38
rs1027859587 Chr.2: 130157189-130157189 on Build GRCh38
rs978201115 Chr.2: 130157488-130157488 on Build GRCh38
rs554980866 Chr.2: 130157119-130157119 on Build GRCh38
rs764012615 Chr.2: 130157210-130157212 on Build GRCh38
rs572147978 Chr.2: 130157291-130157291 on Build GRCh38
rs542245390 Chr.2: 130157334-130157334 on Build GRCh38
rs752332358 Chr.2: 130157310-130157310 on Build GRCh38
rs745654934 Chr.2: 130157235-130157235 on Build GRCh38
rs765809356 Chr.2: 130157279-130157279 on Build GRCh38
rs551025551 Chr.2: 130157165-130157165 on Build GRCh38
rs778717941 Chr.2: 130157233-130157233 on Build GRCh38
rs112437022 Chr.2: 130157209-130157209 on Build GRCh38
rs757019055 Chr.2: 130157405-130157407 on Build GRCh38
rs1012544265 Chr.2: 130157385-130157385 on Build GRCh38
rs775721583 Chr.2: 130157251-130157251 on Build GRCh38
rs756697472 Chr.2: 130157432-130157432 on Build GRCh38
rs560969417 Chr.2: 130157378-130157378 on Build GRCh38
rs142393930 Chr.2: 130157464-130157464 on Build GRCh38
rs762116344 Chr.2: 130157202-130157202 on Build GRCh38
rs2290296 Chr.2: 130157224-130157224 on Build GRCh38
rs780990528 Chr.2: 130157213-130157213 on Build GRCh38
rs762431943 Chr.2: 130157270-130157270 on Build GRCh38
rs948035336 Chr.2: 130157551-130157551 on Build GRCh38
rs895024990 Chr.2: 130157167-130157167 on Build GRCh38
rs765649506 Chr.2: 130157205-130157205 on Build GRCh38
rs771788227 Chr.2: 130157361-130157361 on Build GRCh38
rs768740780 Chr.2: 130157260-130157260 on Build GRCh38
rs187574477 Chr.2: 130157431-130157431 on Build GRCh38
rs1003455863 Chr.2: 130157136-130157136 on Build GRCh38
rs760675524 Chr.2: 130157259-130157259 on Build GRCh38
rs769744557 Chr.2: 130157540-130157540 on Build GRCh38
rs529951621 Chr.2: 130157530-130157530 on Build GRCh38
rs774554309 Chr.2: 130157412-130157412 on Build GRCh38
rs772336350 Chr.2: 130157236-130157236 on Build GRCh38
rs755596059 Chr.2: 130157219-130157219 on Build GRCh38
rs10206469 Chr.2: 130157280-130157280 on Build GRCh38
rs923709257 Chr.2: 130157489-130157489 on Build GRCh38
rs531230637 Chr.2: 130157392-130157392 on Build GRCh38
rs773800582 Chr.2: 130157394-130157394 on Build GRCh38
rs916224020 Chr.2: 130157533-130157533 on Build GRCh38
rs1015641753 Chr.2: 130157214-130157214 on Build GRCh38
rs747205901 Chr.2: 130157375-130157375 on Build GRCh38
rs749196653 Chr.2: 130157227-130157227 on Build GRCh38
rs968170837 Chr.2: 130157481-130157481 on Build GRCh38
rs887604402 Chr.2: 130157407-130157407 on Build GRCh38
rs766729229 Chr.2: 130157402-130157402 on Build GRCh38
rs750797826 Chr.2: 130157210-130157210 on Build GRCh38
rs764950953 Chr.2: 130157419-130157419 on Build GRCh38
rs778807928 Chr.2: 130157348-130157348 on Build GRCh38
rs202201383 Chr.2: 130157386-130157386 on Build GRCh38
rs1056139273 Chr.2: 130157526-130157526 on Build GRCh38
rs564845526 Chr.2: 130157445-130157445 on Build GRCh38
rs540082240 Chr.2: 130157214-130157215 on Build GRCh38
rs765162154 Chr.2: 130157435-130157435 on Build GRCh38
rs537719298 Chr.2: 130157172-130157172 on Build GRCh38
rs770829604 Chr.2: 130157230-130157230 on Build GRCh38
rs189395415 Chr.2: 130157179-130157179 on Build GRCh38
rs985652259 Chr.2: 130157241-130157241 on Build GRCh38
rs748235097 Chr.2: 130157391-130157391 on Build GRCh38
rs180747214 Chr.2: 130157188-130157188 on Build GRCh38
rs201383210 Chr.2: 130157140-130157144 on Build GRCh38
rs939062180 Chr.2: 130157507-130157507 on Build GRCh38
rs895063402 Chr.2: 130157384-130157384 on Build GRCh38
rs541644496 Chr.2: 130157272-130157272 on Build GRCh38
rs986110252 Chr.2: 130157471-130157471 on Build GRCh38
rs745675076 Chr.2: 130157359-130157359 on Build GRCh38
rs1030037244 Chr.2: 130157454-130157454 on Build GRCh38
rs755615020 Chr.2: 130157316-130157316 on Build GRCh38
rs530238773 Chr.2: 130157500-130157500 on Build GRCh38
rs752209449 Chr.2: 130157218-130157218 on Build GRCh38
rs758211544 Chr.2: 130157448-130157448 on Build GRCh38
rs780170230 Chr.2: 130157370-130157370 on Build GRCh38
rs1056031261 Chr.2: 130157356-130157356 on Build GRCh38
rs371216093 Chr.2: 130157180-130157180 on Build GRCh38
rs41269837 Chr.2: 130157331-130157331 on Build GRCh38
rs763420025 Chr.2: 130157396-130157396 on Build GRCh38

View Full Product Details