Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 147933315-147933823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ORC4
Entrez Gene ID
5000
Gene Name
origin recognition complex subunit 4
Gene Aliases
ORC4L, ORC4P
UniGene
Hs.558364
Gene Chromosome Location
Chr.2: 147930397-148021604 on Build GRCh38
Transcripts
NM_001190879.2 NM_001190881.2 NM_001190882.2 NM_002552.4 NM_181741.3 NM_181742.3

SNP Detail

SNP ID Chromosome Location
rs925641744 Chr.2: 147933430-147933430 on Build GRCh38
rs1038469834 Chr.2: 147933793-147933793 on Build GRCh38
rs978485520 Chr.2: 147933628-147933628 on Build GRCh38
rs187377554 Chr.2: 147933524-147933524 on Build GRCh38
rs912969335 Chr.2: 147933376-147933376 on Build GRCh38
rs375155777 Chr.2: 147933532-147933532 on Build GRCh38
rs901461526 Chr.2: 147933457-147933457 on Build GRCh38
rs17232078 Chr.2: 147933687-147933687 on Build GRCh38
rs763779367 Chr.2: 147933477-147933477 on Build GRCh38
rs533815198 Chr.2: 147933588-147933588 on Build GRCh38
rs764522068 Chr.2: 147933389-147933391 on Build GRCh38
rs967509291 Chr.2: 147933622-147933622 on Build GRCh38
rs939476452 Chr.2: 147933794-147933794 on Build GRCh38
rs942916810 Chr.2: 147933432-147933432 on Build GRCh38
rs17232064 Chr.2: 147933776-147933776 on Build GRCh38
rs925587947 Chr.2: 147933667-147933667 on Build GRCh38
rs887179864 Chr.2: 147933510-147933510 on Build GRCh38
rs138015010 Chr.2: 147933679-147933679 on Build GRCh38
rs1034259636 Chr.2: 147933564-147933564 on Build GRCh38
rs538638237 Chr.2: 147933400-147933400 on Build GRCh38
rs1047647053 Chr.2: 147933750-147933750 on Build GRCh38
rs567338886 Chr.2: 147933457-147933458 on Build GRCh38
rs961998835 Chr.2: 147933657-147933657 on Build GRCh38
rs146923314 Chr.2: 147933546-147933546 on Build GRCh38
rs892721961 Chr.2: 147933496-147933496 on Build GRCh38
rs1036045406 Chr.2: 147933444-147933444 on Build GRCh38
rs1047251487 Chr.2: 147933482-147933482 on Build GRCh38
rs920315421 Chr.2: 147933451-147933455 on Build GRCh38
rs760246585 Chr.2: 147933340-147933340 on Build GRCh38
rs569246843 Chr.2: 147933481-147933481 on Build GRCh38
rs556209839 Chr.2: 147933573-147933573 on Build GRCh38
rs535754607 Chr.2: 147933345-147933345 on Build GRCh38
rs1039940743 Chr.2: 147933454-147933454 on Build GRCh38
rs550991254 Chr.2: 147933466-147933466 on Build GRCh38
rs1009961239 Chr.2: 147933501-147933501 on Build GRCh38
rs17232071 Chr.2: 147933728-147933728 on Build GRCh38
rs190664746 Chr.2: 147933621-147933621 on Build GRCh38
rs945839505 Chr.2: 147933388-147933388 on Build GRCh38
rs867050981 Chr.2: 147933638-147933638 on Build GRCh38
rs996960321 Chr.2: 147933547-147933547 on Build GRCh38
rs1016920761 Chr.2: 147933528-147933528 on Build GRCh38
rs942363040 Chr.2: 147933673-147933673 on Build GRCh38
rs922829854 Chr.2: 147933716-147933716 on Build GRCh38
rs941383578 Chr.2: 147933784-147933784 on Build GRCh38
rs930423777 Chr.2: 147933452-147933452 on Build GRCh38

View Full Product Details