Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 148770904-148771404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPC2
Entrez Gene ID
26122
Gene Name
enhancer of polycomb homolog 2
Gene Aliases
EPC-LIKE
UniGene
Hs.23270
Gene Chromosome Location
Chr.2: 148644780-148788491 on Build GRCh38
Transcripts
NM_015630.3 XM_011510941.2 XM_011510943.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
221192 c.1379T>C p.V460A Chr.2: 148771046-148771046 on Build GRCh38
4435757 c.1573C>G p.L525V Chr.2: 148771240-148771240 on Build GRCh38
7186813 c.1361_1362insT p.G455fs*16 Chr.2: 148770922-148770923 on Build GRCh38
274985 c.1693T>G p.L565V Chr.2: 148771360-148771360 on Build GRCh38
7322009 c.1546A>G p.I516V Chr.2: 148771213-148771213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201098073 Chr.2: 148771138-148771138 on Build GRCh38
rs754420639 Chr.2: 148771084-148771084 on Build GRCh38
rs199930117 Chr.2: 148771366-148771366 on Build GRCh38
rs185289361 Chr.2: 148771148-148771148 on Build GRCh38
rs961422592 Chr.2: 148771168-148771168 on Build GRCh38
rs765474517 Chr.2: 148771334-148771334 on Build GRCh38
rs952721990 Chr.2: 148771198-148771198 on Build GRCh38
rs773161874 Chr.2: 148771058-148771058 on Build GRCh38
rs778439385 Chr.2: 148770986-148770986 on Build GRCh38
rs767184969 Chr.2: 148771066-148771066 on Build GRCh38
rs375339855 Chr.2: 148771106-148771106 on Build GRCh38
rs34219412 Chr.2: 148771379-148771380 on Build GRCh38
rs143193226 Chr.2: 148770942-148770942 on Build GRCh38
rs746129367 Chr.2: 148771012-148771012 on Build GRCh38
rs1050368495 Chr.2: 148771077-148771077 on Build GRCh38
rs982977480 Chr.2: 148770997-148770997 on Build GRCh38
rs776361255 Chr.2: 148771019-148771019 on Build GRCh38
rs780057245 Chr.2: 148771233-148771233 on Build GRCh38
rs540745251 Chr.2: 148771024-148771024 on Build GRCh38
rs764605539 Chr.2: 148771182-148771182 on Build GRCh38
rs760356407 Chr.2: 148771177-148771177 on Build GRCh38
rs771636079 Chr.2: 148771298-148771298 on Build GRCh38
rs151138439 Chr.2: 148770975-148770975 on Build GRCh38
rs369459576 Chr.2: 148771294-148771294 on Build GRCh38
rs934461326 Chr.2: 148771162-148771162 on Build GRCh38
rs762495682 Chr.2: 148771139-148771139 on Build GRCh38
rs771470479 Chr.2: 148770941-148770941 on Build GRCh38
rs758805629 Chr.2: 148771362-148771362 on Build GRCh38
rs775119282 Chr.2: 148771299-148771299 on Build GRCh38
rs772310625 Chr.2: 148771018-148771018 on Build GRCh38
rs747278661 Chr.2: 148771238-148771238 on Build GRCh38
rs761606141 Chr.2: 148771028-148771028 on Build GRCh38
rs755360890 Chr.2: 148770963-148770966 on Build GRCh38
rs915608094 Chr.2: 148771067-148771067 on Build GRCh38
rs777611033 Chr.2: 148771260-148771260 on Build GRCh38
rs774350069 Chr.2: 148771271-148771271 on Build GRCh38
rs769508718 Chr.2: 148771031-148771031 on Build GRCh38
rs374201174 Chr.2: 148771344-148771344 on Build GRCh38
rs781588367 Chr.2: 148771112-148771113 on Build GRCh38
rs751993337 Chr.2: 148770960-148770961 on Build GRCh38
rs201024923 Chr.2: 148770998-148770998 on Build GRCh38
rs765870754 Chr.2: 148770954-148770954 on Build GRCh38
rs760446331 Chr.2: 148771310-148771310 on Build GRCh38
rs775315565 Chr.2: 148771175-148771175 on Build GRCh38
rs369986379 Chr.2: 148771129-148771129 on Build GRCh38
rs756553008 Chr.2: 148771075-148771075 on Build GRCh38
rs779005072 Chr.2: 148771003-148771003 on Build GRCh38
rs368220537 Chr.2: 148770973-148770973 on Build GRCh38
rs762256199 Chr.2: 148771330-148771330 on Build GRCh38
rs751048807 Chr.2: 148771060-148771060 on Build GRCh38
rs762450919 Chr.2: 148770952-148770952 on Build GRCh38
rs745753492 Chr.2: 148771286-148771286 on Build GRCh38
rs183800694 Chr.2: 148771328-148771328 on Build GRCh38
rs200771222 Chr.2: 148771197-148771197 on Build GRCh38
rs765764852 Chr.2: 148771219-148771219 on Build GRCh38
rs758611059 Chr.2: 148771120-148771120 on Build GRCh38
rs750824217 Chr.2: 148771221-148771221 on Build GRCh38
rs565102580 Chr.2: 148771107-148771107 on Build GRCh38
rs752339749 Chr.2: 148771073-148771073 on Build GRCh38
rs763837264 Chr.2: 148771319-148771319 on Build GRCh38
rs191190211 Chr.2: 148771269-148771269 on Build GRCh38
rs796970452 Chr.2: 148771321-148771321 on Build GRCh38
rs780323319 Chr.2: 148771125-148771125 on Build GRCh38
rs749094942 Chr.2: 148771266-148771266 on Build GRCh38
rs754635475 Chr.2: 148770961-148770961 on Build GRCh38
rs778245242 Chr.2: 148771079-148771079 on Build GRCh38
rs375081989 Chr.2: 148771163-148771163 on Build GRCh38
rs771168518 Chr.2: 148771000-148771000 on Build GRCh38
rs949778821 Chr.2: 148771112-148771112 on Build GRCh38
rs1026911525 Chr.2: 148771184-148771184 on Build GRCh38
rs758532643 Chr.2: 148771224-148771224 on Build GRCh38
rs754226470 Chr.2: 148771188-148771188 on Build GRCh38
rs991438246 Chr.2: 148771202-148771202 on Build GRCh38
rs34444151 Chr.2: 148771169-148771169 on Build GRCh38
rs756742878 Chr.2: 148770982-148770982 on Build GRCh38
rs765854975 Chr.2: 148771059-148771059 on Build GRCh38
rs770896107 Chr.2: 148771156-148771156 on Build GRCh38
rs751190643 Chr.2: 148770956-148770956 on Build GRCh38
rs914871885 Chr.2: 148771295-148771295 on Build GRCh38
rs372741400 Chr.2: 148771242-148771242 on Build GRCh38
rs757763407 Chr.2: 148771099-148771099 on Build GRCh38

View Full Product Details