Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 151485590-151486032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEB
Entrez Gene ID
4703
Gene Name
nebulin
Gene Aliases
NEB177D, NEM2
UniGene
Hs.588655
Gene Chromosome Location
Chr.2: 151485334-151734487 on Build GRCh38
Transcripts
NM_001164507.1 NM_001164508.1 NM_001271208.1 NM_004543.4 XM_005246590.2 XM_005246591.2 XM_005246592.2 XM_005246593.2 XM_005246594.2 XM_005246596.2 XM_005246597.2 XM_005246598.2 XM_005246599.2 XM_005246600.2 XM_005246601.2 XM_005246602.2 XM_005246603.2 XM_005246604.2 XM_005246606.2 XM_005246608.2 XM_005246610.2 XM_005246611.2 XM_005246612.2 XM_005246613.2 XM_005246615.2 XM_005246617.2 XM_006712541.2 XM_006712542.2 XM_011511225.2 XM_011511226.2 XM_011511227.2 XM_017004177.1 XM_017004178.1 XM_017004179.1 XM_017004180.1 XM_017004181.1 XM_017004182.1 XM_017004183.1 XM_017004184.1 XM_017004185.1

Gene Symbol
RIF1
Entrez Gene ID
55183
Gene Name
replication timing regulatory factor 1
Gene Aliases
-
UniGene
Hs.536537
Gene Chromosome Location
Chr.2: 151409883-151534200 on Build GRCh38
Transcripts
NM_001164507.1 NM_001164508.1 NM_001271208.1 NM_004543.4 XM_005246590.2 XM_005246591.2 XM_005246592.2 XM_005246593.2 XM_005246594.2 XM_005246596.2 XM_005246597.2 XM_005246598.2 XM_005246599.2 XM_005246600.2 XM_005246601.2 XM_005246602.2 XM_005246603.2 XM_005246604.2 XM_005246606.2 XM_005246608.2 XM_005246610.2 XM_005246611.2 XM_005246612.2 XM_005246613.2 XM_005246615.2 XM_005246617.2 XM_006712541.2 XM_006712542.2 XM_011511225.2 XM_011511226.2 XM_011511227.2 XM_017004177.1 XM_017004178.1 XM_017004179.1 XM_017004180.1 XM_017004181.1 XM_017004182.1 XM_017004183.1 XM_017004184.1 XM_017004185.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7099848 c.19911C>G p.I6637M Chr.2: 151485859-151485859 on Build GRCh38
206373 c.19847G>A p.R6616H Chr.2: 151485923-151485923 on Build GRCh38
4706169 c.19949G>A p.G6650D Chr.2: 151485821-151485821 on Build GRCh38
6724090 c.25543G>A p.G8515R Chr.2: 151485795-151485795 on Build GRCh38
4706172 c.19942A>G p.M6648V Chr.2: 151485828-151485828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747518320 Chr.2: 151485969-151485969 on Build GRCh38
rs774603877 Chr.2: 151485893-151485893 on Build GRCh38
rs746779495 Chr.2: 151485961-151485963 on Build GRCh38
rs745444923 Chr.2: 151485870-151485870 on Build GRCh38
rs777522469 Chr.2: 151485757-151485757 on Build GRCh38
rs755934851 Chr.2: 151485946-151485946 on Build GRCh38
rs779805343 Chr.2: 151485741-151485741 on Build GRCh38
rs767629927 Chr.2: 151485805-151485805 on Build GRCh38
rs982464769 Chr.2: 151486007-151486007 on Build GRCh38
rs749003938 Chr.2: 151485823-151485823 on Build GRCh38
rs143932500 Chr.2: 151485962-151485962 on Build GRCh38
rs993833092 Chr.2: 151485891-151485891 on Build GRCh38
rs757923442 Chr.2: 151485964-151485964 on Build GRCh38
rs917471345 Chr.2: 151485983-151485983 on Build GRCh38
rs746382045 Chr.2: 151485775-151485775 on Build GRCh38
rs955292497 Chr.2: 151485658-151485658 on Build GRCh38
rs941040073 Chr.2: 151485683-151485683 on Build GRCh38
rs775823432 Chr.2: 151485783-151485783 on Build GRCh38
rs368167929 Chr.2: 151485809-151485809 on Build GRCh38
rs759841766 Chr.2: 151485729-151485730 on Build GRCh38
rs778502509 Chr.2: 151485868-151485868 on Build GRCh38
rs922490072 Chr.2: 151485633-151485633 on Build GRCh38
rs762107118 Chr.2: 151485913-151485913 on Build GRCh38
rs542325814 Chr.2: 151485752-151485752 on Build GRCh38
rs148839798 Chr.2: 151485949-151485950 on Build GRCh38
rs752707527 Chr.2: 151485943-151485943 on Build GRCh38
rs776645459 Chr.2: 151486004-151486004 on Build GRCh38
rs748407608 Chr.2: 151486002-151486002 on Build GRCh38
rs776932788 Chr.2: 151485795-151485795 on Build GRCh38
rs955699252 Chr.2: 151485972-151485972 on Build GRCh38
rs776650831 Chr.2: 151485906-151485906 on Build GRCh38
rs908181333 Chr.2: 151485669-151485669 on Build GRCh38
rs558019875 Chr.2: 151485773-151485773 on Build GRCh38
rs756024407 Chr.2: 151485816-151485816 on Build GRCh38
rs917418756 Chr.2: 151485742-151485742 on Build GRCh38
rs796213500 Chr.2: 151485965-151485965 on Build GRCh38
rs778467089 Chr.2: 151485963-151485963 on Build GRCh38
rs374505298 Chr.2: 151485722-151485722 on Build GRCh38
rs924281589 Chr.2: 151485717-151485717 on Build GRCh38
rs370369451 Chr.2: 151485792-151485792 on Build GRCh38
rs542706227 Chr.2: 151485924-151485924 on Build GRCh38
rs1051639339 Chr.2: 151485909-151485909 on Build GRCh38
rs754751835 Chr.2: 151485743-151485743 on Build GRCh38
rs751394716 Chr.2: 151485804-151485804 on Build GRCh38
rs549826996 Chr.2: 151485617-151485617 on Build GRCh38
rs1030206780 Chr.2: 151485631-151485631 on Build GRCh38
rs761066364 Chr.2: 151485789-151485789 on Build GRCh38
rs3821324 Chr.2: 151485796-151485796 on Build GRCh38
rs753828020 Chr.2: 151485710-151485710 on Build GRCh38
rs747323786 Chr.2: 151485885-151485885 on Build GRCh38
rs535169903 Chr.2: 151485687-151485687 on Build GRCh38
rs371591500 Chr.2: 151485916-151485916 on Build GRCh38
rs772621475 Chr.2: 151485778-151485778 on Build GRCh38
rs141449286 Chr.2: 151485905-151485905 on Build GRCh38
rs781148078 Chr.2: 151485748-151485748 on Build GRCh38
rs893112731 Chr.2: 151485881-151485881 on Build GRCh38
rs777246173 Chr.2: 151485819-151485819 on Build GRCh38
rs143994653 Chr.2: 151485661-151485661 on Build GRCh38
rs765363580 Chr.2: 151485798-151485798 on Build GRCh38
rs191006696 Chr.2: 151485694-151485694 on Build GRCh38
rs374412508 Chr.2: 151485917-151485917 on Build GRCh38
rs376501860 Chr.2: 151485724-151485725 on Build GRCh38
rs756659726 Chr.2: 151485829-151485829 on Build GRCh38
rs566096042 Chr.2: 151485703-151485703 on Build GRCh38
rs117861109 Chr.2: 151485774-151485774 on Build GRCh38
rs377343559 Chr.2: 151485753-151485753 on Build GRCh38
rs985877971 Chr.2: 151485668-151485668 on Build GRCh38
rs368346105 Chr.2: 151485923-151485923 on Build GRCh38
rs772533651 Chr.2: 151485871-151485871 on Build GRCh38
rs559804968 Chr.2: 151485932-151485932 on Build GRCh38
rs773628949 Chr.2: 151485724-151485724 on Build GRCh38
rs534898543 Chr.2: 151485749-151485749 on Build GRCh38
rs753378567 Chr.2: 151485955-151485955 on Build GRCh38
rs768045509 Chr.2: 151485929-151485930 on Build GRCh38
rs371503160 Chr.2: 151485973-151485973 on Build GRCh38
rs16830090 Chr.2: 151485967-151485967 on Build GRCh38
rs201714437 Chr.2: 151485875-151485875 on Build GRCh38
rs763959138 Chr.2: 151485954-151485954 on Build GRCh38
rs372113031 Chr.2: 151485940-151485940 on Build GRCh38
rs970272286 Chr.2: 151485986-151485986 on Build GRCh38
rs748728868 Chr.2: 151485759-151485759 on Build GRCh38

View Full Product Details