Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 151489733-151490243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEB
Entrez Gene ID
4703
Gene Name
nebulin
Gene Aliases
NEB177D, NEM2
UniGene
Hs.588655
Gene Chromosome Location
Chr.2: 151485334-151734487 on Build GRCh38
Transcripts
NM_001164507.1 NM_001164508.1 NM_001271208.1 NM_004543.4 XM_005246590.2 XM_005246591.2 XM_005246592.2 XM_005246593.2 XM_005246594.2 XM_005246596.2 XM_005246597.2 XM_005246598.2 XM_005246599.2 XM_005246600.2 XM_005246601.2 XM_005246602.2 XM_005246603.2 XM_005246604.2 XM_005246606.2 XM_005246608.2 XM_005246610.2 XM_005246611.2 XM_005246612.2 XM_005246613.2 XM_005246615.2 XM_005246617.2 XM_006712541.2 XM_006712542.2 XM_011511225.2 XM_011511226.2 XM_011511227.2 XM_017004177.1 XM_017004178.1 XM_017004179.1 XM_017004180.1 XM_017004181.1 XM_017004182.1 XM_017004183.1 XM_017004184.1 XM_017004185.1

Gene Symbol
RIF1
Entrez Gene ID
55183
Gene Name
replication timing regulatory factor 1
Gene Aliases
-
UniGene
Hs.536537
Gene Chromosome Location
Chr.2: 151409883-151534200 on Build GRCh38
Transcripts
NM_001164507.1 NM_001164508.1 NM_001271208.1 NM_004543.4 XM_005246590.2 XM_005246591.2 XM_005246592.2 XM_005246593.2 XM_005246594.2 XM_005246596.2 XM_005246597.2 XM_005246598.2 XM_005246599.2 XM_005246600.2 XM_005246601.2 XM_005246602.2 XM_005246603.2 XM_005246604.2 XM_005246606.2 XM_005246608.2 XM_005246610.2 XM_005246611.2 XM_005246612.2 XM_005246613.2 XM_005246615.2 XM_005246617.2 XM_006712541.2 XM_006712542.2 XM_011511225.2 XM_011511226.2 XM_011511227.2 XM_017004177.1 XM_017004178.1 XM_017004179.1 XM_017004180.1 XM_017004181.1 XM_017004182.1 XM_017004183.1 XM_017004184.1 XM_017004185.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1316658 c.25400C>A p.A8467D Chr.2: 151489975-151489975 on Build GRCh38
1590778 c.25370T>C p.V8457A Chr.2: 151490005-151490005 on Build GRCh38
1590777 c.25336C>T p.R8446* Chr.2: 151490039-151490039 on Build GRCh38
7202301 c.25334A>G p.Q8445R Chr.2: 151490041-151490041 on Build GRCh38
5892744 c.25337G>A p.R8446Q Chr.2: 151490038-151490038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779434256 Chr.2: 151490187-151490187 on Build GRCh38
rs781606419 Chr.2: 151489921-151489921 on Build GRCh38
rs376082951 Chr.2: 151489925-151489925 on Build GRCh38
rs78592085 Chr.2: 151490008-151490008 on Build GRCh38
rs1042096421 Chr.2: 151489906-151489906 on Build GRCh38
rs372813378 Chr.2: 151489757-151489758 on Build GRCh38
rs944667720 Chr.2: 151489856-151489856 on Build GRCh38
rs999772907 Chr.2: 151490216-151490216 on Build GRCh38
rs894867682 Chr.2: 151490180-151490181 on Build GRCh38
rs765019089 Chr.2: 151490102-151490102 on Build GRCh38
rs954022747 Chr.2: 151490138-151490138 on Build GRCh38
rs894795433 Chr.2: 151489870-151489870 on Build GRCh38
rs117632493 Chr.2: 151490199-151490199 on Build GRCh38
rs902218572 Chr.2: 151490214-151490214 on Build GRCh38
rs771080616 Chr.2: 151490059-151490059 on Build GRCh38
rs776453091 Chr.2: 151490091-151490091 on Build GRCh38
rs762580492 Chr.2: 151490123-151490123 on Build GRCh38
rs757290966 Chr.2: 151490080-151490081 on Build GRCh38
rs377748897 Chr.2: 151490016-151490016 on Build GRCh38
rs1027833218 Chr.2: 151490013-151490013 on Build GRCh38
rs552439882 Chr.2: 151490090-151490090 on Build GRCh38
rs766255786 Chr.2: 151490097-151490100 on Build GRCh38
rs941882391 Chr.2: 151490126-151490126 on Build GRCh38
rs778104284 Chr.2: 151490045-151490045 on Build GRCh38
rs752994973 Chr.2: 151489823-151489823 on Build GRCh38
rs765995672 Chr.2: 151489839-151489839 on Build GRCh38
rs560634136 Chr.2: 151489758-151489758 on Build GRCh38
rs201934608 Chr.2: 151490093-151490093 on Build GRCh38
rs1013234237 Chr.2: 151489879-151489880 on Build GRCh38
rs569107587 Chr.2: 151490099-151490099 on Build GRCh38
rs761049503 Chr.2: 151489947-151489948 on Build GRCh38
rs946524084 Chr.2: 151490172-151490172 on Build GRCh38
rs376984481 Chr.2: 151489985-151489985 on Build GRCh38
rs34415027 Chr.2: 151490092-151490092 on Build GRCh38
rs771168563 Chr.2: 151489962-151489962 on Build GRCh38
rs965449402 Chr.2: 151490031-151490031 on Build GRCh38
rs749529584 Chr.2: 151490057-151490057 on Build GRCh38
rs750018272 Chr.2: 151490117-151490117 on Build GRCh38
rs902200227 Chr.2: 151489922-151489922 on Build GRCh38
rs923714189 Chr.2: 151490180-151490180 on Build GRCh38
rs921926775 Chr.2: 151490112-151490112 on Build GRCh38
rs774563344 Chr.2: 151490075-151490075 on Build GRCh38
rs958107239 Chr.2: 151489758-151489758 on Build GRCh38
rs753976426 Chr.2: 151490075-151490085 on Build GRCh38
rs763079125 Chr.2: 151489961-151489961 on Build GRCh38
rs566651804 Chr.2: 151489840-151489841 on Build GRCh38
rs773618785 Chr.2: 151489938-151489938 on Build GRCh38
rs764892870 Chr.2: 151490015-151490015 on Build GRCh38
rs546478649 Chr.2: 151489853-151489853 on Build GRCh38
rs13031275 Chr.2: 151489980-151489980 on Build GRCh38
rs781590882 Chr.2: 151490040-151490040 on Build GRCh38
rs745856699 Chr.2: 151489941-151489944 on Build GRCh38
rs751462080 Chr.2: 151490117-151490117 on Build GRCh38
rs764403209 Chr.2: 151490032-151490034 on Build GRCh38
rs778955728 Chr.2: 151489804-151489804 on Build GRCh38
rs562192701 Chr.2: 151489896-151489896 on Build GRCh38
rs200731870 Chr.2: 151490039-151490039 on Build GRCh38
rs748525509 Chr.2: 151490043-151490043 on Build GRCh38
rs1042189022 Chr.2: 151490178-151490178 on Build GRCh38
rs190063063 Chr.2: 151489890-151489890 on Build GRCh38
rs952273879 Chr.2: 151490014-151490014 on Build GRCh38
rs145934392 Chr.2: 151489860-151489860 on Build GRCh38
rs369420017 Chr.2: 151489926-151489926 on Build GRCh38
rs182699929 Chr.2: 151490023-151490023 on Build GRCh38
rs985986221 Chr.2: 151489758-151489761 on Build GRCh38
rs918592168 Chr.2: 151490082-151490082 on Build GRCh38
rs6733688 Chr.2: 151489887-151489887 on Build GRCh38
rs966105402 Chr.2: 151490067-151490067 on Build GRCh38
rs548319430 Chr.2: 151489889-151489889 on Build GRCh38
rs200083849 Chr.2: 151490007-151490007 on Build GRCh38
rs985741163 Chr.2: 151490155-151490155 on Build GRCh38
rs976742353 Chr.2: 151489807-151489810 on Build GRCh38
rs929234690 Chr.2: 151489851-151489851 on Build GRCh38
rs944068256 Chr.2: 151489800-151489800 on Build GRCh38
rs751128054 Chr.2: 151490038-151490038 on Build GRCh38
rs1055313130 Chr.2: 151489869-151489869 on Build GRCh38
rs372703032 Chr.2: 151490077-151490077 on Build GRCh38
rs779552151 Chr.2: 151490028-151490028 on Build GRCh38
rs915096660 Chr.2: 151490157-151490157 on Build GRCh38
rs1003190406 Chr.2: 151489973-151489973 on Build GRCh38
rs186335817 Chr.2: 151489834-151489834 on Build GRCh38
rs922575062 Chr.2: 151489876-151489876 on Build GRCh38
rs929224551 Chr.2: 151490096-151490096 on Build GRCh38

View Full Product Details