Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 156328325-156328825 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR4A2
Entrez Gene ID
4929
Gene Name
nuclear receptor subfamily 4 group A member 2
Gene Aliases
HZF-3, NOT, NURR1, RNR1, TINUR
UniGene
Hs.563344
Gene Chromosome Location
Chr.2: 156324432-156334683 on Build GRCh38
Transcripts
NM_006186.3 XM_005246621.3 XM_005246622.3 XM_006712553.3 XM_011511246.1 XM_017004219.1 XM_017004220.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
162987 c.865C>T p.R289C Chr.2: 156328533-156328533 on Build GRCh38
5058164 c.911A>G p.N304S Chr.2: 156328487-156328487 on Build GRCh38
3364318 c.877A>C p.K293Q Chr.2: 156328521-156328521 on Build GRCh38
4907598 c.903A>C p.T301T Chr.2: 156328528-156328528 on Build GRCh38
6383980 c.866G>T p.R289L Chr.2: 156328532-156328532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985788126 Chr.2: 156328774-156328774 on Build GRCh38
rs779599905 Chr.2: 156328428-156328428 on Build GRCh38
rs1057728 Chr.2: 156328692-156328692 on Build GRCh38
rs960908237 Chr.2: 156328528-156328528 on Build GRCh38
rs770975527 Chr.2: 156328516-156328516 on Build GRCh38
rs200098653 Chr.2: 156328375-156328375 on Build GRCh38
rs756924048 Chr.2: 156328388-156328388 on Build GRCh38
rs369100519 Chr.2: 156328525-156328525 on Build GRCh38
rs553759782 Chr.2: 156328395-156328395 on Build GRCh38
rs951502114 Chr.2: 156328650-156328650 on Build GRCh38
rs868815167 Chr.2: 156328645-156328645 on Build GRCh38
rs375842303 Chr.2: 156328369-156328369 on Build GRCh38
rs568819333 Chr.2: 156328777-156328777 on Build GRCh38
rs997673473 Chr.2: 156328702-156328702 on Build GRCh38
rs935367101 Chr.2: 156328380-156328380 on Build GRCh38
rs746679984 Chr.2: 156328375-156328376 on Build GRCh38
rs747516804 Chr.2: 156328444-156328444 on Build GRCh38
rs887667280 Chr.2: 156328556-156328556 on Build GRCh38
rs545781307 Chr.2: 156328426-156328426 on Build GRCh38
rs769315364 Chr.2: 156328563-156328586 on Build GRCh38
rs531491822 Chr.2: 156328787-156328787 on Build GRCh38
rs912569438 Chr.2: 156328655-156328655 on Build GRCh38
rs544832163 Chr.2: 156328543-156328543 on Build GRCh38
rs753589011 Chr.2: 156328376-156328376 on Build GRCh38
rs773502234 Chr.2: 156328506-156328506 on Build GRCh38
rs764433817 Chr.2: 156328374-156328374 on Build GRCh38
rs1057727 Chr.2: 156328696-156328696 on Build GRCh38
rs757737364 Chr.2: 156328769-156328769 on Build GRCh38
rs3208770 Chr.2: 156328701-156328701 on Build GRCh38
rs35204829 Chr.2: 156328361-156328361 on Build GRCh38
rs369518364 Chr.2: 156328552-156328552 on Build GRCh38
rs1011299537 Chr.2: 156328424-156328424 on Build GRCh38
rs565929368 Chr.2: 156328649-156328649 on Build GRCh38
rs770158698 Chr.2: 156328497-156328497 on Build GRCh38
rs532389053 Chr.2: 156328742-156328742 on Build GRCh38
rs538276241 Chr.2: 156328640-156328640 on Build GRCh38
rs16840253 Chr.2: 156328548-156328548 on Build GRCh38
rs530263388 Chr.2: 156328565-156328565 on Build GRCh38
rs974292407 Chr.2: 156328567-156328567 on Build GRCh38
rs748478277 Chr.2: 156328495-156328495 on Build GRCh38
rs982745590 Chr.2: 156328651-156328651 on Build GRCh38
rs768136480 Chr.2: 156328553-156328560 on Build GRCh38
rs558480553 Chr.2: 156328649-156328650 on Build GRCh38
rs780777042 Chr.2: 156328561-156328562 on Build GRCh38
rs752644905 Chr.2: 156328366-156328366 on Build GRCh38
rs576748818 Chr.2: 156328400-156328400 on Build GRCh38
rs1030106098 Chr.2: 156328722-156328722 on Build GRCh38
rs776990744 Chr.2: 156328638-156328641 on Build GRCh38
rs552104132 Chr.2: 156328762-156328762 on Build GRCh38
rs560598161 Chr.2: 156328735-156328735 on Build GRCh38
rs765008816 Chr.2: 156328561-156328561 on Build GRCh38
rs201103933 Chr.2: 156328483-156328483 on Build GRCh38
rs540352550 Chr.2: 156328594-156328594 on Build GRCh38
rs575964818 Chr.2: 156328513-156328513 on Build GRCh38
rs769178148 Chr.2: 156328464-156328464 on Build GRCh38
rs145436985 Chr.2: 156328545-156328545 on Build GRCh38

View Full Product Details