Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 165293717-165294219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN2A
Entrez Gene ID
6326
Gene Name
sodium voltage-gated channel alpha subunit 2
Gene Aliases
BFIC3, BFIS3, BFNIS, EIEE11, HBA, HBSCI, HBSCII, NAC2, Na(v)1.2, Nav1.2, SCN2A1, SCN2A2
UniGene
Hs.93485
Gene Chromosome Location
Chr.2: 165208056-165392310 on Build GRCh38
Transcripts
NM_001040142.1 NM_021007.2 XM_005246753.3 XM_017004655.1 XM_017004656.1 XM_017004657.1 XM_017004658.1

SNP Detail

SNP ID Chromosome Location
rs780674346 Chr.2: 165294040-165294041 on Build GRCh38
rs949601263 Chr.2: 165293806-165293806 on Build GRCh38
rs969988732 Chr.2: 165293837-165293837 on Build GRCh38
rs886054998 Chr.2: 165294044-165294044 on Build GRCh38
rs867379149 Chr.2: 165294015-165294015 on Build GRCh38
rs886054997 Chr.2: 165294053-165294054 on Build GRCh38
rs1022876772 Chr.2: 165293814-165293814 on Build GRCh38
rs936466321 Chr.2: 165293869-165293869 on Build GRCh38
rs1035560339 Chr.2: 165293889-165293889 on Build GRCh38
rs886054994 Chr.2: 165294039-165294039 on Build GRCh38
rs1011472959 Chr.2: 165293796-165293796 on Build GRCh38
rs746854009 Chr.2: 165293860-165293860 on Build GRCh38
rs886054996 Chr.2: 165294041-165294042 on Build GRCh38
rs67417831 Chr.2: 165294038-165294038 on Build GRCh38
rs886054989 Chr.2: 165293859-165293859 on Build GRCh38
rs961039104 Chr.2: 165294044-165294044 on Build GRCh38
rs974072748 Chr.2: 165294184-165294184 on Build GRCh38
rs568737155 Chr.2: 165293861-165293861 on Build GRCh38
rs940815041 Chr.2: 165293773-165293773 on Build GRCh38
rs886054995 Chr.2: 165294039-165294039 on Build GRCh38
rs972360748 Chr.2: 165293783-165293783 on Build GRCh38
rs12614399 Chr.2: 165293790-165293790 on Build GRCh38
rs76193610 Chr.2: 165294040-165294040 on Build GRCh38
rs758142083 Chr.2: 165293760-165293760 on Build GRCh38
rs886054990 Chr.2: 165294038-165294039 on Build GRCh38
rs962697383 Chr.2: 165294116-165294116 on Build GRCh38
rs886054991 Chr.2: 165294037-165294038 on Build GRCh38
rs886054999 Chr.2: 165294121-165294121 on Build GRCh38
rs879316715 Chr.2: 165294161-165294161 on Build GRCh38
rs771454481 Chr.2: 165294013-165294024 on Build GRCh38
rs886054992 Chr.2: 165294038-165294040 on Build GRCh38
rs749326085 Chr.2: 165293848-165293848 on Build GRCh38
rs35672867 Chr.2: 165294172-165294172 on Build GRCh38
rs1053910718 Chr.2: 165294079-165294079 on Build GRCh38
rs368442485 Chr.2: 165293932-165293932 on Build GRCh38
rs1044773891 Chr.2: 165293852-165293852 on Build GRCh38
rs908525550 Chr.2: 165294056-165294056 on Build GRCh38
rs892319581 Chr.2: 165294193-165294193 on Build GRCh38
rs754857526 Chr.2: 165294158-165294158 on Build GRCh38
rs886054993 Chr.2: 165294039-165294040 on Build GRCh38
rs796053109 Chr.2: 165294115-165294115 on Build GRCh38
rs1004370268 Chr.2: 165294039-165294039 on Build GRCh38
rs982889022 Chr.2: 165294047-165294047 on Build GRCh38
rs1009787252 Chr.2: 165294194-165294194 on Build GRCh38
rs147714927 Chr.2: 165293856-165293856 on Build GRCh38
rs777419988 Chr.2: 165294179-165294179 on Build GRCh38
rs758147516 Chr.2: 165293985-165293985 on Build GRCh38

View Full Product Details