Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 171784409-171784953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A12
Entrez Gene ID
8604
Gene Name
solute carrier family 25 member 12
Gene Aliases
AGC1, ARALAR
UniGene
Hs.470608
Gene Chromosome Location
Chr.2: 171783405-171894306 on Build GRCh38
Transcripts
NM_003705.4 NR_047549.1 XM_011512070.2

SNP Detail

SNP ID Chromosome Location
rs921170429 Chr.2: 171784441-171784441 on Build GRCh38
rs1055741489 Chr.2: 171784833-171784833 on Build GRCh38
rs555515048 Chr.2: 171784683-171784683 on Build GRCh38
rs113965617 Chr.2: 171784580-171784580 on Build GRCh38
rs550984952 Chr.2: 171784625-171784625 on Build GRCh38
rs1045350302 Chr.2: 171784697-171784697 on Build GRCh38
rs181107670 Chr.2: 171784523-171784523 on Build GRCh38
rs34360900 Chr.2: 171784561-171784562 on Build GRCh38
rs975637545 Chr.2: 171784886-171784886 on Build GRCh38
rs886055123 Chr.2: 171784520-171784520 on Build GRCh38
rs1045417514 Chr.2: 171784434-171784434 on Build GRCh38
rs1034656563 Chr.2: 171784769-171784769 on Build GRCh38
rs902557738 Chr.2: 171784916-171784916 on Build GRCh38
rs966231989 Chr.2: 171784654-171784654 on Build GRCh38
rs199931948 Chr.2: 171784715-171784715 on Build GRCh38
rs1055626292 Chr.2: 171784534-171784534 on Build GRCh38
rs914394212 Chr.2: 171784842-171784842 on Build GRCh38
rs572305038 Chr.2: 171784816-171784816 on Build GRCh38
rs1022458705 Chr.2: 171784873-171784873 on Build GRCh38
rs761178693 Chr.2: 171784667-171784667 on Build GRCh38
rs930800472 Chr.2: 171784682-171784682 on Build GRCh38
rs764675274 Chr.2: 171784711-171784711 on Build GRCh38
rs191707233 Chr.2: 171784719-171784719 on Build GRCh38
rs984544799 Chr.2: 171784716-171784716 on Build GRCh38
rs981067904 Chr.2: 171784810-171784810 on Build GRCh38
rs757812385 Chr.2: 171784817-171784817 on Build GRCh38
rs932009945 Chr.2: 171784481-171784481 on Build GRCh38
rs1009277795 Chr.2: 171784864-171784864 on Build GRCh38
rs1003227636 Chr.2: 171784723-171784723 on Build GRCh38
rs1041569286 Chr.2: 171784913-171784913 on Build GRCh38
rs562448681 Chr.2: 171784507-171784507 on Build GRCh38
rs1028217027 Chr.2: 171784912-171784912 on Build GRCh38
rs967940880 Chr.2: 171784877-171784877 on Build GRCh38
rs999909899 Chr.2: 171784595-171784595 on Build GRCh38
rs568986072 Chr.2: 171784914-171784914 on Build GRCh38
rs961840546 Chr.2: 171784784-171784784 on Build GRCh38
rs767853194 Chr.2: 171784544-171784544 on Build GRCh38
rs1044225091 Chr.2: 171784578-171784578 on Build GRCh38
rs996419991 Chr.2: 171784657-171784657 on Build GRCh38
rs1018903231 Chr.2: 171784648-171784648 on Build GRCh38
rs754342085 Chr.2: 171784762-171784762 on Build GRCh38
rs11757 Chr.2: 171784535-171784535 on Build GRCh38

View Full Product Details