Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 179106467-179106974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SESTD1
Entrez Gene ID
91404
Gene Name
SEC14 and spectrin domain containing 1
Gene Aliases
SOLO
UniGene
Hs.30977
Gene Chromosome Location
Chr.2: 179101692-179264723 on Build GRCh38
Transcripts
NM_178123.4 XM_011512139.2 XM_011512141.2 XM_017005250.1 XM_017005251.1

SNP Detail

SNP ID Chromosome Location
rs79789502 Chr.2: 179106875-179106875 on Build GRCh38
rs766088432 Chr.2: 179106545-179106545 on Build GRCh38
rs1025272981 Chr.2: 179106524-179106524 on Build GRCh38
rs534670710 Chr.2: 179106585-179106585 on Build GRCh38
rs974548558 Chr.2: 179106855-179106855 on Build GRCh38
rs772758525 Chr.2: 179106528-179106528 on Build GRCh38
rs906637213 Chr.2: 179106922-179106922 on Build GRCh38
rs985122282 Chr.2: 179106843-179106843 on Build GRCh38
rs940368693 Chr.2: 179106911-179106911 on Build GRCh38
rs550304525 Chr.2: 179106513-179106513 on Build GRCh38
rs1025514601 Chr.2: 179106584-179106584 on Build GRCh38
rs1037312063 Chr.2: 179106642-179106642 on Build GRCh38
rs557474807 Chr.2: 179106741-179106741 on Build GRCh38
rs922386582 Chr.2: 179106801-179106801 on Build GRCh38
rs540984266 Chr.2: 179106919-179106919 on Build GRCh38
rs532898154 Chr.2: 179106502-179106502 on Build GRCh38
rs1013606703 Chr.2: 179106516-179106516 on Build GRCh38
rs2008999 Chr.2: 179106605-179106605 on Build GRCh38
rs773654532 Chr.2: 179106596-179106600 on Build GRCh38
rs111673835 Chr.2: 179106816-179106816 on Build GRCh38
rs952660762 Chr.2: 179106702-179106702 on Build GRCh38
rs186943887 Chr.2: 179106552-179106552 on Build GRCh38
rs904472882 Chr.2: 179106560-179106560 on Build GRCh38
rs549396800 Chr.2: 179106506-179106506 on Build GRCh38
rs920194143 Chr.2: 179106940-179106940 on Build GRCh38
rs935629599 Chr.2: 179106871-179106871 on Build GRCh38
rs891624668 Chr.2: 179106575-179106575 on Build GRCh38
rs796229231 Chr.2: 179106563-179106563 on Build GRCh38
rs751761325 Chr.2: 179106939-179106939 on Build GRCh38
rs191743636 Chr.2: 179106712-179106712 on Build GRCh38
rs1024973115 Chr.2: 179106768-179106768 on Build GRCh38
rs760331367 Chr.2: 179106542-179106542 on Build GRCh38
rs578015354 Chr.2: 179106607-179106607 on Build GRCh38
rs961853163 Chr.2: 179106779-179106779 on Build GRCh38
rs142703170 Chr.2: 179106777-179106777 on Build GRCh38
rs1055191453 Chr.2: 179106728-179106728 on Build GRCh38
rs16866635 Chr.2: 179106876-179106876 on Build GRCh38
rs976942163 Chr.2: 179106797-179106797 on Build GRCh38
rs1013970827 Chr.2: 179106752-179106752 on Build GRCh38
rs1045548549 Chr.2: 179106886-179106886 on Build GRCh38
rs940285550 Chr.2: 179106637-179106637 on Build GRCh38
rs200458669 Chr.2: 179106939-179106940 on Build GRCh38
rs7566810 Chr.2: 179106583-179106583 on Build GRCh38
rs1052939757 Chr.2: 179106574-179106574 on Build GRCh38
rs535693839 Chr.2: 179106534-179106534 on Build GRCh38
rs1052973332 Chr.2: 179106873-179106873 on Build GRCh38
rs753609078 Chr.2: 179106556-179106556 on Build GRCh38
rs921702303 Chr.2: 179106572-179106572 on Build GRCh38
rs758535357 Chr.2: 179106867-179106867 on Build GRCh38
rs570147870 Chr.2: 179106533-179106533 on Build GRCh38
rs995573685 Chr.2: 179106498-179106498 on Build GRCh38
rs1033665165 Chr.2: 179106687-179106687 on Build GRCh38
rs1002227403 Chr.2: 179106666-179106666 on Build GRCh38
rs542077095 Chr.2: 179106860-179106860 on Build GRCh38
rs759607720 Chr.2: 179106564-179106564 on Build GRCh38
rs771759998 Chr.2: 179106505-179106505 on Build GRCh38
rs953803464 Chr.2: 179106821-179106821 on Build GRCh38
rs921639495 Chr.2: 179106871-179106871 on Build GRCh38
rs931431085 Chr.2: 179106940-179106944 on Build GRCh38

View Full Product Details