Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 186638044-186638497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGAV
Entrez Gene ID
3685
Gene Name
integrin subunit alpha V
Gene Aliases
CD51, MSK8, VNRA, VTNR
UniGene
Hs.436873
Gene Chromosome Location
Chr.2: 186590063-186680902 on Build GRCh38
Transcripts
NM_001144999.2 NM_001145000.2 NM_002210.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6688109 c.863G>A p.G288E Chr.2: 186638425-186638425 on Build GRCh38
4511727 c.878C>T p.S293F Chr.2: 186638440-186638440 on Build GRCh38
6688111 c.881T>C p.L294S Chr.2: 186638443-186638443 on Build GRCh38
338710 c.842G>T p.G281V Chr.2: 186638316-186638316 on Build GRCh38
6688113 c.847-1G>T p.? Chr.2: 186638408-186638408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762215864 Chr.2: 186638379-186638379 on Build GRCh38
rs200278337 Chr.2: 186638346-186638346 on Build GRCh38
rs201196607 Chr.2: 186638358-186638358 on Build GRCh38
rs547573471 Chr.2: 186638174-186638174 on Build GRCh38
rs751464182 Chr.2: 186638272-186638272 on Build GRCh38
rs756042949 Chr.2: 186638434-186638434 on Build GRCh38
rs765579889 Chr.2: 186638384-186638384 on Build GRCh38
rs928242443 Chr.2: 186638241-186638241 on Build GRCh38
rs1039670607 Chr.2: 186638446-186638446 on Build GRCh38
rs370679979 Chr.2: 186638140-186638140 on Build GRCh38
rs1030989335 Chr.2: 186638126-186638126 on Build GRCh38
rs757565794 Chr.2: 186638255-186638262 on Build GRCh38
rs375987665 Chr.2: 186638351-186638351 on Build GRCh38
rs954648113 Chr.2: 186638104-186638104 on Build GRCh38
rs199603191 Chr.2: 186638307-186638307 on Build GRCh38
rs111527604 Chr.2: 186638259-186638259 on Build GRCh38
rs13422838 Chr.2: 186638119-186638119 on Build GRCh38
rs769112241 Chr.2: 186638355-186638355 on Build GRCh38
rs1037374027 Chr.2: 186638363-186638363 on Build GRCh38
rs777182270 Chr.2: 186638467-186638467 on Build GRCh38
rs530014949 Chr.2: 186638233-186638233 on Build GRCh38
rs201225541 Chr.2: 186638287-186638287 on Build GRCh38
rs201525624 Chr.2: 186638246-186638246 on Build GRCh38
rs202002317 Chr.2: 186638285-186638285 on Build GRCh38
rs752664881 Chr.2: 186638433-186638433 on Build GRCh38
rs370560229 Chr.2: 186638436-186638436 on Build GRCh38
rs552005760 Chr.2: 186638469-186638469 on Build GRCh38
rs201933397 Chr.2: 186638441-186638441 on Build GRCh38
rs1053070924 Chr.2: 186638156-186638156 on Build GRCh38
rs754911086 Chr.2: 186638281-186638281 on Build GRCh38
rs771551043 Chr.2: 186638462-186638462 on Build GRCh38
rs140473956 Chr.2: 186638459-186638459 on Build GRCh38
rs749663033 Chr.2: 186638254-186638255 on Build GRCh38
rs751613814 Chr.2: 186638398-186638398 on Build GRCh38
rs950445810 Chr.2: 186638362-186638362 on Build GRCh38
rs200643949 Chr.2: 186638394-186638394 on Build GRCh38
rs755073639 Chr.2: 186638399-186638399 on Build GRCh38
rs769686496 Chr.2: 186638293-186638293 on Build GRCh38
rs780112636 Chr.2: 186638261-186638261 on Build GRCh38
rs997543795 Chr.2: 186638146-186638146 on Build GRCh38
rs980925560 Chr.2: 186638193-186638193 on Build GRCh38
rs891865616 Chr.2: 186638161-186638164 on Build GRCh38
rs901811784 Chr.2: 186638137-186638137 on Build GRCh38
rs186998784 Chr.2: 186638361-186638361 on Build GRCh38
rs752672613 Chr.2: 186638196-186638196 on Build GRCh38
rs201930300 Chr.2: 186638460-186638460 on Build GRCh38
rs763177841 Chr.2: 186638392-186638392 on Build GRCh38
rs781325608 Chr.2: 186638421-186638421 on Build GRCh38
rs754155165 Chr.2: 186638232-186638232 on Build GRCh38
rs200557809 Chr.2: 186638291-186638291 on Build GRCh38
rs199654707 Chr.2: 186638457-186638457 on Build GRCh38
rs748910834 Chr.2: 186638451-186638451 on Build GRCh38
rs779135374 Chr.2: 186638394-186638395 on Build GRCh38
rs199807927 Chr.2: 186638386-186638386 on Build GRCh38
rs1049251404 Chr.2: 186638091-186638091 on Build GRCh38
rs748968952 Chr.2: 186638345-186638345 on Build GRCh38
rs201025247 Chr.2: 186638305-186638305 on Build GRCh38
rs899011476 Chr.2: 186638376-186638376 on Build GRCh38
rs765483746 Chr.2: 186638247-186638247 on Build GRCh38
rs202077086 Chr.2: 186638258-186638258 on Build GRCh38
rs777629355 Chr.2: 186638338-186638338 on Build GRCh38
rs761108726 Chr.2: 186638352-186638352 on Build GRCh38
rs777559283 Chr.2: 186638449-186638449 on Build GRCh38
rs750643703 Chr.2: 186638250-186638250 on Build GRCh38
rs565986069 Chr.2: 186638186-186638186 on Build GRCh38
rs761292740 Chr.2: 186638128-186638128 on Build GRCh38
rs112951216 Chr.2: 186638136-186638136 on Build GRCh38

View Full Product Details