Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196199118-196199659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECW2
Entrez Gene ID
57520
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2
Gene Aliases
NEDL2
UniGene
Hs.654742
Gene Chromosome Location
Chr.2: 196194382-196593692 on Build GRCh38
Transcripts
NM_001304840.1 NM_020760.2 XM_006712646.3 XM_006712648.3 XM_017004563.1

SNP Detail

SNP ID Chromosome Location
rs867424154 Chr.2: 196199185-196199185 on Build GRCh38
rs934527977 Chr.2: 196199526-196199526 on Build GRCh38
rs972618986 Chr.2: 196199342-196199342 on Build GRCh38
rs184542531 Chr.2: 196199163-196199163 on Build GRCh38
rs976072252 Chr.2: 196199495-196199495 on Build GRCh38
rs909353142 Chr.2: 196199610-196199610 on Build GRCh38
rs767097408 Chr.2: 196199553-196199553 on Build GRCh38
rs554107656 Chr.2: 196199483-196199483 on Build GRCh38
rs919351640 Chr.2: 196199383-196199383 on Build GRCh38
rs923440177 Chr.2: 196199153-196199153 on Build GRCh38
rs182181336 Chr.2: 196199502-196199502 on Build GRCh38
rs900926655 Chr.2: 196199598-196199598 on Build GRCh38
rs536145108 Chr.2: 196199525-196199525 on Build GRCh38
rs953692859 Chr.2: 196199187-196199187 on Build GRCh38
rs1045590961 Chr.2: 196199353-196199353 on Build GRCh38
rs141855178 Chr.2: 196199434-196199434 on Build GRCh38
rs373618484 Chr.2: 196199205-196199205 on Build GRCh38
rs375663596 Chr.2: 196199493-196199493 on Build GRCh38
rs560819642 Chr.2: 196199309-196199309 on Build GRCh38
rs533738519 Chr.2: 196199477-196199477 on Build GRCh38
rs190079843 Chr.2: 196199254-196199254 on Build GRCh38
rs1011642003 Chr.2: 196199423-196199423 on Build GRCh38
rs941106150 Chr.2: 196199231-196199231 on Build GRCh38
rs10782 Chr.2: 196199420-196199420 on Build GRCh38
rs373852413 Chr.2: 196199270-196199270 on Build GRCh38
rs140831433 Chr.2: 196199190-196199190 on Build GRCh38
rs745838821 Chr.2: 196199338-196199338 on Build GRCh38
rs1020296878 Chr.2: 196199425-196199425 on Build GRCh38
rs1053320840 Chr.2: 196199539-196199539 on Build GRCh38
rs771834423 Chr.2: 196199343-196199343 on Build GRCh38
rs142328555 Chr.2: 196199149-196199149 on Build GRCh38
rs986406221 Chr.2: 196199211-196199211 on Build GRCh38
rs778773336 Chr.2: 196199182-196199182 on Build GRCh38
rs932174503 Chr.2: 196199161-196199161 on Build GRCh38
rs1034170939 Chr.2: 196199412-196199412 on Build GRCh38
rs1001814994 Chr.2: 196199407-196199407 on Build GRCh38
rs998384939 Chr.2: 196199627-196199627 on Build GRCh38
rs923557328 Chr.2: 196199500-196199500 on Build GRCh38
rs948704008 Chr.2: 196199445-196199445 on Build GRCh38
rs963592308 Chr.2: 196199261-196199261 on Build GRCh38
rs765810996 Chr.2: 196199611-196199611 on Build GRCh38
rs907112206 Chr.2: 196199395-196199395 on Build GRCh38
rs556369873 Chr.2: 196199597-196199597 on Build GRCh38
rs571280779 Chr.2: 196199474-196199474 on Build GRCh38
rs151265443 Chr.2: 196199421-196199421 on Build GRCh38
rs1044148 Chr.2: 196199616-196199616 on Build GRCh38

View Full Product Details