Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 197500882-197501381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPE1
Entrez Gene ID
3336
Gene Name
heat shock protein family E (Hsp10) member 1
Gene Aliases
CPN10, EPF, GROES, HSP10
UniGene
Hs.1197
Gene Chromosome Location
Chr.2: 197499997-197503463 on Build GRCh38
Transcripts
NM_002157.2 NM_001202485.1

Gene Symbol
HSPE1-MOB4
Entrez Gene ID
100529241
Gene Name
HSPE1-MOB4 readthrough
Gene Aliases
HSPE1-PHOCN
UniGene
Hs.633165
Gene Chromosome Location
Chr.2: 197499997-197553699 on Build GRCh38
Transcripts
NM_002157.2 NM_001202485.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6455940 c.152C>T p.S51L Chr.2: 197501222-197501222 on Build GRCh38
3990963 c.87A>G p.G29G Chr.2: 197501157-197501157 on Build GRCh38
6289409 c.44G>A p.R15Q Chr.2: 197501114-197501114 on Build GRCh38
3576074 c.122T>C p.V41A Chr.2: 197501192-197501192 on Build GRCh38
5058538 c.134C>G p.T45R Chr.2: 197501204-197501204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924977211 Chr.2: 197501291-197501291 on Build GRCh38
rs374253511 Chr.2: 197501070-197501070 on Build GRCh38
rs112568177 Chr.2: 197501226-197501226 on Build GRCh38
rs781536932 Chr.2: 197501280-197501280 on Build GRCh38
rs142886549 Chr.2: 197501306-197501306 on Build GRCh38
rs755327763 Chr.2: 197501351-197501351 on Build GRCh38
rs374783471 Chr.2: 197500906-197500906 on Build GRCh38
rs139114485 Chr.2: 197501176-197501176 on Build GRCh38
rs779633648 Chr.2: 197501058-197501058 on Build GRCh38
rs372780408 Chr.2: 197501124-197501124 on Build GRCh38
rs770044838 Chr.2: 197501139-197501139 on Build GRCh38
rs960600265 Chr.2: 197501193-197501193 on Build GRCh38
rs777798969 Chr.2: 197501250-197501250 on Build GRCh38
rs776111902 Chr.2: 197501144-197501144 on Build GRCh38
rs80032346 Chr.2: 197500923-197500924 on Build GRCh38
rs893086607 Chr.2: 197501197-197501197 on Build GRCh38
rs772319518 Chr.2: 197501098-197501098 on Build GRCh38
rs747094150 Chr.2: 197501263-197501263 on Build GRCh38
rs113357080 Chr.2: 197501324-197501324 on Build GRCh38
rs1015896262 Chr.2: 197501256-197501256 on Build GRCh38
rs754852521 Chr.2: 197501217-197501217 on Build GRCh38
rs1000014618 Chr.2: 197500984-197500984 on Build GRCh38
rs945035214 Chr.2: 197500959-197500959 on Build GRCh38
rs752498035 Chr.2: 197501245-197501245 on Build GRCh38
rs546209685 Chr.2: 197500926-197500926 on Build GRCh38
rs776654956 Chr.2: 197501132-197501132 on Build GRCh38
rs757412168 Chr.2: 197501276-197501276 on Build GRCh38
rs2305560 Chr.2: 197501064-197501064 on Build GRCh38
rs1030120330 Chr.2: 197501271-197501271 on Build GRCh38
rs185690747 Chr.2: 197501181-197501181 on Build GRCh38
rs748743564 Chr.2: 197501189-197501191 on Build GRCh38
rs943029682 Chr.2: 197501297-197501297 on Build GRCh38
rs758433207 Chr.2: 197501247-197501247 on Build GRCh38
rs766989335 Chr.2: 197501037-197501037 on Build GRCh38
rs570971820 Chr.2: 197501262-197501263 on Build GRCh38
rs778457721 Chr.2: 197501174-197501174 on Build GRCh38
rs985677881 Chr.2: 197501020-197501020 on Build GRCh38
rs770231817 Chr.2: 197501282-197501282 on Build GRCh38
rs1018380262 Chr.2: 197501293-197501296 on Build GRCh38
rs1007041636 Chr.2: 197501290-197501292 on Build GRCh38
rs370386655 Chr.2: 197501065-197501065 on Build GRCh38
rs902617778 Chr.2: 197500976-197500976 on Build GRCh38
rs1016179427 Chr.2: 197501107-197501107 on Build GRCh38
rs375552499 Chr.2: 197501244-197501244 on Build GRCh38
rs760573266 Chr.2: 197501180-197501180 on Build GRCh38
rs918245126 Chr.2: 197501079-197501079 on Build GRCh38
rs755791347 Chr.2: 197501054-197501054 on Build GRCh38
rs763415808 Chr.2: 197501166-197501166 on Build GRCh38
rs761441218 Chr.2: 197501026-197501026 on Build GRCh38
rs901578072 Chr.2: 197501349-197501349 on Build GRCh38
rs1025319540 Chr.2: 197501334-197501334 on Build GRCh38
rs955693131 Chr.2: 197500998-197500999 on Build GRCh38
rs377415140 Chr.2: 197501052-197501052 on Build GRCh38
rs775744688 Chr.2: 197501290-197501290 on Build GRCh38
rs973734042 Chr.2: 197501067-197501067 on Build GRCh38
rs766851508 Chr.2: 197501184-197501184 on Build GRCh38
rs771058605 Chr.2: 197501012-197501012 on Build GRCh38
rs766023643 Chr.2: 197501190-197501190 on Build GRCh38
rs1039189116 Chr.2: 197500998-197500998 on Build GRCh38
rs1007279933 Chr.2: 197500991-197500991 on Build GRCh38
rs368813176 Chr.2: 197501109-197501109 on Build GRCh38
rs903605115 Chr.2: 197500983-197500983 on Build GRCh38
rs748015639 Chr.2: 197501082-197501082 on Build GRCh38
rs760016331 Chr.2: 197501133-197501133 on Build GRCh38
rs902503936 Chr.2: 197501246-197501246 on Build GRCh38
rs763104748 Chr.2: 197500968-197500968 on Build GRCh38
rs760120008 Chr.2: 197501189-197501189 on Build GRCh38
rs778032552 Chr.2: 197501103-197501103 on Build GRCh38
rs148530634 Chr.2: 197501208-197501208 on Build GRCh38
rs913806409 Chr.2: 197500954-197500954 on Build GRCh38
rs750112711 Chr.2: 197501048-197501048 on Build GRCh38

View Full Product Details