Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201625852-201626361 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM237
Entrez Gene ID
65062
Gene Name
transmembrane protein 237
Gene Aliases
ALS2CR4, JBTS14
UniGene
Hs.12319
Gene Chromosome Location
Chr.2: 201620184-201643529 on Build GRCh38
Transcripts
NM_001044385.2 NM_152388.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4734486 c.1128_1131delGTCT p.L376fs*12 Chr.2: 201626054-201626057 on Build GRCh38
4653419 c.1119G>T p.W373C Chr.2: 201626066-201626066 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758712743 Chr.2: 201626037-201626037 on Build GRCh38
rs923985649 Chr.2: 201626332-201626332 on Build GRCh38
rs201921833 Chr.2: 201625981-201625981 on Build GRCh38
rs753804873 Chr.2: 201626011-201626011 on Build GRCh38
rs555926215 Chr.2: 201625931-201625931 on Build GRCh38
rs755985117 Chr.2: 201625999-201625999 on Build GRCh38
rs1016481933 Chr.2: 201626188-201626188 on Build GRCh38
rs13011356 Chr.2: 201626304-201626304 on Build GRCh38
rs892868640 Chr.2: 201625942-201625942 on Build GRCh38
rs1012533709 Chr.2: 201626241-201626241 on Build GRCh38
rs757309896 Chr.2: 201626174-201626174 on Build GRCh38
rs1047227861 Chr.2: 201626115-201626115 on Build GRCh38
rs936460763 Chr.2: 201625909-201625909 on Build GRCh38
rs748301153 Chr.2: 201626034-201626034 on Build GRCh38
rs77570029 Chr.2: 201626051-201626051 on Build GRCh38
rs928915995 Chr.2: 201625902-201625902 on Build GRCh38
rs747158935 Chr.2: 201626050-201626050 on Build GRCh38
rs543786476 Chr.2: 201626189-201626189 on Build GRCh38
rs751952525 Chr.2: 201626118-201626119 on Build GRCh38
rs753716785 Chr.2: 201626116-201626116 on Build GRCh38
rs763774534 Chr.2: 201625893-201625893 on Build GRCh38
rs886055447 Chr.2: 201626070-201626070 on Build GRCh38
rs770483878 Chr.2: 201626196-201626196 on Build GRCh38
rs756738471 Chr.2: 201626101-201626101 on Build GRCh38
rs766935734 Chr.2: 201625986-201625986 on Build GRCh38
rs934926603 Chr.2: 201626269-201626269 on Build GRCh38
rs746307378 Chr.2: 201626201-201626201 on Build GRCh38
rs373965403 Chr.2: 201626075-201626075 on Build GRCh38
rs1045981401 Chr.2: 201625900-201625900 on Build GRCh38
rs1053632038 Chr.2: 201625969-201625969 on Build GRCh38
rs781429961 Chr.2: 201626184-201626184 on Build GRCh38
rs752545565 Chr.2: 201626134-201626134 on Build GRCh38
rs978122378 Chr.2: 201626288-201626288 on Build GRCh38
rs776066221 Chr.2: 201626083-201626083 on Build GRCh38
rs928864245 Chr.2: 201626221-201626221 on Build GRCh38
rs750009423 Chr.2: 201625994-201625994 on Build GRCh38
rs1008510854 Chr.2: 201626025-201626025 on Build GRCh38
rs758523904 Chr.2: 201626153-201626153 on Build GRCh38
rs751757163 Chr.2: 201626166-201626166 on Build GRCh38
rs192172407 Chr.2: 201625905-201625905 on Build GRCh38
rs201817510 Chr.2: 201626004-201626004 on Build GRCh38
rs770221961 Chr.2: 201626080-201626080 on Build GRCh38
rs760192361 Chr.2: 201626096-201626096 on Build GRCh38
rs138509553 Chr.2: 201626095-201626095 on Build GRCh38
rs370795793 Chr.2: 201626071-201626071 on Build GRCh38
rs562262700 Chr.2: 201626216-201626216 on Build GRCh38
rs754717992 Chr.2: 201626023-201626023 on Build GRCh38
rs989091210 Chr.2: 201626270-201626270 on Build GRCh38
rs73989521 Chr.2: 201626089-201626089 on Build GRCh38
rs766664373 Chr.2: 201626009-201626009 on Build GRCh38
rs200863054 Chr.2: 201626044-201626044 on Build GRCh38
rs765897159 Chr.2: 201626104-201626104 on Build GRCh38
rs149240122 Chr.2: 201626120-201626120 on Build GRCh38
rs374578541 Chr.2: 201626193-201626193 on Build GRCh38
rs749208698 Chr.2: 201625943-201625943 on Build GRCh38
rs778693371 Chr.2: 201626024-201626024 on Build GRCh38
rs765010894 Chr.2: 201626128-201626128 on Build GRCh38
rs764930527 Chr.2: 201626217-201626217 on Build GRCh38
rs75707201 Chr.2: 201626281-201626281 on Build GRCh38

View Full Product Details