Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 209507461-209507969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP2
Entrez Gene ID
4133
Gene Name
microtubule associated protein 2
Gene Aliases
MAP2A, MAP2B, MAP2C
UniGene
Hs.368281
Gene Chromosome Location
Chr.2: 209424047-209734118 on Build GRCh38
Transcripts
NM_001039538.1

SNP Detail

SNP ID Chromosome Location
rs900155589 Chr.2: 209507615-209507615 on Build GRCh38
rs972369926 Chr.2: 209507792-209507792 on Build GRCh38
rs939767261 Chr.2: 209507785-209507785 on Build GRCh38
rs567490253 Chr.2: 209507628-209507628 on Build GRCh38
rs1021663437 Chr.2: 209507535-209507535 on Build GRCh38
rs554452766 Chr.2: 209507728-209507728 on Build GRCh38
rs758574313 Chr.2: 209507796-209507796 on Build GRCh38
rs1031650786 Chr.2: 209507659-209507659 on Build GRCh38
rs1010512190 Chr.2: 209507624-209507624 on Build GRCh38
rs182736369 Chr.2: 209507710-209507710 on Build GRCh38
rs755955296 Chr.2: 209507918-209507918 on Build GRCh38
rs765240527 Chr.2: 209507731-209507731 on Build GRCh38
rs569099779 Chr.2: 209507494-209507494 on Build GRCh38
rs960091490 Chr.2: 209507671-209507671 on Build GRCh38
rs752766116 Chr.2: 209507755-209507755 on Build GRCh38
rs12992468 Chr.2: 209507908-209507908 on Build GRCh38
rs12327904 Chr.2: 209507501-209507501 on Build GRCh38
rs553530530 Chr.2: 209507780-209507780 on Build GRCh38
rs902421665 Chr.2: 209507647-209507647 on Build GRCh38
rs567812934 Chr.2: 209507891-209507891 on Build GRCh38
rs998782872 Chr.2: 209507657-209507657 on Build GRCh38
rs951657468 Chr.2: 209507911-209507911 on Build GRCh38
rs759682761 Chr.2: 209507672-209507672 on Build GRCh38
rs1012308842 Chr.2: 209507737-209507737 on Build GRCh38
rs754793486 Chr.2: 209507779-209507779 on Build GRCh38
rs1023658406 Chr.2: 209507781-209507781 on Build GRCh38
rs993138802 Chr.2: 209507677-209507677 on Build GRCh38
rs974086360 Chr.2: 209507826-209507826 on Build GRCh38

View Full Product Details