Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 216096130-216096678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM169
Entrez Gene ID
92691
Gene Name
transmembrane protein 169
Gene Aliases
-
UniGene
Hs.334916
Gene Chromosome Location
Chr.2: 216081866-216102783 on Build GRCh38
Transcripts
NM_001142310.1 NM_001142311.1 NM_001142312.1 NM_138390.3 XM_006712851.1 XM_011512173.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5075542 c.271+10T>C p.? Chr.2: 216096244-216096244 on Build GRCh38
2717545 c.199G>A p.G67R Chr.2: 216096162-216096162 on Build GRCh38
6324400 c.180G>A p.E60E Chr.2: 216096143-216096143 on Build GRCh38
7164702 c.215G>A p.G72E Chr.2: 216096178-216096178 on Build GRCh38
292475 c.198C>T p.P66P Chr.2: 216096161-216096161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750336955 Chr.2: 216096165-216096165 on Build GRCh38
rs562632396 Chr.2: 216096527-216096527 on Build GRCh38
rs747860302 Chr.2: 216096404-216096404 on Build GRCh38
rs774332295 Chr.2: 216096159-216096159 on Build GRCh38
rs778140486 Chr.2: 216096219-216096219 on Build GRCh38
rs941071909 Chr.2: 216096371-216096371 on Build GRCh38
rs148759144 Chr.2: 216096288-216096288 on Build GRCh38
rs558048355 Chr.2: 216096362-216096362 on Build GRCh38
rs1024189627 Chr.2: 216096628-216096628 on Build GRCh38
rs546039047 Chr.2: 216096297-216096297 on Build GRCh38
rs7576830 Chr.2: 216096472-216096472 on Build GRCh38
rs775890136 Chr.2: 216096272-216096272 on Build GRCh38
rs778515573 Chr.2: 216096192-216096192 on Build GRCh38
rs1007056989 Chr.2: 216096344-216096346 on Build GRCh38
rs1018423436 Chr.2: 216096353-216096353 on Build GRCh38
rs375885920 Chr.2: 216096261-216096261 on Build GRCh38
rs372960907 Chr.2: 216096263-216096263 on Build GRCh38
rs544067207 Chr.2: 216096525-216096525 on Build GRCh38
rs998418745 Chr.2: 216096470-216096470 on Build GRCh38
rs374430787 Chr.2: 216096162-216096162 on Build GRCh38
rs944835769 Chr.2: 216096495-216096495 on Build GRCh38
rs573727011 Chr.2: 216096511-216096511 on Build GRCh38
rs772418359 Chr.2: 216096474-216096474 on Build GRCh38
rs551667102 Chr.2: 216096568-216096568 on Build GRCh38
rs1048612579 Chr.2: 216096337-216096337 on Build GRCh38
rs894426017 Chr.2: 216096607-216096607 on Build GRCh38
rs763205653 Chr.2: 216096273-216096273 on Build GRCh38
rs369121768 Chr.2: 216096585-216096585 on Build GRCh38
rs908256006 Chr.2: 216096368-216096368 on Build GRCh38
rs201188491 Chr.2: 216096265-216096265 on Build GRCh38
rs971227584 Chr.2: 216096634-216096634 on Build GRCh38
rs540257532 Chr.2: 216096450-216096450 on Build GRCh38
rs974145481 Chr.2: 216096384-216096384 on Build GRCh38
rs932225836 Chr.2: 216096394-216096394 on Build GRCh38
rs757312967 Chr.2: 216096306-216096306 on Build GRCh38
rs753343321 Chr.2: 216096181-216096181 on Build GRCh38
rs765865523 Chr.2: 216096177-216096177 on Build GRCh38
rs533183661 Chr.2: 216096546-216096546 on Build GRCh38
rs747758906 Chr.2: 216096202-216096202 on Build GRCh38
rs982555745 Chr.2: 216096334-216096334 on Build GRCh38
rs747527011 Chr.2: 216096241-216096241 on Build GRCh38
rs756023565 Chr.2: 216096166-216096166 on Build GRCh38
rs916392141 Chr.2: 216096641-216096641 on Build GRCh38
rs560474417 Chr.2: 216096592-216096592 on Build GRCh38
rs888244217 Chr.2: 216096342-216096342 on Build GRCh38
rs542247651 Chr.2: 216096519-216096519 on Build GRCh38
rs771462296 Chr.2: 216096244-216096244 on Build GRCh38
rs528455228 Chr.2: 216096613-216096613 on Build GRCh38
rs1012858194 Chr.2: 216096608-216096608 on Build GRCh38
rs754666930 Chr.2: 216096188-216096188 on Build GRCh38
rs920881806 Chr.2: 216096389-216096389 on Build GRCh38
rs572468891 Chr.2: 216096434-216096434 on Build GRCh38
rs758766962 Chr.2: 216096208-216096208 on Build GRCh38
rs148746587 Chr.2: 216096161-216096161 on Build GRCh38

View Full Product Details