Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 216675886-216676385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGFBP5
Entrez Gene ID
3488
Gene Name
insulin like growth factor binding protein 5
Gene Aliases
IBP5
UniGene
Hs.607212
Gene Chromosome Location
Chr.2: 216672105-216695549 on Build GRCh38
Transcripts
NM_000599.3

SNP Detail

SNP ID Chromosome Location
rs923860545 Chr.2: 216676253-216676253 on Build GRCh38
rs868362842 Chr.2: 216675955-216675955 on Build GRCh38
rs372646119 Chr.2: 216676056-216676056 on Build GRCh38
rs976636132 Chr.2: 216676363-216676363 on Build GRCh38
rs192524838 Chr.2: 216676198-216676198 on Build GRCh38
rs78303303 Chr.2: 216676320-216676320 on Build GRCh38
rs560047522 Chr.2: 216676165-216676165 on Build GRCh38
rs1055847890 Chr.2: 216676079-216676079 on Build GRCh38
rs752796421 Chr.2: 216676013-216676013 on Build GRCh38
rs150305201 Chr.2: 216676208-216676208 on Build GRCh38
rs911159475 Chr.2: 216676014-216676014 on Build GRCh38
rs1038409212 Chr.2: 216676096-216676096 on Build GRCh38
rs937027319 Chr.2: 216676054-216676054 on Build GRCh38
rs976583790 Chr.2: 216676020-216676020 on Build GRCh38
rs994404122 Chr.2: 216676115-216676115 on Build GRCh38
rs942694936 Chr.2: 216676088-216676088 on Build GRCh38
rs529363703 Chr.2: 216676271-216676271 on Build GRCh38
rs757018240 Chr.2: 216676308-216676308 on Build GRCh38
rs962142626 Chr.2: 216675964-216675964 on Build GRCh38
rs986693577 Chr.2: 216676293-216676293 on Build GRCh38
rs959263941 Chr.2: 216675937-216675937 on Build GRCh38
rs898739633 Chr.2: 216676110-216676110 on Build GRCh38
rs953563254 Chr.2: 216676304-216676304 on Build GRCh38
rs953412548 Chr.2: 216675991-216675991 on Build GRCh38
rs761624441 Chr.2: 216676332-216676332 on Build GRCh38
rs11575207 Chr.2: 216676236-216676236 on Build GRCh38
rs1012312656 Chr.2: 216676199-216676199 on Build GRCh38
rs184623727 Chr.2: 216675957-216675957 on Build GRCh38
rs955149684 Chr.2: 216676292-216676292 on Build GRCh38
rs113360140 Chr.2: 216675986-216675986 on Build GRCh38
rs929992214 Chr.2: 216676359-216676359 on Build GRCh38
rs911291159 Chr.2: 216676309-216676309 on Build GRCh38
rs913050306 Chr.2: 216676025-216676025 on Build GRCh38
rs1047322151 Chr.2: 216676130-216676130 on Build GRCh38
rs569239657 Chr.2: 216676321-216676321 on Build GRCh38
rs919904520 Chr.2: 216676341-216676341 on Build GRCh38
rs867484772 Chr.2: 216676200-216676200 on Build GRCh38
rs1015085649 Chr.2: 216675961-216675961 on Build GRCh38
rs765443315 Chr.2: 216675947-216675947 on Build GRCh38
rs915134736 Chr.2: 216675976-216675976 on Build GRCh38
rs141143461 Chr.2: 216676015-216676015 on Build GRCh38
rs896034070 Chr.2: 216675912-216675912 on Build GRCh38
rs911310190 Chr.2: 216676065-216676065 on Build GRCh38
rs986496752 Chr.2: 216676007-216676007 on Build GRCh38
rs933823208 Chr.2: 216676053-216676053 on Build GRCh38
rs967723724 Chr.2: 216676222-216676222 on Build GRCh38
rs907353355 Chr.2: 216676143-216676143 on Build GRCh38
rs187569565 Chr.2: 216676026-216676026 on Build GRCh38
rs1014442304 Chr.2: 216675917-216675917 on Build GRCh38
rs965289586 Chr.2: 216676335-216676335 on Build GRCh38
rs1042806713 Chr.2: 216676049-216676049 on Build GRCh38
rs1035143493 Chr.2: 216676178-216676178 on Build GRCh38

View Full Product Details