Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218580538-218581083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT9
Entrez Gene ID
9125
Gene Name
CCR4-NOT transcription complex subunit 9
Gene Aliases
CAF40, CT129, RCD-1, RCD1, RQCD1
UniGene
Hs.148767
Gene Chromosome Location
Chr.2: 218568579-218596435 on Build GRCh38
Transcripts
NM_001271634.1 NM_001271635.1 NM_005444.2 NR_073390.1 XM_011512138.2 XM_017005248.1 XM_017005249.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6573047 c.25-6C>G p.? Chr.2: 218580555-218580555 on Build GRCh38
1016384 c.199T>C p.L67L Chr.2: 218580735-218580735 on Build GRCh38
4552773 c.56G>T p.R19I Chr.2: 218580592-218580592 on Build GRCh38
1224282 c.149C>A p.P50H Chr.2: 218580685-218580685 on Build GRCh38
4816669 c.66C>G p.I22M Chr.2: 218580602-218580602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528237718 Chr.2: 218580760-218580760 on Build GRCh38
rs771337558 Chr.2: 218580585-218580585 on Build GRCh38
rs183957340 Chr.2: 218580892-218580892 on Build GRCh38
rs919256850 Chr.2: 218580949-218580949 on Build GRCh38
rs539438465 Chr.2: 218580999-218580999 on Build GRCh38
rs767519716 Chr.2: 218580695-218580695 on Build GRCh38
rs963584808 Chr.2: 218580886-218580886 on Build GRCh38
rs866418872 Chr.2: 218580712-218580712 on Build GRCh38
rs558244425 Chr.2: 218580977-218580977 on Build GRCh38
rs760332092 Chr.2: 218580701-218580701 on Build GRCh38
rs913772090 Chr.2: 218580584-218580584 on Build GRCh38
rs1052141781 Chr.2: 218580686-218580686 on Build GRCh38
rs747552817 Chr.2: 218580788-218580788 on Build GRCh38
rs937849965 Chr.2: 218580626-218580626 on Build GRCh38
rs750409757 Chr.2: 218580696-218580696 on Build GRCh38
rs138953648 Chr.2: 218580756-218580756 on Build GRCh38
rs1054944949 Chr.2: 218581022-218581022 on Build GRCh38
rs769980167 Chr.2: 218580627-218580627 on Build GRCh38
rs775884060 Chr.2: 218580990-218580990 on Build GRCh38
rs986723543 Chr.2: 218580863-218580863 on Build GRCh38
rs915104075 Chr.2: 218581025-218581025 on Build GRCh38
rs546424981 Chr.2: 218580827-218580827 on Build GRCh38
rs772958449 Chr.2: 218581043-218581043 on Build GRCh38
rs987338537 Chr.2: 218581033-218581033 on Build GRCh38
rs1026353888 Chr.2: 218580844-218580844 on Build GRCh38
rs970354661 Chr.2: 218580896-218580896 on Build GRCh38
rs868797174 Chr.2: 218580721-218580721 on Build GRCh38
rs778082401 Chr.2: 218580570-218580570 on Build GRCh38
rs62191561 Chr.2: 218580834-218580834 on Build GRCh38
rs781661777 Chr.2: 218580953-218580953 on Build GRCh38
rs945223293 Chr.2: 218581006-218581006 on Build GRCh38
rs899136184 Chr.2: 218580822-218580822 on Build GRCh38
rs374453349 Chr.2: 218580773-218580773 on Build GRCh38
rs989270726 Chr.2: 218580978-218580978 on Build GRCh38
rs761810000 Chr.2: 218580693-218580693 on Build GRCh38
rs750672423 Chr.2: 218580770-218580770 on Build GRCh38
rs201648255 Chr.2: 218580586-218580586 on Build GRCh38
rs937833542 Chr.2: 218581003-218581003 on Build GRCh38
rs1042333271 Chr.2: 218581040-218581040 on Build GRCh38
rs893744801 Chr.2: 218580761-218580761 on Build GRCh38
rs747285694 Chr.2: 218580575-218580575 on Build GRCh38
rs778192515 Chr.2: 218580787-218580787 on Build GRCh38
rs149423654 Chr.2: 218580853-218580853 on Build GRCh38
rs1055018024 Chr.2: 218580745-218580745 on Build GRCh38
rs772123573 Chr.2: 218581020-218581020 on Build GRCh38
rs760732839 Chr.2: 218581057-218581057 on Build GRCh38
rs369699855 Chr.2: 218580566-218580566 on Build GRCh38
rs752247114 Chr.2: 218580779-218580779 on Build GRCh38
rs1014362653 Chr.2: 218580869-218580869 on Build GRCh38
rs1020617473 Chr.2: 218580800-218580800 on Build GRCh38
rs1039353866 Chr.2: 218581049-218581049 on Build GRCh38
rs77659528 Chr.2: 218581034-218581034 on Build GRCh38
rs954873589 Chr.2: 218580858-218580858 on Build GRCh38
rs754721435 Chr.2: 218580764-218580764 on Build GRCh38
rs538328558 Chr.2: 218580790-218580790 on Build GRCh38
rs375292936 Chr.2: 218580981-218580981 on Build GRCh38
rs147458382 Chr.2: 218580971-218580971 on Build GRCh38
rs780594180 Chr.2: 218581011-218581011 on Build GRCh38
rs778376738 Chr.2: 218580765-218580765 on Build GRCh38
rs997908258 Chr.2: 218580838-218580838 on Build GRCh38
rs757937760 Chr.2: 218580784-218580784 on Build GRCh38
rs980723525 Chr.2: 218580944-218580944 on Build GRCh38
rs902217830 Chr.2: 218580808-218580808 on Build GRCh38
rs558949088 Chr.2: 218581009-218581009 on Build GRCh38
rs1039034786 Chr.2: 218580785-218580785 on Build GRCh38
rs775761220 Chr.2: 218580671-218580671 on Build GRCh38
rs759865221 Chr.2: 218580620-218580620 on Build GRCh38
rs1017753152 Chr.2: 218580885-218580885 on Build GRCh38

View Full Product Details