Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218822909-218823414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR9500
Entrez Gene ID
103504730
Gene Name
microRNA 9500
Gene Aliases
hsa-mir-9500
UniGene
Gene Chromosome Location
Chr.2: 218823090-218823154 on Build GRCh38
Transcripts
NR_128707.1 NM_017431.2 XM_017004343.1

Gene Symbol
PRKAG3
Entrez Gene ID
53632
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 3
Gene Aliases
AMPKG3
UniGene
Hs.591634
Gene Chromosome Location
Chr.2: 218821853-218831803 on Build GRCh38
Transcripts
NR_128707.1 NM_017431.2 XM_017004343.1

SNP Detail

SNP ID Chromosome Location
rs189998712 Chr.2: 218823021-218823021 on Build GRCh38
rs1009836930 Chr.2: 218823110-218823110 on Build GRCh38
rs991702880 Chr.2: 218823202-218823202 on Build GRCh38
rs191770659 Chr.2: 218823251-218823251 on Build GRCh38
rs34654778 Chr.2: 218823192-218823193 on Build GRCh38
rs34377776 Chr.2: 218823137-218823138 on Build GRCh38
rs948352618 Chr.2: 218823010-218823010 on Build GRCh38
rs997385965 Chr.2: 218823057-218823057 on Build GRCh38
rs536492494 Chr.2: 218823205-218823205 on Build GRCh38
rs1053866497 Chr.2: 218823053-218823053 on Build GRCh38
rs901178205 Chr.2: 218823123-218823123 on Build GRCh38
rs932944246 Chr.2: 218823311-218823311 on Build GRCh38
rs1041660544 Chr.2: 218823119-218823119 on Build GRCh38
rs925731069 Chr.2: 218823315-218823315 on Build GRCh38
rs540653338 Chr.2: 218823319-218823319 on Build GRCh38
rs767440336 Chr.2: 218823287-218823287 on Build GRCh38
rs1005965854 Chr.2: 218823374-218823374 on Build GRCh38
rs533686984 Chr.2: 218822994-218822994 on Build GRCh38
rs571519772 Chr.2: 218823135-218823135 on Build GRCh38
rs147201106 Chr.2: 218823273-218823273 on Build GRCh38
rs1050330704 Chr.2: 218823312-218823312 on Build GRCh38
rs150852890 Chr.2: 218823125-218823125 on Build GRCh38
rs917113018 Chr.2: 218823265-218823265 on Build GRCh38
rs952759910 Chr.2: 218823148-218823148 on Build GRCh38
rs892614126 Chr.2: 218823091-218823091 on Build GRCh38
rs543110527 Chr.2: 218823255-218823255 on Build GRCh38
rs978855409 Chr.2: 218823249-218823249 on Build GRCh38
rs371359505 Chr.2: 218823289-218823289 on Build GRCh38
rs527906757 Chr.2: 218823075-218823075 on Build GRCh38
rs961473801 Chr.2: 218823219-218823219 on Build GRCh38
rs139292718 Chr.2: 218823232-218823232 on Build GRCh38
rs888658868 Chr.2: 218823336-218823336 on Build GRCh38
rs947442242 Chr.2: 218823235-218823235 on Build GRCh38
rs1005412724 Chr.2: 218823168-218823168 on Build GRCh38
rs936872742 Chr.2: 218823050-218823050 on Build GRCh38
rs759443009 Chr.2: 218823051-218823051 on Build GRCh38
rs988493314 Chr.2: 218823375-218823375 on Build GRCh38
rs901391377 Chr.2: 218823270-218823270 on Build GRCh38
rs997252445 Chr.2: 218823124-218823124 on Build GRCh38
rs567458039 Chr.2: 218823099-218823099 on Build GRCh38
rs1043026610 Chr.2: 218822940-218822940 on Build GRCh38
rs149935762 Chr.2: 218823256-218823256 on Build GRCh38
rs553810151 Chr.2: 218823195-218823195 on Build GRCh38
rs762074730 Chr.2: 218823118-218823118 on Build GRCh38
rs1028502139 Chr.2: 218823062-218823062 on Build GRCh38
rs537942618 Chr.2: 218823102-218823102 on Build GRCh38
rs560758977 Chr.2: 218823024-218823024 on Build GRCh38
rs572356646 Chr.2: 218823204-218823204 on Build GRCh38
rs1035567476 Chr.2: 218823171-218823171 on Build GRCh38
rs763045317 Chr.2: 218823345-218823345 on Build GRCh38
rs980165641 Chr.2: 218823299-218823299 on Build GRCh38
rs934630102 Chr.2: 218823259-218823259 on Build GRCh38
rs187317587 Chr.2: 218823188-218823188 on Build GRCh38
rs961740881 Chr.2: 218823127-218823127 on Build GRCh38
rs1015985649 Chr.2: 218823385-218823385 on Build GRCh38
rs1041315871 Chr.2: 218823014-218823014 on Build GRCh38
rs925700059 Chr.2: 218823026-218823026 on Build GRCh38
rs182377046 Chr.2: 218823076-218823076 on Build GRCh38

View Full Product Details