Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219178687-219179221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM134A
Entrez Gene ID
79137
Gene Name
family with sequence similarity 134 member A
Gene Aliases
C2orf17, MAG-2
UniGene
Hs.516707
Gene Chromosome Location
Chr.2: 219178190-219185475 on Build GRCh38
Transcripts
NM_001321109.1 NM_001321110.1 NM_024293.5 XM_005246848.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7172046 c.367C>A p.R123S Chr.2: 219179007-219179007 on Build GRCh38
3577898 c.290C>T p.S97F Chr.2: 219178930-219178930 on Build GRCh38
1752355 c.296C>G p.S99W Chr.2: 219178936-219178936 on Build GRCh38
7296470 c.302T>C p.L101P Chr.2: 219178942-219178942 on Build GRCh38
6658458 c.341A>G p.Y114C Chr.2: 219178981-219178981 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781467127 Chr.2: 219179009-219179009 on Build GRCh38
rs907159073 Chr.2: 219178816-219178816 on Build GRCh38
rs562035872 Chr.2: 219178789-219178789 on Build GRCh38
rs745942058 Chr.2: 219178902-219178902 on Build GRCh38
rs769712735 Chr.2: 219179122-219179122 on Build GRCh38
rs550958752 Chr.2: 219178928-219178928 on Build GRCh38
rs923885945 Chr.2: 219179036-219179043 on Build GRCh38
rs781180856 Chr.2: 219178839-219178839 on Build GRCh38
rs759080456 Chr.2: 219178959-219178959 on Build GRCh38
rs777981680 Chr.2: 219178878-219178878 on Build GRCh38
rs528306075 Chr.2: 219178743-219178743 on Build GRCh38
rs745682400 Chr.2: 219178948-219178948 on Build GRCh38
rs372160347 Chr.2: 219179076-219179076 on Build GRCh38
rs931046390 Chr.2: 219179190-219179190 on Build GRCh38
rs758693368 Chr.2: 219178876-219178876 on Build GRCh38
rs1032328198 Chr.2: 219179015-219179015 on Build GRCh38
rs559102953 Chr.2: 219178710-219178710 on Build GRCh38
rs1013675346 Chr.2: 219178881-219178881 on Build GRCh38
rs757528871 Chr.2: 219179005-219179005 on Build GRCh38
rs943833685 Chr.2: 219179165-219179165 on Build GRCh38
rs956679630 Chr.2: 219179110-219179110 on Build GRCh38
rs900288970 Chr.2: 219178715-219178715 on Build GRCh38
rs753187196 Chr.2: 219178969-219178969 on Build GRCh38
rs762586877 Chr.2: 219179039-219179047 on Build GRCh38
rs765665115 Chr.2: 219178964-219178964 on Build GRCh38
rs745380052 Chr.2: 219179064-219179064 on Build GRCh38
rs1048047641 Chr.2: 219178766-219178766 on Build GRCh38
rs368573403 Chr.2: 219178981-219178981 on Build GRCh38
rs887911190 Chr.2: 219179194-219179194 on Build GRCh38
rs750538853 Chr.2: 219179014-219179014 on Build GRCh38
rs921052300 Chr.2: 219179172-219179172 on Build GRCh38
rs375522296 Chr.2: 219178956-219178956 on Build GRCh38
rs567052785 Chr.2: 219179092-219179092 on Build GRCh38
rs150316142 Chr.2: 219178745-219178745 on Build GRCh38
rs533462258 Chr.2: 219178986-219178986 on Build GRCh38
rs148389515 Chr.2: 219178790-219178791 on Build GRCh38
rs747929479 Chr.2: 219178831-219178832 on Build GRCh38
rs888052113 Chr.2: 219178855-219178855 on Build GRCh38
rs182226169 Chr.2: 219178974-219178974 on Build GRCh38
rs777759884 Chr.2: 219179046-219179046 on Build GRCh38
rs746785827 Chr.2: 219179048-219179048 on Build GRCh38
rs551863613 Chr.2: 219179170-219179170 on Build GRCh38
rs1022112674 Chr.2: 219178897-219178897 on Build GRCh38
rs1040490347 Chr.2: 219178714-219178714 on Build GRCh38
rs57922572 Chr.2: 219178793-219178794 on Build GRCh38
rs187189509 Chr.2: 219179078-219179078 on Build GRCh38
rs1005531569 Chr.2: 219178863-219178863 on Build GRCh38
rs772134414 Chr.2: 219179058-219179058 on Build GRCh38
rs757354652 Chr.2: 219178893-219178893 on Build GRCh38
rs200151781 Chr.2: 219178907-219178907 on Build GRCh38
rs770684363 Chr.2: 219178946-219178946 on Build GRCh38
rs772725311 Chr.2: 219178885-219178888 on Build GRCh38
rs764566524 Chr.2: 219179079-219179079 on Build GRCh38
rs770772129 Chr.2: 219179055-219179055 on Build GRCh38
rs763307175 Chr.2: 219179176-219179176 on Build GRCh38
rs372335730 Chr.2: 219178891-219178891 on Build GRCh38
rs912397826 Chr.2: 219179146-219179146 on Build GRCh38
rs577888287 Chr.2: 219179038-219179038 on Build GRCh38
rs769296535 Chr.2: 219179075-219179075 on Build GRCh38
rs1049249397 Chr.2: 219178779-219178780 on Build GRCh38
rs995165422 Chr.2: 219178739-219178739 on Build GRCh38
rs979468872 Chr.2: 219178939-219178939 on Build GRCh38
rs781293817 Chr.2: 219178894-219178894 on Build GRCh38
rs760364405 Chr.2: 219178940-219178940 on Build GRCh38
rs747902504 Chr.2: 219178909-219178909 on Build GRCh38
rs748071753 Chr.2: 219179017-219179017 on Build GRCh38
rs1015136827 Chr.2: 219178791-219178791 on Build GRCh38
rs753067949 Chr.2: 219178875-219178875 on Build GRCh38
rs555737587 Chr.2: 219179171-219179171 on Build GRCh38
rs758300164 Chr.2: 219179039-219179039 on Build GRCh38
rs756154899 Chr.2: 219179016-219179016 on Build GRCh38

View Full Product Details