Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 222201629-222202138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAX3
Entrez Gene ID
5077
Gene Name
paired box 3
Gene Aliases
CDHS, HUP2, WS1, WS3
UniGene
Hs.42146
Gene Chromosome Location
Chr.2: 222199887-222298996 on Build GRCh38
Transcripts
NM_001127366.2 NM_181457.3 NM_181458.3 NM_181459.3 NM_181460.3 NM_181461.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4795983 c.1369G>A p.G457S Chr.2: 222201995-222201995 on Build GRCh38
6576174 c.1380G>T p.M460I Chr.2: 222201984-222201984 on Build GRCh38
1017001 c.1361G>T p.S454I Chr.2: 222202003-222202003 on Build GRCh38
4781844 c.1231G>A p.A411T Chr.2: 222202133-222202133 on Build GRCh38
4772966 c.1272G>T p.T424T Chr.2: 222202092-222202092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027769694 Chr.2: 222201835-222201835 on Build GRCh38
rs186207055 Chr.2: 222201889-222201889 on Build GRCh38
rs754795764 Chr.2: 222202051-222202051 on Build GRCh38
rs889542927 Chr.2: 222201997-222201997 on Build GRCh38
rs888262810 Chr.2: 222201839-222201839 on Build GRCh38
rs944943242 Chr.2: 222202058-222202058 on Build GRCh38
rs945514356 Chr.2: 222201951-222201951 on Build GRCh38
rs776696114 Chr.2: 222201981-222201981 on Build GRCh38
rs995327866 Chr.2: 222201833-222201833 on Build GRCh38
rs1047857427 Chr.2: 222201817-222201817 on Build GRCh38
rs778669266 Chr.2: 222202053-222202053 on Build GRCh38
rs959065396 Chr.2: 222201899-222201899 on Build GRCh38
rs1028504308 Chr.2: 222201676-222201676 on Build GRCh38
rs937767149 Chr.2: 222201692-222201692 on Build GRCh38
rs759042262 Chr.2: 222201877-222201877 on Build GRCh38
rs34690509 Chr.2: 222201747-222201748 on Build GRCh38
rs752127930 Chr.2: 222202010-222202010 on Build GRCh38
rs372266013 Chr.2: 222202000-222202000 on Build GRCh38
rs368725878 Chr.2: 222201818-222201818 on Build GRCh38
rs540254930 Chr.2: 222201921-222201921 on Build GRCh38
rs972225778 Chr.2: 222201733-222201733 on Build GRCh38
rs1005216793 Chr.2: 222202067-222202067 on Build GRCh38
rs377157062 Chr.2: 222202032-222202032 on Build GRCh38
rs765696723 Chr.2: 222201985-222201985 on Build GRCh38
rs369886550 Chr.2: 222202087-222202087 on Build GRCh38
rs879468927 Chr.2: 222202005-222202005 on Build GRCh38
rs774274104 Chr.2: 222201935-222201935 on Build GRCh38
rs775861804 Chr.2: 222202107-222202107 on Build GRCh38
rs749014584 Chr.2: 222202046-222202046 on Build GRCh38
rs998446991 Chr.2: 222201923-222201923 on Build GRCh38
rs919452415 Chr.2: 222201738-222201738 on Build GRCh38
rs376147620 Chr.2: 222202116-222202116 on Build GRCh38
rs528350431 Chr.2: 222202115-222202115 on Build GRCh38
rs746597491 Chr.2: 222201811-222201811 on Build GRCh38
rs374089919 Chr.2: 222201933-222201933 on Build GRCh38
rs45624434 Chr.2: 222201724-222201724 on Build GRCh38
rs963660169 Chr.2: 222201732-222201732 on Build GRCh38
rs555977435 Chr.2: 222201792-222201792 on Build GRCh38
rs779693694 Chr.2: 222202039-222202039 on Build GRCh38
rs764632216 Chr.2: 222202008-222202008 on Build GRCh38
rs944659680 Chr.2: 222201782-222201782 on Build GRCh38
rs767090833 Chr.2: 222202015-222202015 on Build GRCh38
rs746069520 Chr.2: 222202092-222202092 on Build GRCh38
rs573451372 Chr.2: 222201743-222201743 on Build GRCh38
rs981757136 Chr.2: 222201661-222201661 on Build GRCh38
rs559630928 Chr.2: 222202059-222202059 on Build GRCh38
rs552778546 Chr.2: 222201817-222201818 on Build GRCh38
rs930688047 Chr.2: 222202118-222202118 on Build GRCh38
rs774896277 Chr.2: 222202031-222202031 on Build GRCh38
rs749209813 Chr.2: 222201924-222201924 on Build GRCh38
rs367865139 Chr.2: 222202020-222202020 on Build GRCh38
rs778236891 Chr.2: 222202111-222202111 on Build GRCh38
rs181872465 Chr.2: 222201723-222201723 on Build GRCh38
rs1016652010 Chr.2: 222202069-222202069 on Build GRCh38
rs747022299 Chr.2: 222201955-222201955 on Build GRCh38
rs989287440 Chr.2: 222201697-222201697 on Build GRCh38
rs756696533 Chr.2: 222202012-222202012 on Build GRCh38
rs373281216 Chr.2: 222201816-222201817 on Build GRCh38
rs926343135 Chr.2: 222201668-222201668 on Build GRCh38
rs970628895 Chr.2: 222201851-222201851 on Build GRCh38
rs886055674 Chr.2: 222201819-222201819 on Build GRCh38
rs763685386 Chr.2: 222201879-222201879 on Build GRCh38
rs375182072 Chr.2: 222201908-222201908 on Build GRCh38
rs763426571 Chr.2: 222201992-222201992 on Build GRCh38
rs776059693 Chr.2: 222201989-222201989 on Build GRCh38
rs753228919 Chr.2: 222201873-222201873 on Build GRCh38
rs953715918 Chr.2: 222201689-222201689 on Build GRCh38
rs937991969 Chr.2: 222201819-222201819 on Build GRCh38
rs770087251 Chr.2: 222202093-222202093 on Build GRCh38
rs768521579 Chr.2: 222201930-222201930 on Build GRCh38
rs544726519 Chr.2: 222201841-222201841 on Build GRCh38
rs560996755 Chr.2: 222201742-222201742 on Build GRCh38
rs369495504 Chr.2: 222201912-222201912 on Build GRCh38
rs900977338 Chr.2: 222201909-222201909 on Build GRCh38
rs770885464 Chr.2: 222202073-222202073 on Build GRCh38
rs199611711 Chr.2: 222201903-222201903 on Build GRCh38
rs780657657 Chr.2: 222201900-222201900 on Build GRCh38
rs771066036 Chr.2: 222201960-222201960 on Build GRCh38
rs78035924 Chr.2: 222201758-222201758 on Build GRCh38
rs1039060214 Chr.2: 222201795-222201795 on Build GRCh38
rs142988099 Chr.2: 222201840-222201840 on Build GRCh38

View Full Product Details