Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 224473368-224473867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL3
Entrez Gene ID
8452
Gene Name
cullin 3
Gene Aliases
CUL-3, PHA2E
UniGene
Hs.372286
Gene Chromosome Location
Chr.2: 224470150-224585397 on Build GRCh38
Transcripts
NM_001257197.1 NM_001257198.1 NM_003590.4 XM_006712800.2 XM_011511994.2 XM_011511995.1 XM_011511996.2

SNP Detail

SNP ID Chromosome Location
rs972547647 Chr.2: 224473420-224473423 on Build GRCh38
rs904756720 Chr.2: 224473625-224473625 on Build GRCh38
rs745531588 Chr.2: 224473631-224473631 on Build GRCh38
rs568774589 Chr.2: 224473487-224473487 on Build GRCh38
rs531395316 Chr.2: 224473509-224473509 on Build GRCh38
rs1016879346 Chr.2: 224473822-224473822 on Build GRCh38
rs572958976 Chr.2: 224473741-224473741 on Build GRCh38
rs376961030 Chr.2: 224473809-224473809 on Build GRCh38
rs960597314 Chr.2: 224473484-224473484 on Build GRCh38
rs553445597 Chr.2: 224473640-224473640 on Build GRCh38
rs927731743 Chr.2: 224473428-224473428 on Build GRCh38
rs887528034 Chr.2: 224473821-224473821 on Build GRCh38
rs781259483 Chr.2: 224473794-224473794 on Build GRCh38
rs1050399787 Chr.2: 224473683-224473683 on Build GRCh38
rs993238973 Chr.2: 224473393-224473393 on Build GRCh38
rs945204111 Chr.2: 224473566-224473566 on Build GRCh38
rs147054027 Chr.2: 224473576-224473576 on Build GRCh38
rs901305339 Chr.2: 224473725-224473725 on Build GRCh38
rs537781010 Chr.2: 224473494-224473494 on Build GRCh38
rs548761621 Chr.2: 224473438-224473438 on Build GRCh38
rs1041045216 Chr.2: 224473722-224473722 on Build GRCh38
rs539421515 Chr.2: 224473613-224473613 on Build GRCh38
rs1027821171 Chr.2: 224473402-224473402 on Build GRCh38
rs369378295 Chr.2: 224473695-224473695 on Build GRCh38
rs939079424 Chr.2: 224473458-224473458 on Build GRCh38
rs886055693 Chr.2: 224473700-224473700 on Build GRCh38
rs762720877 Chr.2: 224473454-224473456 on Build GRCh38
rs751530025 Chr.2: 224473670-224473670 on Build GRCh38
rs894773973 Chr.2: 224473535-224473535 on Build GRCh38
rs926333413 Chr.2: 224473463-224473463 on Build GRCh38
rs115972743 Chr.2: 224473687-224473687 on Build GRCh38
rs778642011 Chr.2: 224473581-224473581 on Build GRCh38
rs561931113 Chr.2: 224473405-224473405 on Build GRCh38
rs913710776 Chr.2: 224473526-224473526 on Build GRCh38
rs541625442 Chr.2: 224473783-224473783 on Build GRCh38
rs992495920 Chr.2: 224473488-224473488 on Build GRCh38
rs577196781 Chr.2: 224473589-224473589 on Build GRCh38
rs897955766 Chr.2: 224473784-224473784 on Build GRCh38
rs1027814917 Chr.2: 224473807-224473807 on Build GRCh38
rs755297097 Chr.2: 224473779-224473779 on Build GRCh38
rs980398909 Chr.2: 224473455-224473455 on Build GRCh38
rs998185840 Chr.2: 224473764-224473764 on Build GRCh38

View Full Product Details