Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 227342623-227343150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFF
Entrez Gene ID
56947
Gene Name
mitochondrial fission factor
Gene Aliases
C2orf33, GL004
UniGene
Hs.471528
Gene Chromosome Location
Chr.2: 227325151-227357836 on Build GRCh38
Transcripts
NM_001277061.1 NM_001277062.1 NM_001277063.1 NM_001277064.1 NM_001277065.1 NM_001277066.1 NM_001277067.1 NM_001277068.1 NM_020194.5 NR_102266.1 XM_005246681.3 XM_005246682.1 XM_005246683.1 XM_005246684.1 XM_005246685.3 XM_005246686.3 XM_005246687.3 XM_005246688.1 XM_006712637.1 XM_006712638.1 XM_006712639.2 XM_011511500.1 XM_011511501.1 XM_017004512.1 XM_017004513.1 XM_017004514.1 XM_017004515.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4500456 c.565C>T p.Q189* Chr.2: 227342791-227342791 on Build GRCh38
1017384 c.570A>T p.A190A Chr.2: 227342796-227342796 on Build GRCh38
1017383 c.545G>T p.R182I Chr.2: 227342771-227342771 on Build GRCh38
7103918 c.585G>C p.P195P Chr.2: 227342811-227342811 on Build GRCh38
5927234 c.557C>T p.S186L Chr.2: 227342783-227342783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs79113813 Chr.2: 227343006-227343006 on Build GRCh38
rs763899321 Chr.2: 227342807-227342807 on Build GRCh38
rs1015267578 Chr.2: 227342932-227342932 on Build GRCh38
rs939321092 Chr.2: 227343073-227343073 on Build GRCh38
rs368130845 Chr.2: 227342838-227342838 on Build GRCh38
rs573872401 Chr.2: 227342899-227342899 on Build GRCh38
rs555295345 Chr.2: 227342855-227342855 on Build GRCh38
rs918987869 Chr.2: 227342946-227342946 on Build GRCh38
rs752944748 Chr.2: 227342794-227342794 on Build GRCh38
rs1011056821 Chr.2: 227342831-227342831 on Build GRCh38
rs779734462 Chr.2: 227342832-227342832 on Build GRCh38
rs1022788094 Chr.2: 227343036-227343036 on Build GRCh38
rs973889520 Chr.2: 227342653-227342653 on Build GRCh38
rs77817189 Chr.2: 227343008-227343008 on Build GRCh38
rs755576850 Chr.2: 227342825-227342825 on Build GRCh38
rs761805211 Chr.2: 227342729-227342729 on Build GRCh38
rs939435641 Chr.2: 227342747-227342747 on Build GRCh38
rs201492539 Chr.2: 227342819-227342819 on Build GRCh38
rs752060091 Chr.2: 227342810-227342810 on Build GRCh38
rs1009430607 Chr.2: 227343114-227343114 on Build GRCh38
rs759646757 Chr.2: 227342753-227342753 on Build GRCh38
rs973843892 Chr.2: 227343029-227343029 on Build GRCh38
rs780736008 Chr.2: 227342701-227342701 on Build GRCh38
rs143777853 Chr.2: 227343053-227343053 on Build GRCh38
rs375100372 Chr.2: 227342848-227342850 on Build GRCh38
rs745321254 Chr.2: 227342703-227342703 on Build GRCh38
rs533603160 Chr.2: 227343067-227343069 on Build GRCh38
rs772077425 Chr.2: 227342845-227342845 on Build GRCh38
rs367621130 Chr.2: 227342811-227342811 on Build GRCh38
rs111634972 Chr.2: 227342784-227342784 on Build GRCh38
rs962452995 Chr.2: 227342934-227342934 on Build GRCh38
rs779701519 Chr.2: 227342652-227342652 on Build GRCh38
rs927975777 Chr.2: 227343067-227343067 on Build GRCh38
rs374544546 Chr.2: 227342728-227342728 on Build GRCh38
rs747841482 Chr.2: 227342842-227342842 on Build GRCh38
rs764512060 Chr.2: 227342801-227342801 on Build GRCh38
rs749105770 Chr.2: 227342725-227342725 on Build GRCh38
rs775088720 Chr.2: 227342717-227342717 on Build GRCh38
rs748908577 Chr.2: 227342834-227342834 on Build GRCh38
rs11688163 Chr.2: 227342895-227342895 on Build GRCh38
rs776864848 Chr.2: 227342736-227342736 on Build GRCh38
rs997539408 Chr.2: 227342880-227342880 on Build GRCh38
rs773233850 Chr.2: 227342848-227342848 on Build GRCh38
rs6740870 Chr.2: 227343060-227343060 on Build GRCh38
rs892526780 Chr.2: 227343100-227343100 on Build GRCh38
rs745416315 Chr.2: 227342820-227342820 on Build GRCh38
rs765887189 Chr.2: 227342673-227342673 on Build GRCh38
rs545146698 Chr.2: 227343042-227343042 on Build GRCh38
rs577397233 Chr.2: 227342931-227342931 on Build GRCh38
rs759739309 Chr.2: 227342849-227342849 on Build GRCh38
rs11675936 Chr.2: 227342804-227342804 on Build GRCh38
rs79229930 Chr.2: 227343007-227343007 on Build GRCh38
rs915804619 Chr.2: 227342674-227342674 on Build GRCh38
rs759976495 Chr.2: 227342723-227342724 on Build GRCh38
rs569706627 Chr.2: 227342683-227342683 on Build GRCh38
rs780896178 Chr.2: 227343024-227343024 on Build GRCh38
rs913949997 Chr.2: 227343122-227343122 on Build GRCh38
rs769445244 Chr.2: 227342714-227342714 on Build GRCh38
rs765376315 Chr.2: 227342863-227342863 on Build GRCh38
rs948653842 Chr.2: 227342697-227342697 on Build GRCh38
rs768479084 Chr.2: 227342727-227342727 on Build GRCh38
rs879261200 Chr.2: 227342651-227342651 on Build GRCh38
rs375796833 Chr.2: 227342839-227342839 on Build GRCh38
rs758873739 Chr.2: 227342799-227342799 on Build GRCh38
rs6738078 Chr.2: 227342909-227342909 on Build GRCh38
rs1047664323 Chr.2: 227342996-227342996 on Build GRCh38
rs910172331 Chr.2: 227343006-227343007 on Build GRCh38
rs1035034302 Chr.2: 227342684-227342684 on Build GRCh38
rs62279174 Chr.2: 227343126-227343126 on Build GRCh38
rs1003953035 Chr.2: 227342927-227342927 on Build GRCh38
rs775918563 Chr.2: 227342870-227342870 on Build GRCh38
rs766429439 Chr.2: 227342730-227342730 on Build GRCh38
rs192486184 Chr.2: 227343077-227343077 on Build GRCh38
rs368588231 Chr.2: 227342809-227342809 on Build GRCh38
rs1045251674 Chr.2: 227343051-227343051 on Build GRCh38
rs879838989 Chr.2: 227342995-227342996 on Build GRCh38
rs766986319 Chr.2: 227342768-227342769 on Build GRCh38
rs754195660 Chr.2: 227343089-227343089 on Build GRCh38
rs900046792 Chr.2: 227342878-227342878 on Build GRCh38
rs563432794 Chr.2: 227343049-227343049 on Build GRCh38
rs910215058 Chr.2: 227342860-227342860 on Build GRCh38
rs956578954 Chr.2: 227342790-227342790 on Build GRCh38
rs148986019 Chr.2: 227342675-227342675 on Build GRCh38

View Full Product Details