Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232456486-232456990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPI
Entrez Gene ID
248
Gene Name
alkaline phosphatase, intestinal
Gene Aliases
IAP
UniGene
Hs.37009
Gene Chromosome Location
Chr.2: 232456112-232460745 on Build GRCh38
Transcripts
NM_001631.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1692165 c.328G>A p.D110N Chr.2: 232456926-232456926 on Build GRCh38
3579082 c.362G>A p.G121E Chr.2: 232456960-232456960 on Build GRCh38
4965099 c.301-4G>A p.? Chr.2: 232456895-232456895 on Build GRCh38
6999431 c.334G>A p.A112T Chr.2: 232456932-232456932 on Build GRCh38
3720778 c.347C>T p.T116M Chr.2: 232456945-232456945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543094160 Chr.2: 232456571-232456571 on Build GRCh38
rs759187502 Chr.2: 232456810-232456810 on Build GRCh38
rs777135488 Chr.2: 232456882-232456882 on Build GRCh38
rs201149811 Chr.2: 232456680-232456680 on Build GRCh38
rs757859054 Chr.2: 232456604-232456604 on Build GRCh38
rs745727808 Chr.2: 232456631-232456631 on Build GRCh38
rs746380280 Chr.2: 232456952-232456952 on Build GRCh38
rs747070491 Chr.2: 232456670-232456670 on Build GRCh38
rs566677043 Chr.2: 232456886-232456886 on Build GRCh38
rs375512663 Chr.2: 232456719-232456719 on Build GRCh38
rs974771004 Chr.2: 232456930-232456930 on Build GRCh38
rs758698499 Chr.2: 232456867-232456867 on Build GRCh38
rs200694055 Chr.2: 232456863-232456863 on Build GRCh38
rs761853515 Chr.2: 232456584-232456584 on Build GRCh38
rs567673319 Chr.2: 232456846-232456846 on Build GRCh38
rs184809548 Chr.2: 232456894-232456894 on Build GRCh38
rs370515857 Chr.2: 232456864-232456864 on Build GRCh38
rs751723259 Chr.2: 232456934-232456934 on Build GRCh38
rs545317166 Chr.2: 232456750-232456750 on Build GRCh38
rs374217058 Chr.2: 232456869-232456869 on Build GRCh38
rs750645792 Chr.2: 232456607-232456607 on Build GRCh38
rs199922420 Chr.2: 232456891-232456891 on Build GRCh38
rs779020997 Chr.2: 232456652-232456652 on Build GRCh38
rs148744505 Chr.2: 232456961-232456961 on Build GRCh38
rs752381527 Chr.2: 232456596-232456596 on Build GRCh38
rs778025369 Chr.2: 232456933-232456933 on Build GRCh38
rs772076928 Chr.2: 232456661-232456661 on Build GRCh38
rs764394136 Chr.2: 232456957-232456957 on Build GRCh38
rs565071856 Chr.2: 232456814-232456814 on Build GRCh38
rs781205092 Chr.2: 232456946-232456946 on Build GRCh38
rs145841277 Chr.2: 232456586-232456586 on Build GRCh38
rs145699808 Chr.2: 232456595-232456595 on Build GRCh38
rs770421872 Chr.2: 232456615-232456615 on Build GRCh38
rs781090459 Chr.2: 232456550-232456550 on Build GRCh38
rs920492047 Chr.2: 232456588-232456588 on Build GRCh38
rs775006273 Chr.2: 232456720-232456720 on Build GRCh38
rs150302120 Chr.2: 232456689-232456689 on Build GRCh38
rs770379151 Chr.2: 232456958-232456958 on Build GRCh38
rs541317610 Chr.2: 232456959-232456959 on Build GRCh38
rs758615388 Chr.2: 232456967-232456967 on Build GRCh38
rs1013955699 Chr.2: 232456765-232456765 on Build GRCh38
rs146038441 Chr.2: 232456908-232456908 on Build GRCh38
rs974055526 Chr.2: 232456883-232456883 on Build GRCh38
rs762622182 Chr.2: 232456723-232456723 on Build GRCh38
rs752188879 Chr.2: 232456515-232456515 on Build GRCh38
rs759862463 Chr.2: 232456594-232456594 on Build GRCh38
rs976253861 Chr.2: 232456955-232456955 on Build GRCh38
rs61732024 Chr.2: 232456669-232456669 on Build GRCh38
rs904245994 Chr.2: 232456721-232456721 on Build GRCh38
rs368815505 Chr.2: 232456611-232456611 on Build GRCh38
rs60677335 Chr.2: 232456801-232456801 on Build GRCh38
rs71421649 Chr.2: 232456646-232456646 on Build GRCh38
rs774137247 Chr.2: 232456895-232456895 on Build GRCh38
rs777726761 Chr.2: 232456510-232456510 on Build GRCh38
rs765285295 Chr.2: 232456590-232456602 on Build GRCh38
rs751573907 Chr.2: 232456612-232456612 on Build GRCh38
rs774633391 Chr.2: 232456569-232456569 on Build GRCh38
rs770082201 Chr.2: 232456890-232456890 on Build GRCh38
rs377393587 Chr.2: 232456892-232456892 on Build GRCh38
rs761424272 Chr.2: 232456906-232456906 on Build GRCh38
rs746902902 Chr.2: 232456535-232456535 on Build GRCh38
rs767623702 Chr.2: 232456738-232456738 on Build GRCh38
rs750421707 Chr.2: 232456911-232456911 on Build GRCh38
rs757332360 Chr.2: 232456945-232456945 on Build GRCh38
rs546698962 Chr.2: 232456868-232456868 on Build GRCh38
rs558991529 Chr.2: 232456516-232456516 on Build GRCh38
rs756826696 Chr.2: 232456737-232456737 on Build GRCh38
rs550725974 Chr.2: 232456842-232456842 on Build GRCh38
rs759950805 Chr.2: 232456692-232456692 on Build GRCh38
rs201896655 Chr.2: 232456684-232456684 on Build GRCh38
rs530918095 Chr.2: 232456816-232456816 on Build GRCh38
rs376171344 Chr.2: 232456565-232456565 on Build GRCh38
rs61732032 Chr.2: 232456931-232456931 on Build GRCh38
rs781250451 Chr.2: 232456619-232456619 on Build GRCh38
rs371281193 Chr.2: 232456584-232456584 on Build GRCh38
rs376854130 Chr.2: 232456862-232456862 on Build GRCh38
rs763531821 Chr.2: 232456733-232456733 on Build GRCh38
rs142092116 Chr.2: 232456744-232456744 on Build GRCh38
rs983346861 Chr.2: 232456896-232456896 on Build GRCh38
rs376557863 Chr.2: 232456509-232456509 on Build GRCh38
rs760613268 Chr.2: 232456919-232456919 on Build GRCh38
rs911873047 Chr.2: 232456654-232456654 on Build GRCh38
rs371458940 Chr.2: 232456587-232456587 on Build GRCh38
rs113267808 Chr.2: 232456551-232456551 on Build GRCh38
rs766754479 Chr.2: 232456591-232456591 on Build GRCh38
rs778604684 Chr.2: 232456514-232456514 on Build GRCh38
rs1031357012 Chr.2: 232456537-232456537 on Build GRCh38
rs761521664 Chr.2: 232456968-232456968 on Build GRCh38
rs371086927 Chr.2: 232456932-232456932 on Build GRCh38
rs1048012688 Chr.2: 232456603-232456603 on Build GRCh38
rs900759974 Chr.2: 232456675-232456675 on Build GRCh38
rs142586150 Chr.2: 232456653-232456653 on Build GRCh38
rs768847599 Chr.2: 232456962-232456962 on Build GRCh38
rs181077741 Chr.2: 232456735-232456735 on Build GRCh38
rs755963042 Chr.2: 232456637-232456637 on Build GRCh38
rs757210358 Chr.2: 232456616-232456616 on Build GRCh38
rs1043933709 Chr.2: 232456707-232456707 on Build GRCh38
rs777628025 Chr.2: 232456517-232456517 on Build GRCh38
rs774155707 Chr.2: 232456964-232456964 on Build GRCh38
rs868198329 Chr.2: 232456701-232456701 on Build GRCh38
rs771730709 Chr.2: 232456969-232456969 on Build GRCh38
rs765578422 Chr.2: 232456704-232456704 on Build GRCh38
rs369841817 Chr.2: 232456543-232456543 on Build GRCh38

View Full Product Details