Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 234492962-234493467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL4C
Entrez Gene ID
10123
Gene Name
ADP ribosylation factor like GTPase 4C
Gene Aliases
ARL7, LAK
UniGene
Hs.111554
Gene Chromosome Location
Chr.2: 234493041-234497053 on Build GRCh38
Transcripts
NM_001282431.1 NM_005737.3

SNP Detail

SNP ID Chromosome Location
rs575707191 Chr.2: 234492998-234492999 on Build GRCh38
rs759172572 Chr.2: 234493185-234493185 on Build GRCh38
rs181857551 Chr.2: 234492997-234492997 on Build GRCh38
rs940709656 Chr.2: 234493430-234493430 on Build GRCh38
rs564449925 Chr.2: 234493162-234493162 on Build GRCh38
rs895258283 Chr.2: 234493169-234493169 on Build GRCh38
rs968781745 Chr.2: 234493007-234493007 on Build GRCh38
rs920218235 Chr.2: 234493242-234493242 on Build GRCh38
rs1049618097 Chr.2: 234493428-234493428 on Build GRCh38
rs996369660 Chr.2: 234493283-234493283 on Build GRCh38
rs1019082754 Chr.2: 234492989-234492989 on Build GRCh38
rs545303578 Chr.2: 234493352-234493352 on Build GRCh38
rs749713268 Chr.2: 234493306-234493306 on Build GRCh38
rs397960087 Chr.2: 234493005-234493006 on Build GRCh38
rs569485811 Chr.2: 234493163-234493163 on Build GRCh38
rs533401166 Chr.2: 234493332-234493332 on Build GRCh38
rs763601192 Chr.2: 234493440-234493441 on Build GRCh38
rs900095928 Chr.2: 234493307-234493307 on Build GRCh38
rs775887575 Chr.2: 234493090-234493094 on Build GRCh38
rs1005154578 Chr.2: 234492988-234492988 on Build GRCh38
rs201917159 Chr.2: 234493006-234493006 on Build GRCh38
rs34977209 Chr.2: 234493164-234493165 on Build GRCh38
rs1022015814 Chr.2: 234493006-234493006 on Build GRCh38
rs75179573 Chr.2: 234493268-234493268 on Build GRCh38
rs886439915 Chr.2: 234493238-234493238 on Build GRCh38
rs939171542 Chr.2: 234493352-234493352 on Build GRCh38
rs1055616911 Chr.2: 234493151-234493151 on Build GRCh38
rs989654290 Chr.2: 234493300-234493300 on Build GRCh38
rs993056855 Chr.2: 234493126-234493126 on Build GRCh38
rs774550553 Chr.2: 234493405-234493405 on Build GRCh38
rs769239731 Chr.2: 234493397-234493397 on Build GRCh38
rs945271585 Chr.2: 234493037-234493043 on Build GRCh38
rs745364388 Chr.2: 234493177-234493177 on Build GRCh38
rs372227136 Chr.2: 234493357-234493357 on Build GRCh38
rs558853201 Chr.2: 234493115-234493115 on Build GRCh38
rs998856566 Chr.2: 234493033-234493033 on Build GRCh38
rs397727215 Chr.2: 234493004-234493005 on Build GRCh38
rs997473973 Chr.2: 234493321-234493321 on Build GRCh38
rs1042288983 Chr.2: 234493049-234493049 on Build GRCh38
rs886431393 Chr.2: 234493429-234493429 on Build GRCh38
rs900065223 Chr.2: 234492995-234492995 on Build GRCh38
rs959898441 Chr.2: 234493097-234493097 on Build GRCh38
rs533926856 Chr.2: 234493276-234493276 on Build GRCh38
rs966358409 Chr.2: 234493331-234493331 on Build GRCh38
rs12055 Chr.2: 234493266-234493266 on Build GRCh38
rs989191111 Chr.2: 234493011-234493011 on Build GRCh38
rs868430937 Chr.2: 234493102-234493102 on Build GRCh38
rs539344985 Chr.2: 234493051-234493051 on Build GRCh38
rs921060420 Chr.2: 234492999-234492999 on Build GRCh38
rs973102848 Chr.2: 234493241-234493241 on Build GRCh38
rs11386401 Chr.2: 234492999-234493000 on Build GRCh38
rs930704461 Chr.2: 234493423-234493423 on Build GRCh38
rs904571165 Chr.2: 234493337-234493337 on Build GRCh38
rs147285577 Chr.2: 234493267-234493267 on Build GRCh38
rs1031714355 Chr.2: 234493072-234493072 on Build GRCh38
rs557300861 Chr.2: 234493101-234493101 on Build GRCh38
rs939223800 Chr.2: 234493106-234493106 on Build GRCh38
rs779728776 Chr.2: 234493288-234493288 on Build GRCh38
rs369235982 Chr.2: 234493297-234493297 on Build GRCh38
rs35206193 Chr.2: 234493292-234493293 on Build GRCh38
rs928955594 Chr.2: 234493253-234493253 on Build GRCh38
rs573026655 Chr.2: 234493335-234493335 on Build GRCh38

View Full Product Details