Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 237340748-237341249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A3
Entrez Gene ID
1293
Gene Name
collagen type VI alpha 3 chain
Gene Aliases
BTHLM1, DYT27, UCMD1
UniGene
Hs.233240
Gene Chromosome Location
Chr.2: 237324012-237434822 on Build GRCh38
Transcripts
NM_004369.3 NM_057166.4 NM_057167.3 XM_005246065.1 XM_005246066.1 XM_006712253.1 XM_011510574.1 XM_017003303.1 XM_017003304.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
477135 c.7881T>G p.A2627A Chr.2: 237341035-237341035 on Build GRCh38
122301 c.7943A>G p.Q2648R Chr.2: 237340973-237340973 on Build GRCh38
4558881 c.7848G>A p.V2616V Chr.2: 237341068-237341068 on Build GRCh38
720111 c.7780G>A p.D2594N Chr.2: 237341136-237341136 on Build GRCh38
3579584 c.8063C>T p.S2688F Chr.2: 237340853-237340853 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781775472 Chr.2: 237340992-237340992 on Build GRCh38
rs775967180 Chr.2: 237341076-237341076 on Build GRCh38
rs145136426 Chr.2: 237341038-237341038 on Build GRCh38
rs747976144 Chr.2: 237341009-237341009 on Build GRCh38
rs34558385 Chr.2: 237341074-237341074 on Build GRCh38
rs765987305 Chr.2: 237340893-237340893 on Build GRCh38
rs886044041 Chr.2: 237340940-237340940 on Build GRCh38
rs973630902 Chr.2: 237341121-237341121 on Build GRCh38
rs760154245 Chr.2: 237340956-237340956 on Build GRCh38
rs766954929 Chr.2: 237340878-237340878 on Build GRCh38
rs1053569255 Chr.2: 237341104-237341104 on Build GRCh38
rs963639269 Chr.2: 237341116-237341116 on Build GRCh38
rs755777148 Chr.2: 237340772-237340772 on Build GRCh38
rs145884404 Chr.2: 237341073-237341073 on Build GRCh38
rs753981395 Chr.2: 237341066-237341066 on Build GRCh38
rs765014790 Chr.2: 237341039-237341039 on Build GRCh38
rs551116650 Chr.2: 237340912-237340912 on Build GRCh38
rs957705259 Chr.2: 237340802-237340802 on Build GRCh38
rs755750913 Chr.2: 237340886-237340886 on Build GRCh38
rs754664272 Chr.2: 237340986-237340986 on Build GRCh38
rs1037630319 Chr.2: 237340869-237340869 on Build GRCh38
rs767471920 Chr.2: 237340953-237340953 on Build GRCh38
rs759129620 Chr.2: 237341078-237341078 on Build GRCh38
rs142851023 Chr.2: 237340907-237340907 on Build GRCh38
rs749957183 Chr.2: 237340880-237340880 on Build GRCh38
rs886044342 Chr.2: 237340884-237340884 on Build GRCh38
rs890268146 Chr.2: 237340968-237340968 on Build GRCh38
rs771974295 Chr.2: 237341011-237341011 on Build GRCh38
rs35288868 Chr.2: 237341076-237341077 on Build GRCh38
rs1031349800 Chr.2: 237340789-237340789 on Build GRCh38
rs780804543 Chr.2: 237341200-237341200 on Build GRCh38
rs921987199 Chr.2: 237340879-237340879 on Build GRCh38
rs771203220 Chr.2: 237340806-237340806 on Build GRCh38
rs115757876 Chr.2: 237340819-237340819 on Build GRCh38
rs761740861 Chr.2: 237340871-237340871 on Build GRCh38
rs148017477 Chr.2: 237340899-237340899 on Build GRCh38
rs750826266 Chr.2: 237341058-237341058 on Build GRCh38
rs111595697 Chr.2: 237340988-237340988 on Build GRCh38
rs779104216 Chr.2: 237341144-237341144 on Build GRCh38
rs775630107 Chr.2: 237340926-237340926 on Build GRCh38
rs745793481 Chr.2: 237341164-237341164 on Build GRCh38
rs761045227 Chr.2: 237341131-237341131 on Build GRCh38
rs747926608 Chr.2: 237341072-237341072 on Build GRCh38
rs758750067 Chr.2: 237340901-237340901 on Build GRCh38
rs774092134 Chr.2: 237341006-237341006 on Build GRCh38
rs762702382 Chr.2: 237340828-237340828 on Build GRCh38
rs768129302 Chr.2: 237341056-237341056 on Build GRCh38
rs377679398 Chr.2: 237341013-237341013 on Build GRCh38
rs745354138 Chr.2: 237341225-237341225 on Build GRCh38
rs372278071 Chr.2: 237341148-237341148 on Build GRCh38
rs973495446 Chr.2: 237341005-237341005 on Build GRCh38
rs535306425 Chr.2: 237341083-237341083 on Build GRCh38
rs150554876 Chr.2: 237341108-237341108 on Build GRCh38
rs398124135 Chr.2: 237340908-237340908 on Build GRCh38
rs138285547 Chr.2: 237341067-237341067 on Build GRCh38
rs987550949 Chr.2: 237341177-237341177 on Build GRCh38
rs777305410 Chr.2: 237341046-237341046 on Build GRCh38
rs775318733 Chr.2: 237340825-237340825 on Build GRCh38
rs780182250 Chr.2: 237340996-237340996 on Build GRCh38
rs779154094 Chr.2: 237341070-237341070 on Build GRCh38
rs886042623 Chr.2: 237341119-237341120 on Build GRCh38
rs765740728 Chr.2: 237341034-237341034 on Build GRCh38
rs149403952 Chr.2: 237340894-237340894 on Build GRCh38
rs528369978 Chr.2: 237340909-237340909 on Build GRCh38
rs886044691 Chr.2: 237341008-237341008 on Build GRCh38
rs748491825 Chr.2: 237340776-237340776 on Build GRCh38
rs80193928 Chr.2: 237340921-237340921 on Build GRCh38
rs902231679 Chr.2: 237340915-237340915 on Build GRCh38
rs755107410 Chr.2: 237341139-237341139 on Build GRCh38
rs751945310 Chr.2: 237341041-237341041 on Build GRCh38
rs369537287 Chr.2: 237340963-237340963 on Build GRCh38
rs776220123 Chr.2: 237341037-237341037 on Build GRCh38
rs984056484 Chr.2: 237341097-237341097 on Build GRCh38
rs753501207 Chr.2: 237340965-237340965 on Build GRCh38
rs767932907 Chr.2: 237341117-237341117 on Build GRCh38
rs879503815 Chr.2: 237341223-237341223 on Build GRCh38
rs779948294 Chr.2: 237341062-237341062 on Build GRCh38
rs530561703 Chr.2: 237340952-237340952 on Build GRCh38
rs114806654 Chr.2: 237341084-237341084 on Build GRCh38
rs112104700 Chr.2: 237341043-237341043 on Build GRCh38
rs772602377 Chr.2: 237340779-237340779 on Build GRCh38
rs751561315 Chr.2: 237340842-237340842 on Build GRCh38
rs778195223 Chr.2: 237340794-237340794 on Build GRCh38
rs777383519 Chr.2: 237341169-237341169 on Build GRCh38
rs35763271 Chr.2: 237340771-237340771 on Build GRCh38
rs750819402 Chr.2: 237341122-237341122 on Build GRCh38
rs770746208 Chr.2: 237341189-237341189 on Build GRCh38
rs75349214 Chr.2: 237340919-237340919 on Build GRCh38
rs747073702 Chr.2: 237340804-237340804 on Build GRCh38
rs749408852 Chr.2: 237340999-237340999 on Build GRCh38
rs960490247 Chr.2: 237340885-237340885 on Build GRCh38
rs746855574 Chr.2: 237341173-237341173 on Build GRCh38
rs766947740 Chr.2: 237341136-237341136 on Build GRCh38
rs756654349 Chr.2: 237341061-237341061 on Build GRCh38
rs536700038 Chr.2: 237340851-237340851 on Build GRCh38
rs375222168 Chr.2: 237340933-237340933 on Build GRCh38
rs886055803 Chr.2: 237340866-237340866 on Build GRCh38
rs575544658 Chr.2: 237341197-237341197 on Build GRCh38
rs757801355 Chr.2: 237340989-237340989 on Build GRCh38
rs144249704 Chr.2: 237341137-237341137 on Build GRCh38
rs79313758 Chr.2: 237340906-237340906 on Build GRCh38
rs751453783 Chr.2: 237340818-237340818 on Build GRCh38
rs907941419 Chr.2: 237340815-237340815 on Build GRCh38
rs368811075 Chr.2: 237341191-237341191 on Build GRCh38
rs776858630 Chr.2: 237340818-237340818 on Build GRCh38
rs772676484 Chr.2: 237340954-237340954 on Build GRCh38
rs922019493 Chr.2: 237340911-237340911 on Build GRCh38
rs4433949 Chr.2: 237340987-237340987 on Build GRCh38
rs760230894 Chr.2: 237341031-237341031 on Build GRCh38
rs768944937 Chr.2: 237341002-237341002 on Build GRCh38
rs886042689 Chr.2: 237340843-237340843 on Build GRCh38
rs759414368 Chr.2: 237341182-237341183 on Build GRCh38
rs756212605 Chr.2: 237340993-237340993 on Build GRCh38
rs371571488 Chr.2: 237340841-237340841 on Build GRCh38
rs372793439 Chr.2: 237341194-237341194 on Build GRCh38
rs969160751 Chr.2: 237340834-237340834 on Build GRCh38
rs773192776 Chr.2: 237341018-237341018 on Build GRCh38
rs779286281 Chr.2: 237340775-237340775 on Build GRCh38
rs1024862202 Chr.2: 237340964-237340964 on Build GRCh38
rs749100905 Chr.2: 237340932-237340932 on Build GRCh38

View Full Product Details