Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 240596212-240596704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN10
Entrez Gene ID
11132
Gene Name
calpain 10
Gene Aliases
CANP10, NIDDM1
UniGene
Hs.728234
Gene Chromosome Location
Chr.2: 240586716-240599109 on Build GRCh38
Transcripts
NM_023083.3 NM_023085.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3380105 c.1464C>G p.T488T Chr.2: 240596504-240596504 on Build GRCh38
4834049 c.1353C>T p.Y451Y Chr.2: 240596393-240596393 on Build GRCh38
6286759 c.1348G>T p.A450S Chr.2: 240596388-240596388 on Build GRCh38
4092877 c.1483G>A p.A495T Chr.2: 240596682-240596682 on Build GRCh38
2909699 c.1355A>G p.D452G Chr.2: 240596395-240596395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202215064 Chr.2: 240596536-240596536 on Build GRCh38
rs1034621952 Chr.2: 240596524-240596524 on Build GRCh38
rs904726596 Chr.2: 240596487-240596487 on Build GRCh38
rs751132481 Chr.2: 240596547-240596547 on Build GRCh38
rs754583407 Chr.2: 240596468-240596468 on Build GRCh38
rs2975765 Chr.2: 240596380-240596380 on Build GRCh38
rs918695213 Chr.2: 240596390-240596390 on Build GRCh38
rs756668839 Chr.2: 240596483-240596483 on Build GRCh38
rs777520933 Chr.2: 240596653-240596653 on Build GRCh38
rs771284091 Chr.2: 240596397-240596397 on Build GRCh38
rs764755613 Chr.2: 240596535-240596535 on Build GRCh38
rs758093042 Chr.2: 240596538-240596538 on Build GRCh38
rs748756061 Chr.2: 240596328-240596328 on Build GRCh38
rs773316366 Chr.2: 240596434-240596434 on Build GRCh38
rs929138796 Chr.2: 240596235-240596235 on Build GRCh38
rs368588347 Chr.2: 240596471-240596471 on Build GRCh38
rs1039880585 Chr.2: 240596571-240596571 on Build GRCh38
rs774232664 Chr.2: 240596428-240596428 on Build GRCh38
rs548546733 Chr.2: 240596387-240596387 on Build GRCh38
rs553967526 Chr.2: 240596630-240596630 on Build GRCh38
rs754709387 Chr.2: 240596383-240596383 on Build GRCh38
rs752192395 Chr.2: 240596509-240596509 on Build GRCh38
rs747760729 Chr.2: 240596393-240596393 on Build GRCh38
rs757272199 Chr.2: 240596304-240596304 on Build GRCh38
rs375653976 Chr.2: 240596379-240596379 on Build GRCh38
rs761970401 Chr.2: 240596429-240596429 on Build GRCh38
rs534379076 Chr.2: 240596505-240596505 on Build GRCh38
rs780388987 Chr.2: 240596484-240596484 on Build GRCh38
rs759741817 Chr.2: 240596285-240596285 on Build GRCh38
rs200003517 Chr.2: 240596335-240596335 on Build GRCh38
rs201807668 Chr.2: 240596332-240596332 on Build GRCh38
rs770943505 Chr.2: 240596670-240596670 on Build GRCh38
rs374143083 Chr.2: 240596412-240596412 on Build GRCh38
rs930006352 Chr.2: 240596392-240596392 on Build GRCh38
rs759430755 Chr.2: 240596682-240596682 on Build GRCh38
rs775131440 Chr.2: 240596268-240596268 on Build GRCh38
rs909939907 Chr.2: 240596560-240596560 on Build GRCh38
rs780556583 Chr.2: 240596554-240596554 on Build GRCh38
rs538913310 Chr.2: 240596515-240596515 on Build GRCh38
rs777102177 Chr.2: 240596284-240596284 on Build GRCh38
rs1047535074 Chr.2: 240596257-240596257 on Build GRCh38
rs776914728 Chr.2: 240596338-240596338 on Build GRCh38
rs767517219 Chr.2: 240596431-240596431 on Build GRCh38
rs763272807 Chr.2: 240596356-240596356 on Build GRCh38
rs1055475743 Chr.2: 240596234-240596234 on Build GRCh38
rs200314283 Chr.2: 240596375-240596375 on Build GRCh38
rs756285083 Chr.2: 240596283-240596284 on Build GRCh38
rs980412388 Chr.2: 240596344-240596344 on Build GRCh38
rs765450449 Chr.2: 240596291-240596291 on Build GRCh38
rs752717259 Chr.2: 240596294-240596294 on Build GRCh38
rs376384709 Chr.2: 240596408-240596408 on Build GRCh38
rs373039172 Chr.2: 240596295-240596295 on Build GRCh38
rs367919148 Chr.2: 240596549-240596549 on Build GRCh38
rs868763560 Chr.2: 240596354-240596354 on Build GRCh38
rs868447092 Chr.2: 240596684-240596684 on Build GRCh38
rs751715639 Chr.2: 240596366-240596366 on Build GRCh38
rs747090224 Chr.2: 240596659-240596659 on Build GRCh38
rs1020715828 Chr.2: 240596334-240596334 on Build GRCh38
rs375878738 Chr.2: 240596533-240596533 on Build GRCh38
rs1000192409 Chr.2: 240596646-240596646 on Build GRCh38
rs1010378179 Chr.2: 240596308-240596308 on Build GRCh38
rs766256299 Chr.2: 240596457-240596457 on Build GRCh38
rs749530877 Chr.2: 240596533-240596548 on Build GRCh38
rs760677320 Chr.2: 240596531-240596531 on Build GRCh38
rs751423611 Chr.2: 240596303-240596303 on Build GRCh38
rs750305591 Chr.2: 240596313-240596313 on Build GRCh38
rs777996904 Chr.2: 240596654-240596654 on Build GRCh38
rs775718991 Chr.2: 240596351-240596351 on Build GRCh38
rs1048029128 Chr.2: 240596398-240596398 on Build GRCh38
rs184439941 Chr.2: 240596418-240596418 on Build GRCh38
rs529302384 Chr.2: 240596381-240596381 on Build GRCh38
rs371296378 Chr.2: 240596477-240596477 on Build GRCh38
rs758832246 Chr.2: 240596367-240596367 on Build GRCh38
rs760736608 Chr.2: 240596449-240596449 on Build GRCh38
rs1030065769 Chr.2: 240596296-240596296 on Build GRCh38
rs781679015 Chr.2: 240596406-240596406 on Build GRCh38
rs901310671 Chr.2: 240596608-240596608 on Build GRCh38
rs375179542 Chr.2: 240596310-240596310 on Build GRCh38
rs768985963 Chr.2: 240596423-240596423 on Build GRCh38
rs748382378 Chr.2: 240596637-240596637 on Build GRCh38
rs202074382 Chr.2: 240596534-240596534 on Build GRCh38
rs2953171 Chr.2: 240596636-240596636 on Build GRCh38
rs764055347 Chr.2: 240596299-240596299 on Build GRCh38
rs768080984 Chr.2: 240596329-240596329 on Build GRCh38
rs769954158 Chr.2: 240596350-240596350 on Build GRCh38
rs755756511 Chr.2: 240596325-240596325 on Build GRCh38
rs114535069 Chr.2: 240596460-240596460 on Build GRCh38
rs377454835 Chr.2: 240596618-240596618 on Build GRCh38
rs201984748 Chr.2: 240596491-240596491 on Build GRCh38
rs774659955 Chr.2: 240596504-240596504 on Build GRCh38
rs773475963 Chr.2: 240596529-240596529 on Build GRCh38
rs749570105 Chr.2: 240596555-240596555 on Build GRCh38
rs148303768 Chr.2: 240596469-240596469 on Build GRCh38
rs752063033 Chr.2: 240596413-240596413 on Build GRCh38
rs765606395 Chr.2: 240596289-240596289 on Build GRCh38
rs916988137 Chr.2: 240596267-240596267 on Build GRCh38
rs369499868 Chr.2: 240596283-240596283 on Build GRCh38
rs370360650 Chr.2: 240596378-240596378 on Build GRCh38
rs370774447 Chr.2: 240596409-240596409 on Build GRCh38
rs759907402 Chr.2: 240596339-240596339 on Build GRCh38
rs369837274 Chr.2: 240596331-240596331 on Build GRCh38
rs372030642 Chr.2: 240596269-240596269 on Build GRCh38
rs768706621 Chr.2: 240596502-240596502 on Build GRCh38
rs776661578 Chr.2: 240596681-240596681 on Build GRCh38
rs764054638 Chr.2: 240596363-240596363 on Build GRCh38
rs761058925 Chr.2: 240596282-240596282 on Build GRCh38
rs373123426 Chr.2: 240596384-240596384 on Build GRCh38
rs1007031224 Chr.2: 240596302-240596302 on Build GRCh38
rs777656879 Chr.2: 240596360-240596361 on Build GRCh38
rs375186294 Chr.2: 240596280-240596280 on Build GRCh38
rs1051351442 Chr.2: 240596470-240596470 on Build GRCh38
rs200700918 Chr.2: 240596521-240596521 on Build GRCh38

View Full Product Details