Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241750064-241750567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
D2HGDH
Entrez Gene ID
728294
Gene Name
D-2-hydroxyglutarate dehydrogenase
Gene Aliases
D2HGD
UniGene
Hs.516813
Gene Chromosome Location
Chr.2: 241734579-241768816 on Build GRCh38
Transcripts
NM_001287249.1 NM_152783.4 NR_109778.1 XM_011511734.1 XM_011511735.1 XM_011511736.1 XM_011511737.2 XM_011511738.2 XM_011511743.1 XM_011511744.1 XM_011511745.2 XM_011511749.2 XM_011511750.2 XM_011511751.1 XM_011511753.2 XM_011511754.1 XM_011511756.1 XM_011511757.2 XM_011511759.2 XM_017004828.1 XM_017004829.1 XM_017004830.1 XM_017004831.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5728143 c.931G>A p.E311K Chr.2: 241750228-241750228 on Build GRCh38
3391636 c.854-8C>T p.? Chr.2: 241750143-241750143 on Build GRCh38
6317836 c.854G>T p.G285V Chr.2: 241750151-241750151 on Build GRCh38
6091147 c.963C>G p.V321V Chr.2: 241750260-241750260 on Build GRCh38
3580145 c.975C>T p.L325L Chr.2: 241750272-241750272 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs4675882 Chr.2: 241750542-241750542 on Build GRCh38
rs533922628 Chr.2: 241750384-241750384 on Build GRCh38
rs750661147 Chr.2: 241750322-241750322 on Build GRCh38
rs760161842 Chr.2: 241750235-241750235 on Build GRCh38
rs1028700535 Chr.2: 241750092-241750092 on Build GRCh38
rs143405918 Chr.2: 241750291-241750291 on Build GRCh38
rs761375145 Chr.2: 241750106-241750106 on Build GRCh38
rs373721818 Chr.2: 241750344-241750344 on Build GRCh38
rs774061348 Chr.2: 241750218-241750218 on Build GRCh38
rs762202718 Chr.2: 241750317-241750317 on Build GRCh38
rs867428268 Chr.2: 241750182-241750182 on Build GRCh38
rs907800320 Chr.2: 241750478-241750478 on Build GRCh38
rs749051812 Chr.2: 241750204-241750204 on Build GRCh38
rs771622834 Chr.2: 241750227-241750227 on Build GRCh38
rs772733736 Chr.2: 241750228-241750228 on Build GRCh38
rs750536429 Chr.2: 241750288-241750288 on Build GRCh38
rs761427826 Chr.2: 241750221-241750221 on Build GRCh38
rs758093567 Chr.2: 241750335-241750335 on Build GRCh38
rs754386160 Chr.2: 241750118-241750118 on Build GRCh38
rs141475702 Chr.2: 241750190-241750190 on Build GRCh38
rs961915115 Chr.2: 241750484-241750484 on Build GRCh38
rs757574221 Chr.2: 241750286-241750286 on Build GRCh38
rs753004727 Chr.2: 241750134-241750134 on Build GRCh38
rs553730051 Chr.2: 241750475-241750475 on Build GRCh38
rs368579744 Chr.2: 241750309-241750309 on Build GRCh38
rs772791154 Chr.2: 241750307-241750307 on Build GRCh38
rs371493676 Chr.2: 241750312-241750313 on Build GRCh38
rs764644757 Chr.2: 241750314-241750314 on Build GRCh38
rs370839952 Chr.2: 241750132-241750132 on Build GRCh38
rs756216496 Chr.2: 241750324-241750324 on Build GRCh38
rs187567086 Chr.2: 241750439-241750439 on Build GRCh38
rs754773784 Chr.2: 241750302-241750302 on Build GRCh38
rs533576337 Chr.2: 241750361-241750361 on Build GRCh38
rs370039160 Chr.2: 241750306-241750306 on Build GRCh38
rs758172973 Chr.2: 241750337-241750337 on Build GRCh38
rs963098589 Chr.2: 241750409-241750409 on Build GRCh38
rs1008887378 Chr.2: 241750109-241750109 on Build GRCh38
rs779764333 Chr.2: 241750343-241750344 on Build GRCh38
rs763494373 Chr.2: 241750312-241750312 on Build GRCh38
rs779976615 Chr.2: 241750196-241750196 on Build GRCh38
rs976747238 Chr.2: 241750528-241750528 on Build GRCh38
rs768199463 Chr.2: 241750103-241750103 on Build GRCh38
rs957762107 Chr.2: 241750208-241750208 on Build GRCh38
rs764487647 Chr.2: 241750264-241750264 on Build GRCh38
rs749247772 Chr.2: 241750300-241750300 on Build GRCh38
rs770396692 Chr.2: 241750345-241750345 on Build GRCh38
rs529526867 Chr.2: 241750305-241750305 on Build GRCh38
rs774886115 Chr.2: 241750315-241750315 on Build GRCh38
rs367678493 Chr.2: 241750469-241750469 on Build GRCh38
rs751827106 Chr.2: 241750155-241750155 on Build GRCh38
rs1023158117 Chr.2: 241750450-241750450 on Build GRCh38
rs781397354 Chr.2: 241750185-241750185 on Build GRCh38
rs540361877 Chr.2: 241750532-241750532 on Build GRCh38
rs111252594 Chr.2: 241750394-241750394 on Build GRCh38
rs141524359 Chr.2: 241750260-241750260 on Build GRCh38
rs112789889 Chr.2: 241750333-241750333 on Build GRCh38
rs931424565 Chr.2: 241750497-241750497 on Build GRCh38
rs779915637 Chr.2: 241750292-241750292 on Build GRCh38
rs1018387852 Chr.2: 241750406-241750406 on Build GRCh38
rs767013987 Chr.2: 241750117-241750117 on Build GRCh38
rs767867548 Chr.2: 241750320-241750320 on Build GRCh38
rs988069320 Chr.2: 241750265-241750265 on Build GRCh38
rs768485205 Chr.2: 241750212-241750212 on Build GRCh38
rs34802755 Chr.2: 241750448-241750448 on Build GRCh38
rs551272234 Chr.2: 241750326-241750326 on Build GRCh38
rs957792799 Chr.2: 241750422-241750422 on Build GRCh38
rs572979011 Chr.2: 241750485-241750485 on Build GRCh38
rs910276236 Chr.2: 241750304-241750304 on Build GRCh38
rs375093289 Chr.2: 241750328-241750328 on Build GRCh38
rs763289689 Chr.2: 241750263-241750263 on Build GRCh38
rs1023042150 Chr.2: 241750156-241750156 on Build GRCh38
rs956064915 Chr.2: 241750488-241750488 on Build GRCh38
rs71430381 Chr.2: 241750418-241750418 on Build GRCh38
rs769653234 Chr.2: 241750193-241750193 on Build GRCh38
rs777464269 Chr.2: 241750341-241750341 on Build GRCh38
rs548980299 Chr.2: 241750388-241750388 on Build GRCh38
rs566564994 Chr.2: 241750383-241750383 on Build GRCh38
rs556423385 Chr.2: 241750443-241750443 on Build GRCh38
rs372767424 Chr.2: 241750342-241750342 on Build GRCh38
rs34815577 Chr.2: 241750413-241750414 on Build GRCh38
rs778785432 Chr.2: 241750332-241750332 on Build GRCh38
rs889781455 Chr.2: 241750352-241750352 on Build GRCh38
rs572895480 Chr.2: 241750403-241750403 on Build GRCh38
rs1041680264 Chr.2: 241750387-241750387 on Build GRCh38
rs142073267 Chr.2: 241750287-241750287 on Build GRCh38
rs751879599 Chr.2: 241750280-241750280 on Build GRCh38
rs757451444 Chr.2: 241750159-241750159 on Build GRCh38
rs551151066 Chr.2: 241750362-241750362 on Build GRCh38
rs886055848 Chr.2: 241750220-241750220 on Build GRCh38
rs999667320 Chr.2: 241750395-241750395 on Build GRCh38
rs773997064 Chr.2: 241750104-241750104 on Build GRCh38
rs113529779 Chr.2: 241750261-241750261 on Build GRCh38
rs775900141 Chr.2: 241750311-241750311 on Build GRCh38
rs762771273 Chr.2: 241750100-241750100 on Build GRCh38
rs758727451 Chr.2: 241750143-241750143 on Build GRCh38
rs754967821 Chr.2: 241750329-241750329 on Build GRCh38
rs184060439 Chr.2: 241750129-241750129 on Build GRCh38
rs1057378101 Chr.2: 241750421-241750421 on Build GRCh38

View Full Product Details