Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 326162-326660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOX12
Entrez Gene ID
6666
Gene Name
SRY-box 12
Gene Aliases
SOX22
UniGene
Hs.43627
Gene Chromosome Location
Chr.20: 325571-330228 on Build GRCh38
Transcripts
NM_006943.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7251526 c.335G>T p.R112L Chr.20: 326259-326259 on Build GRCh38
4612908 c.682_687delGAGGAA p.E232_E233delEE Chr.20: 326606-326611 on Build GRCh38
4637014 c.383G>C p.G128A Chr.20: 326307-326307 on Build GRCh38
7254924 c.526G>C p.V176L Chr.20: 326450-326450 on Build GRCh38
4988192 c.305C>T p.A102V Chr.20: 326229-326229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982266240 Chr.20: 326558-326563 on Build GRCh38
rs1022017585 Chr.20: 326617-326617 on Build GRCh38
rs780154689 Chr.20: 326206-326206 on Build GRCh38
rs967433229 Chr.20: 326399-326399 on Build GRCh38
rs906397602 Chr.20: 326564-326564 on Build GRCh38
rs532036846 Chr.20: 326605-326605 on Build GRCh38
rs200558979 Chr.20: 326314-326314 on Build GRCh38
rs916714328 Chr.20: 326336-326336 on Build GRCh38
rs768711017 Chr.20: 326218-326218 on Build GRCh38
rs921509695 Chr.20: 326500-326500 on Build GRCh38
rs147752920 Chr.20: 326597-326605 on Build GRCh38
rs760101905 Chr.20: 326266-326266 on Build GRCh38
rs1019709029 Chr.20: 326631-326631 on Build GRCh38
rs908317093 Chr.20: 326557-326557 on Build GRCh38
rs1041476189 Chr.20: 326448-326448 on Build GRCh38
rs753328938 Chr.20: 326257-326257 on Build GRCh38
rs761494795 Chr.20: 326227-326227 on Build GRCh38
rs991407227 Chr.20: 326316-326316 on Build GRCh38
rs958538885 Chr.20: 326535-326535 on Build GRCh38
rs770657048 Chr.20: 326418-326418 on Build GRCh38
rs184118017 Chr.20: 326626-326626 on Build GRCh38
rs1050342387 Chr.20: 326609-326611 on Build GRCh38
rs764383462 Chr.20: 326309-326309 on Build GRCh38
rs535111667 Chr.20: 326342-326342 on Build GRCh38
rs557418764 Chr.20: 326531-326531 on Build GRCh38
rs902922633 Chr.20: 326452-326452 on Build GRCh38
rs866230814 Chr.20: 326586-326586 on Build GRCh38
rs748008617 Chr.20: 326414-326414 on Build GRCh38
rs866311137 Chr.20: 326271-326271 on Build GRCh38
rs867428648 Chr.20: 326259-326259 on Build GRCh38
rs890248293 Chr.20: 326422-326422 on Build GRCh38
rs1011981897 Chr.20: 326346-326346 on Build GRCh38
rs571702058 Chr.20: 326488-326488 on Build GRCh38
rs771694928 Chr.20: 326440-326440 on Build GRCh38
rs765814571 Chr.20: 326253-326253 on Build GRCh38
rs982230130 Chr.20: 326339-326339 on Build GRCh38
rs374987024 Chr.20: 326557-326558 on Build GRCh38
rs1046190720 Chr.20: 326310-326310 on Build GRCh38
rs778917309 Chr.20: 326637-326637 on Build GRCh38
rs562098614 Chr.20: 326558-326558 on Build GRCh38
rs1002103858 Chr.20: 326328-326328 on Build GRCh38
rs1055083034 Chr.20: 326570-326570 on Build GRCh38
rs770666496 Chr.20: 326558-326560 on Build GRCh38
rs1008567981 Chr.20: 326273-326273 on Build GRCh38
rs770850889 Chr.20: 326198-326198 on Build GRCh38
rs769785308 Chr.20: 326612-326614 on Build GRCh38
rs755018354 Chr.20: 326390-326390 on Build GRCh38
rs978436990 Chr.20: 326277-326277 on Build GRCh38
rs745915966 Chr.20: 326357-326357 on Build GRCh38
rs939858413 Chr.20: 326307-326307 on Build GRCh38
rs758639743 Chr.20: 326306-326306 on Build GRCh38
rs778948503 Chr.20: 326402-326402 on Build GRCh38
rs764425109 Chr.20: 326566-326566 on Build GRCh38
rs35462946 Chr.20: 326258-326258 on Build GRCh38
rs931003404 Chr.20: 326514-326514 on Build GRCh38
rs527670259 Chr.20: 326633-326633 on Build GRCh38
rs757594648 Chr.20: 326600-326600 on Build GRCh38
rs1008682476 Chr.20: 326431-326431 on Build GRCh38
rs372611150 Chr.20: 326457-326457 on Build GRCh38
rs1000287490 Chr.20: 326462-326462 on Build GRCh38
rs546352861 Chr.20: 326560-326560 on Build GRCh38
rs920527550 Chr.20: 326387-326387 on Build GRCh38
rs866806497 Chr.20: 326492-326492 on Build GRCh38
rs556205020 Chr.20: 326274-326274 on Build GRCh38
rs563044724 Chr.20: 326572-326572 on Build GRCh38
rs749297999 Chr.20: 326215-326215 on Build GRCh38
rs181303084 Chr.20: 326583-326583 on Build GRCh38
rs750556965 Chr.20: 326622-326622 on Build GRCh38
rs760233296 Chr.20: 326248-326248 on Build GRCh38
rs36103952 Chr.20: 326539-326539 on Build GRCh38
rs1023888940 Chr.20: 326545-326545 on Build GRCh38
rs1019825613 Chr.20: 326454-326454 on Build GRCh38
rs765753698 Chr.20: 326542-326542 on Build GRCh38
rs772697483 Chr.20: 326239-326239 on Build GRCh38
rs975730143 Chr.20: 326499-326499 on Build GRCh38
rs1033993488 Chr.20: 326338-326338 on Build GRCh38
rs774332168 Chr.20: 326221-326221 on Build GRCh38
rs746695082 Chr.20: 326478-326478 on Build GRCh38
rs554982419 Chr.20: 326434-326434 on Build GRCh38
rs771781431 Chr.20: 326236-326236 on Build GRCh38
rs753152460 Chr.20: 326276-326276 on Build GRCh38
rs1037691902 Chr.20: 326368-326368 on Build GRCh38
rs751675521 Chr.20: 326312-326312 on Build GRCh38
rs772928372 Chr.20: 326441-326441 on Build GRCh38
rs113440192 Chr.20: 326512-326512 on Build GRCh38
rs773988152 Chr.20: 326470-326470 on Build GRCh38

View Full Product Details