Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1164925-1165432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMF1
Entrez Gene ID
9491
Gene Name
proteasome inhibitor subunit 1
Gene Aliases
PI31
UniGene
Hs.471917
Gene Chromosome Location
Chr.20: 1113229-1172246 on Build GRCh38
Transcripts
NM_001323407.1 NM_001323408.1 NM_001323409.1 NM_001323410.1 NM_006814.4 NM_178578.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4096641 c.807G>A p.M269I Chr.20: 1165071-1165071 on Build GRCh38
3544102 c.783C>T p.L261L Chr.20: 1165047-1165047 on Build GRCh38
4508695 c.784C>T p.P262S Chr.20: 1165048-1165048 on Build GRCh38
1024494 c.799G>A p.D267N Chr.20: 1165063-1165063 on Build GRCh38
5456804 c.791C>T p.P264L Chr.20: 1165055-1165055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759055739 Chr.20: 1164979-1164979 on Build GRCh38
rs765711831 Chr.20: 1164990-1164990 on Build GRCh38
rs772132109 Chr.20: 1165389-1165389 on Build GRCh38
rs895123721 Chr.20: 1165026-1165026 on Build GRCh38
rs946599975 Chr.20: 1165300-1165300 on Build GRCh38
rs552235484 Chr.20: 1165136-1165136 on Build GRCh38
rs367774059 Chr.20: 1165103-1165103 on Build GRCh38
rs370516617 Chr.20: 1165021-1165021 on Build GRCh38
rs781367296 Chr.20: 1165043-1165043 on Build GRCh38
rs750512137 Chr.20: 1165047-1165047 on Build GRCh38
rs1016420572 Chr.20: 1165272-1165272 on Build GRCh38
rs748009139 Chr.20: 1165056-1165056 on Build GRCh38
rs115433407 Chr.20: 1165121-1165121 on Build GRCh38
rs763498006 Chr.20: 1165105-1165105 on Build GRCh38
rs575550221 Chr.20: 1165406-1165406 on Build GRCh38
rs764489136 Chr.20: 1165013-1165013 on Build GRCh38
rs958994234 Chr.20: 1165238-1165238 on Build GRCh38
rs745426724 Chr.20: 1165088-1165088 on Build GRCh38
rs568802161 Chr.20: 1165172-1165172 on Build GRCh38
rs994728874 Chr.20: 1165143-1165143 on Build GRCh38
rs1028681256 Chr.20: 1165179-1165179 on Build GRCh38
rs536463292 Chr.20: 1165358-1165358 on Build GRCh38
rs746694596 Chr.20: 1165079-1165079 on Build GRCh38
rs775867301 Chr.20: 1165102-1165102 on Build GRCh38
rs762239459 Chr.20: 1165122-1165122 on Build GRCh38
rs777227111 Chr.20: 1164986-1164987 on Build GRCh38
rs1026085511 Chr.20: 1165204-1165204 on Build GRCh38
rs981857645 Chr.20: 1165222-1165222 on Build GRCh38
rs984547687 Chr.20: 1165254-1165254 on Build GRCh38
rs777492424 Chr.20: 1165072-1165072 on Build GRCh38
rs796802123 Chr.20: 1165039-1165039 on Build GRCh38
rs879845824 Chr.20: 1165245-1165245 on Build GRCh38
rs557234229 Chr.20: 1165278-1165278 on Build GRCh38
rs17716261 Chr.20: 1165404-1165404 on Build GRCh38
rs189360363 Chr.20: 1165205-1165205 on Build GRCh38
rs201939911 Chr.20: 1165093-1165093 on Build GRCh38
rs770529679 Chr.20: 1165084-1165084 on Build GRCh38
rs778779823 Chr.20: 1165051-1165051 on Build GRCh38
rs915151430 Chr.20: 1165262-1165262 on Build GRCh38
rs1042511498 Chr.20: 1164980-1164980 on Build GRCh38
rs372979271 Chr.20: 1165287-1165287 on Build GRCh38
rs756199742 Chr.20: 1165050-1165050 on Build GRCh38
rs139171551 Chr.20: 1165353-1165353 on Build GRCh38
rs1006887452 Chr.20: 1165395-1165395 on Build GRCh38
rs903716699 Chr.20: 1165063-1165063 on Build GRCh38
rs775963251 Chr.20: 1164978-1164978 on Build GRCh38
rs934075598 Chr.20: 1165359-1165359 on Build GRCh38
rs1042596005 Chr.20: 1165343-1165343 on Build GRCh38
rs962950764 Chr.20: 1165332-1165332 on Build GRCh38
rs764577093 Chr.20: 1165114-1165114 on Build GRCh38
rs746404766 Chr.20: 1165046-1165047 on Build GRCh38
rs927642793 Chr.20: 1165226-1165226 on Build GRCh38
rs566584380 Chr.20: 1165057-1165057 on Build GRCh38
rs898722322 Chr.20: 1165407-1165407 on Build GRCh38
rs918876884 Chr.20: 1165387-1165387 on Build GRCh38
rs374138693 Chr.20: 1165135-1165135 on Build GRCh38
rs1051139142 Chr.20: 1165383-1165383 on Build GRCh38
rs374673008 Chr.20: 1164993-1164993 on Build GRCh38
rs1018283897 Chr.20: 1165223-1165223 on Build GRCh38
rs776322535 Chr.20: 1165085-1165085 on Build GRCh38
rs985741992 Chr.20: 1165212-1165212 on Build GRCh38
rs757506454 Chr.20: 1165042-1165042 on Build GRCh38
rs372553802 Chr.20: 1164991-1164991 on Build GRCh38

View Full Product Details