Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 70438606-70439115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTH
Entrez Gene ID
1491
Gene Name
cystathionine gamma-lyase
Gene Aliases
-
UniGene
Hs.19904
Gene Chromosome Location
Chr.1: 70411218-70441949 on Build GRCh38
Transcripts
NM_001190463.1 NM_001902.5 NM_153742.4 XM_005270509.3 XM_017000416.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1196850 c.1067A>G p.H356R Chr.1: 70438702-70438702 on Build GRCh38
1202576 c.1070C>T p.A357V Chr.1: 70438705-70438705 on Build GRCh38
6054513 c.1136G>T p.G379V Chr.1: 70438771-70438771 on Build GRCh38
7241144 c.1075G>A p.V359I Chr.1: 70438710-70438710 on Build GRCh38
5007913 c.1123delC p.R375fs*6 Chr.1: 70438758-70438758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879046306 Chr.1: 70438841-70438850 on Build GRCh38
rs781141588 Chr.1: 70439072-70439072 on Build GRCh38
rs756277614 Chr.1: 70438758-70438758 on Build GRCh38
rs766372153 Chr.1: 70438770-70438770 on Build GRCh38
rs374498069 Chr.1: 70438747-70438747 on Build GRCh38
rs756118802 Chr.1: 70438650-70438650 on Build GRCh38
rs886764325 Chr.1: 70439057-70439057 on Build GRCh38
rs978764947 Chr.1: 70438904-70438904 on Build GRCh38
rs773965646 Chr.1: 70438872-70438872 on Build GRCh38
rs746748634 Chr.1: 70438678-70438678 on Build GRCh38
rs762518831 Chr.1: 70438704-70438704 on Build GRCh38
rs768853335 Chr.1: 70438672-70438672 on Build GRCh38
rs774996710 Chr.1: 70438839-70438840 on Build GRCh38
rs746818113 Chr.1: 70438699-70438699 on Build GRCh38
rs781631349 Chr.1: 70438637-70438637 on Build GRCh38
rs1053237824 Chr.1: 70438972-70438972 on Build GRCh38
rs142226680 Chr.1: 70438848-70438849 on Build GRCh38
rs878882001 Chr.1: 70438850-70438851 on Build GRCh38
rs939821396 Chr.1: 70439061-70439061 on Build GRCh38
rs369641854 Chr.1: 70438887-70438888 on Build GRCh38
rs775009574 Chr.1: 70438699-70438699 on Build GRCh38
rs771041647 Chr.1: 70438679-70438679 on Build GRCh38
rs765005172 Chr.1: 70438871-70438872 on Build GRCh38
rs1004352906 Chr.1: 70438706-70438706 on Build GRCh38
rs753537579 Chr.1: 70438863-70438870 on Build GRCh38
rs766857581 Chr.1: 70439058-70439058 on Build GRCh38
rs776678178 Chr.1: 70438682-70438682 on Build GRCh38
rs143053803 Chr.1: 70438975-70438975 on Build GRCh38
rs759183340 Chr.1: 70438687-70438687 on Build GRCh38
rs112004441 Chr.1: 70438840-70438841 on Build GRCh38
rs755053825 Chr.1: 70439060-70439060 on Build GRCh38
rs958031563 Chr.1: 70438792-70438792 on Build GRCh38
rs758278230 Chr.1: 70439081-70439081 on Build GRCh38
rs750929169 Chr.1: 70438744-70438744 on Build GRCh38
rs773495840 Chr.1: 70439050-70439050 on Build GRCh38
rs753911466 Chr.1: 70438771-70438771 on Build GRCh38
rs1046187746 Chr.1: 70438996-70438996 on Build GRCh38
rs758634523 Chr.1: 70439068-70439068 on Build GRCh38
rs780392655 Chr.1: 70438655-70438655 on Build GRCh38
rs763763933 Chr.1: 70438705-70438705 on Build GRCh38
rs762234423 Chr.1: 70438873-70438873 on Build GRCh38
rs113615213 Chr.1: 70439048-70439048 on Build GRCh38
rs761114601 Chr.1: 70439053-70439053 on Build GRCh38
rs756918121 Chr.1: 70438866-70438867 on Build GRCh38
rs762148181 Chr.1: 70438724-70438724 on Build GRCh38
rs3221630 Chr.1: 70438843-70438844 on Build GRCh38
rs753910024 Chr.1: 70439059-70439059 on Build GRCh38
rs562535861 Chr.1: 70439003-70439003 on Build GRCh38
rs75559784 Chr.1: 70438870-70438870 on Build GRCh38
rs770606747 Chr.1: 70438817-70438817 on Build GRCh38
rs963458808 Chr.1: 70438840-70438840 on Build GRCh38
rs58815241 Chr.1: 70438869-70438870 on Build GRCh38
rs149061359 Chr.1: 70438899-70438899 on Build GRCh38
rs199832524 Chr.1: 70439062-70439062 on Build GRCh38
rs991422524 Chr.1: 70438969-70438969 on Build GRCh38
rs915802860 Chr.1: 70438974-70438974 on Build GRCh38
rs749482337 Chr.1: 70438657-70438657 on Build GRCh38
rs926016396 Chr.1: 70438958-70438958 on Build GRCh38
rs763561019 Chr.1: 70438846-70438847 on Build GRCh38
rs745808575 Chr.1: 70438803-70438803 on Build GRCh38
rs376152054 Chr.1: 70438692-70438692 on Build GRCh38
rs982589572 Chr.1: 70438893-70438893 on Build GRCh38
rs756028356 Chr.1: 70439082-70439082 on Build GRCh38
rs202074968 Chr.1: 70438791-70438791 on Build GRCh38
rs12124009 Chr.1: 70438671-70438671 on Build GRCh38
rs777739313 Chr.1: 70438673-70438673 on Build GRCh38
rs938509024 Chr.1: 70438961-70438961 on Build GRCh38
rs368682761 Chr.1: 70438761-70438761 on Build GRCh38
rs769707444 Chr.1: 70438822-70438822 on Build GRCh38
rs767856919 Chr.1: 70438874-70438874 on Build GRCh38
rs752952965 Chr.1: 70439064-70439064 on Build GRCh38
rs541334870 Chr.1: 70438879-70438879 on Build GRCh38
rs745637431 Chr.1: 70439074-70439074 on Build GRCh38
rs144875036 Chr.1: 70438807-70438807 on Build GRCh38
rs370657238 Chr.1: 70438789-70438789 on Build GRCh38
rs374296307 Chr.1: 70438802-70438802 on Build GRCh38
rs760136564 Chr.1: 70438841-70438842 on Build GRCh38
rs779255023 Chr.1: 70438783-70438783 on Build GRCh38
rs750553951 Chr.1: 70438644-70438644 on Build GRCh38
rs755173328 Chr.1: 70438776-70438776 on Build GRCh38
rs531740937 Chr.1: 70439008-70439008 on Build GRCh38
rs1048075034 Chr.1: 70439036-70439036 on Build GRCh38
rs560822814 Chr.1: 70438895-70438895 on Build GRCh38
rs989452771 Chr.1: 70438827-70438827 on Build GRCh38
rs544773981 Chr.1: 70438796-70438796 on Build GRCh38
rs750162995 Chr.1: 70439075-70439076 on Build GRCh38
rs774720932 Chr.1: 70438708-70438708 on Build GRCh38
rs370492686 Chr.1: 70438727-70438727 on Build GRCh38
rs748783233 Chr.1: 70438839-70438839 on Build GRCh38
rs905129154 Chr.1: 70439073-70439074 on Build GRCh38
rs966807304 Chr.1: 70438888-70438888 on Build GRCh38

View Full Product Details