Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 71064748-71065247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZRANB2
Entrez Gene ID
9406
Gene Name
zinc finger RANBP2-type containing 2
Gene Aliases
ZIS, ZIS1, ZIS2, ZNF265
UniGene
Hs.194718
Gene Chromosome Location
Chr.1: 71063291-71081289 on Build GRCh38
Transcripts
NM_005455.4 NM_203350.2 NR_038420.1

Gene Symbol
ZRANB2-AS1
Entrez Gene ID
100132618
Gene Name
ZRANB2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.720513
Gene Chromosome Location
Chr.1: 71046506-71067184 on Build GRCh38
Transcripts
NM_005455.4 NM_203350.2 NR_038420.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
293179 c.932G>A p.R311H Chr.1: 71065135-71065135 on Build GRCh38
2241138 c.977G>T p.S326I Chr.1: 71065090-71065090 on Build GRCh38
1344207 c.987G>T p.K329N Chr.1: 71065080-71065080 on Build GRCh38
4745362 c.970C>T p.R324C Chr.1: 71065097-71065097 on Build GRCh38
227410 c.944C>T p.S315L Chr.1: 71065123-71065123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767164175 Chr.1: 71065066-71065066 on Build GRCh38
rs201924525 Chr.1: 71065109-71065109 on Build GRCh38
rs113395401 Chr.1: 71065188-71065188 on Build GRCh38
rs761085397 Chr.1: 71065087-71065087 on Build GRCh38
rs765336022 Chr.1: 71065094-71065094 on Build GRCh38
rs895071639 Chr.1: 71064972-71064972 on Build GRCh38
rs181548907 Chr.1: 71065169-71065169 on Build GRCh38
rs761800037 Chr.1: 71065151-71065151 on Build GRCh38
rs764015441 Chr.1: 71065165-71065165 on Build GRCh38
rs796371479 Chr.1: 71065050-71065053 on Build GRCh38
rs939216792 Chr.1: 71064961-71064961 on Build GRCh38
rs546434261 Chr.1: 71064867-71064867 on Build GRCh38
rs1039388028 Chr.1: 71065171-71065171 on Build GRCh38
rs757370186 Chr.1: 71065173-71065173 on Build GRCh38
rs772045580 Chr.1: 71064959-71064959 on Build GRCh38
rs768674308 Chr.1: 71065132-71065132 on Build GRCh38
rs189228363 Chr.1: 71065046-71065046 on Build GRCh38
rs778042706 Chr.1: 71065102-71065102 on Build GRCh38
rs150149903 Chr.1: 71065091-71065091 on Build GRCh38
rs377531298 Chr.1: 71065008-71065008 on Build GRCh38
rs137863951 Chr.1: 71065136-71065136 on Build GRCh38
rs571102816 Chr.1: 71065149-71065149 on Build GRCh38
rs373641078 Chr.1: 71065183-71065183 on Build GRCh38
rs944004073 Chr.1: 71064987-71064987 on Build GRCh38
rs998372565 Chr.1: 71065080-71065081 on Build GRCh38
rs528544138 Chr.1: 71064970-71064970 on Build GRCh38
rs752746050 Chr.1: 71065096-71065096 on Build GRCh38
rs1053831094 Chr.1: 71065219-71065219 on Build GRCh38
rs888475935 Chr.1: 71065011-71065011 on Build GRCh38
rs112462014 Chr.1: 71064944-71064944 on Build GRCh38
rs148862349 Chr.1: 71065097-71065097 on Build GRCh38
rs892561831 Chr.1: 71065223-71065223 on Build GRCh38
rs759989507 Chr.1: 71065092-71065092 on Build GRCh38
rs548299127 Chr.1: 71065000-71065000 on Build GRCh38
rs901248890 Chr.1: 71065212-71065212 on Build GRCh38
rs370108422 Chr.1: 71065148-71065148 on Build GRCh38
rs372855432 Chr.1: 71064818-71064818 on Build GRCh38
rs562573141 Chr.1: 71064810-71064810 on Build GRCh38
rs530849164 Chr.1: 71065086-71065086 on Build GRCh38
rs377326726 Chr.1: 71065160-71065160 on Build GRCh38
rs746439417 Chr.1: 71065136-71065139 on Build GRCh38
rs760175069 Chr.1: 71065144-71065144 on Build GRCh38
rs199511818 Chr.1: 71065108-71065108 on Build GRCh38
rs1039721821 Chr.1: 71064999-71064999 on Build GRCh38
rs772910245 Chr.1: 71065140-71065140 on Build GRCh38
rs988379292 Chr.1: 71064791-71064791 on Build GRCh38
rs966379959 Chr.1: 71065123-71065123 on Build GRCh38
rs977813121 Chr.1: 71065135-71065135 on Build GRCh38
rs773949852 Chr.1: 71065032-71065032 on Build GRCh38
rs146951263 Chr.1: 71064866-71064866 on Build GRCh38
rs778605260 Chr.1: 71065133-71065133 on Build GRCh38
rs749257543 Chr.1: 71065119-71065119 on Build GRCh38
rs919076950 Chr.1: 71064881-71064881 on Build GRCh38
rs766860131 Chr.1: 71065090-71065090 on Build GRCh38
rs926335887 Chr.1: 71064855-71064855 on Build GRCh38
rs892635951 Chr.1: 71065072-71065072 on Build GRCh38
rs930548073 Chr.1: 71065163-71065163 on Build GRCh38
rs930597272 Chr.1: 71064991-71064991 on Build GRCh38
rs879467161 Chr.1: 71064804-71064804 on Build GRCh38
rs899892793 Chr.1: 71065006-71065006 on Build GRCh38
rs963401954 Chr.1: 71065138-71065138 on Build GRCh38
rs373037504 Chr.1: 71065152-71065152 on Build GRCh38
rs146523749 Chr.1: 71065076-71065076 on Build GRCh38
rs375781578 Chr.1: 71065141-71065141 on Build GRCh38
rs761589155 Chr.1: 71065058-71065058 on Build GRCh38
rs372571673 Chr.1: 71065137-71065137 on Build GRCh38
rs138154674 Chr.1: 71065195-71065195 on Build GRCh38
rs779935381 Chr.1: 71065111-71065111 on Build GRCh38
rs776997728 Chr.1: 71064838-71064838 on Build GRCh38
rs564267368 Chr.1: 71064837-71064837 on Build GRCh38
rs58499881 Chr.1: 71065047-71065050 on Build GRCh38
rs1009740912 Chr.1: 71065088-71065088 on Build GRCh38
rs936702018 Chr.1: 71065196-71065196 on Build GRCh38
rs370181295 Chr.1: 71064821-71064821 on Build GRCh38
rs772496727 Chr.1: 71065165-71065166 on Build GRCh38

View Full Product Details