Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 71610887-71611432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEGR1
Entrez Gene ID
257194
Gene Name
neuronal growth regulator 1
Gene Aliases
DMML2433, IGLON4, KILON, Ntra
UniGene
Hs.146542
Gene Chromosome Location
Chr.1: 71402942-72282850 on Build GRCh38
Transcripts
NM_173808.2 XM_011541200.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5434945 c.715C>T p.R239C Chr.1: 71611099-71611099 on Build GRCh38
295284 c.753G>A p.P251P Chr.1: 71611061-71611061 on Build GRCh38
4009816 c.786G>A p.K262K Chr.1: 71611028-71611028 on Build GRCh38
4009815 c.788A>C p.K263T Chr.1: 71611026-71611026 on Build GRCh38
912044 c.713delG p.G238fs*5 Chr.1: 71611101-71611101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907498033 Chr.1: 71611405-71611405 on Build GRCh38
rs773803468 Chr.1: 71611171-71611171 on Build GRCh38
rs368150568 Chr.1: 71611020-71611020 on Build GRCh38
rs761756380 Chr.1: 71611371-71611371 on Build GRCh38
rs572399407 Chr.1: 71611149-71611149 on Build GRCh38
rs192019593 Chr.1: 71611337-71611337 on Build GRCh38
rs541513538 Chr.1: 71611175-71611175 on Build GRCh38
rs537927549 Chr.1: 71611269-71611269 on Build GRCh38
rs763876467 Chr.1: 71610977-71610978 on Build GRCh38
rs117315763 Chr.1: 71611270-71611270 on Build GRCh38
rs1042848424 Chr.1: 71611246-71611246 on Build GRCh38
rs990499804 Chr.1: 71611104-71611104 on Build GRCh38
rs760362193 Chr.1: 71611195-71611195 on Build GRCh38
rs755128597 Chr.1: 71611000-71611000 on Build GRCh38
rs748357135 Chr.1: 71611006-71611006 on Build GRCh38
rs576347795 Chr.1: 71611102-71611102 on Build GRCh38
rs773893499 Chr.1: 71611092-71611092 on Build GRCh38
rs749275382 Chr.1: 71611163-71611163 on Build GRCh38
rs984863515 Chr.1: 71611402-71611402 on Build GRCh38
rs889312605 Chr.1: 71611221-71611221 on Build GRCh38
rs991342328 Chr.1: 71611030-71611030 on Build GRCh38
rs754424709 Chr.1: 71611105-71611105 on Build GRCh38
rs761433632 Chr.1: 71611097-71611097 on Build GRCh38
rs548564946 Chr.1: 71611076-71611076 on Build GRCh38
rs745680449 Chr.1: 71611040-71611040 on Build GRCh38
rs780706498 Chr.1: 71610979-71610979 on Build GRCh38
rs190127810 Chr.1: 71610962-71610962 on Build GRCh38
rs768570798 Chr.1: 71611170-71611170 on Build GRCh38
rs778071072 Chr.1: 71611116-71611116 on Build GRCh38
rs967020779 Chr.1: 71611060-71611060 on Build GRCh38
rs523835 Chr.1: 71611211-71611211 on Build GRCh38
rs1023329914 Chr.1: 71611365-71611365 on Build GRCh38
rs148555653 Chr.1: 71611219-71611219 on Build GRCh38
rs1035246330 Chr.1: 71611302-71611302 on Build GRCh38
rs776710041 Chr.1: 71611156-71611159 on Build GRCh38
rs373419972 Chr.1: 71611099-71611099 on Build GRCh38
rs769521243 Chr.1: 71611053-71611053 on Build GRCh38
rs772936719 Chr.1: 71611184-71611184 on Build GRCh38
rs545038964 Chr.1: 71611107-71611107 on Build GRCh38
rs567510565 Chr.1: 71610939-71610939 on Build GRCh38
rs369461565 Chr.1: 71611231-71611231 on Build GRCh38
rs940712254 Chr.1: 71611176-71611176 on Build GRCh38
rs779095459 Chr.1: 71611002-71611002 on Build GRCh38
rs1022334401 Chr.1: 71611268-71611268 on Build GRCh38
rs972008737 Chr.1: 71611183-71611183 on Build GRCh38
rs947518908 Chr.1: 71611229-71611229 on Build GRCh38
rs563548424 Chr.1: 71611248-71611248 on Build GRCh38
rs901642712 Chr.1: 71611217-71611217 on Build GRCh38
rs761387776 Chr.1: 71611174-71611174 on Build GRCh38
rs756643165 Chr.1: 71610978-71610978 on Build GRCh38
rs767198961 Chr.1: 71611098-71611098 on Build GRCh38
rs554178019 Chr.1: 71611291-71611291 on Build GRCh38
rs923554260 Chr.1: 71611201-71611201 on Build GRCh38
rs747152886 Chr.1: 71611138-71611138 on Build GRCh38
rs536481896 Chr.1: 71610943-71610943 on Build GRCh38
rs61729931 Chr.1: 71611112-71611112 on Build GRCh38
rs369017807 Chr.1: 71611061-71611061 on Build GRCh38
rs889901914 Chr.1: 71611225-71611225 on Build GRCh38
rs749751648 Chr.1: 71610980-71610980 on Build GRCh38
rs142995669 Chr.1: 71611399-71611399 on Build GRCh38
rs867277651 Chr.1: 71611063-71611063 on Build GRCh38
rs750790702 Chr.1: 71610975-71610975 on Build GRCh38
rs763636088 Chr.1: 71611082-71611082 on Build GRCh38
rs766797101 Chr.1: 71611103-71611103 on Build GRCh38
rs145840571 Chr.1: 71611111-71611111 on Build GRCh38
rs182424786 Chr.1: 71611212-71611212 on Build GRCh38
rs895483338 Chr.1: 71611276-71611276 on Build GRCh38
rs567842076 Chr.1: 71611338-71611338 on Build GRCh38
rs1013285615 Chr.1: 71611330-71611330 on Build GRCh38
rs776347091 Chr.1: 71611028-71611028 on Build GRCh38
rs761837398 Chr.1: 71611155-71611156 on Build GRCh38
rs187422572 Chr.1: 71611265-71611265 on Build GRCh38
rs779795906 Chr.1: 71611158-71611158 on Build GRCh38
rs532465511 Chr.1: 71611261-71611261 on Build GRCh38

View Full Product Details