Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 71697873-71698403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEGR1
Entrez Gene ID
257194
Gene Name
neuronal growth regulator 1
Gene Aliases
DMML2433, IGLON4, KILON, Ntra
UniGene
Hs.146542
Gene Chromosome Location
Chr.1: 71402942-72282850 on Build GRCh38
Transcripts
NM_173808.2 XM_011541200.2 XM_011541201.2 XM_017000961.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6527657 c.614C>G p.A205G Chr.1: 71698061-71698061 on Build GRCh38
4009818 c.538A>G p.K180E Chr.1: 71698137-71698137 on Build GRCh38
6939459 c.575G>A p.G192E Chr.1: 71698100-71698100 on Build GRCh38
4009817 c.548A>C p.E183A Chr.1: 71698127-71698127 on Build GRCh38
6937113 c.598G>A p.E200K Chr.1: 71698077-71698077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866634961 Chr.1: 71698026-71698026 on Build GRCh38
rs964488973 Chr.1: 71698153-71698153 on Build GRCh38
rs766888116 Chr.1: 71697957-71697957 on Build GRCh38
rs1037718574 Chr.1: 71698305-71698305 on Build GRCh38
rs370964275 Chr.1: 71697997-71697997 on Build GRCh38
rs767735536 Chr.1: 71698170-71698170 on Build GRCh38
rs370464016 Chr.1: 71698167-71698167 on Build GRCh38
rs754232246 Chr.1: 71697969-71697969 on Build GRCh38
rs187159183 Chr.1: 71698182-71698182 on Build GRCh38
rs1012489096 Chr.1: 71698299-71698299 on Build GRCh38
rs926252431 Chr.1: 71697977-71698001 on Build GRCh38
rs773691059 Chr.1: 71698098-71698098 on Build GRCh38
rs1026713844 Chr.1: 71698360-71698360 on Build GRCh38
rs765266693 Chr.1: 71698167-71698167 on Build GRCh38
rs377679053 Chr.1: 71697994-71697994 on Build GRCh38
rs764285516 Chr.1: 71698143-71698143 on Build GRCh38
rs576344350 Chr.1: 71698206-71698206 on Build GRCh38
rs561902724 Chr.1: 71698157-71698157 on Build GRCh38
rs779885533 Chr.1: 71698032-71698032 on Build GRCh38
rs751848901 Chr.1: 71698010-71698010 on Build GRCh38
rs372699137 Chr.1: 71698263-71698264 on Build GRCh38
rs767134486 Chr.1: 71698052-71698052 on Build GRCh38
rs778919318 Chr.1: 71698051-71698051 on Build GRCh38
rs377713205 Chr.1: 71698072-71698072 on Build GRCh38
rs975978821 Chr.1: 71698047-71698047 on Build GRCh38
rs757450838 Chr.1: 71698023-71698023 on Build GRCh38
rs536071086 Chr.1: 71698175-71698175 on Build GRCh38
rs771640260 Chr.1: 71698054-71698054 on Build GRCh38
rs751629193 Chr.1: 71698144-71698144 on Build GRCh38
rs890740273 Chr.1: 71698363-71698363 on Build GRCh38
rs879252255 Chr.1: 71698323-71698323 on Build GRCh38
rs760178310 Chr.1: 71697974-71697974 on Build GRCh38
rs768377820 Chr.1: 71698042-71698042 on Build GRCh38
rs895422218 Chr.1: 71698278-71698278 on Build GRCh38
rs201170592 Chr.1: 71698078-71698078 on Build GRCh38
rs765470454 Chr.1: 71698107-71698107 on Build GRCh38
rs1044271386 Chr.1: 71698239-71698239 on Build GRCh38
rs538215185 Chr.1: 71698217-71698217 on Build GRCh38
rs1002236699 Chr.1: 71697929-71697929 on Build GRCh38
rs573330495 Chr.1: 71697966-71697966 on Build GRCh38
rs925612075 Chr.1: 71698161-71698161 on Build GRCh38
rs1056748280 Chr.1: 71698270-71698270 on Build GRCh38
rs772562752 Chr.1: 71698060-71698060 on Build GRCh38
rs202121169 Chr.1: 71698091-71698091 on Build GRCh38
rs765835588 Chr.1: 71697985-71697985 on Build GRCh38
rs778060963 Chr.1: 71697910-71697910 on Build GRCh38
rs758528413 Chr.1: 71697995-71697995 on Build GRCh38
rs763339570 Chr.1: 71698125-71698125 on Build GRCh38
rs899186787 Chr.1: 71698306-71698306 on Build GRCh38
rs184229753 Chr.1: 71698174-71698174 on Build GRCh38
rs373210517 Chr.1: 71698036-71698036 on Build GRCh38
rs148707586 Chr.1: 71698089-71698089 on Build GRCh38
rs147799646 Chr.1: 71698324-71698324 on Build GRCh38
rs34844534 Chr.1: 71698123-71698123 on Build GRCh38
rs887939892 Chr.1: 71698327-71698327 on Build GRCh38
rs558526444 Chr.1: 71698254-71698254 on Build GRCh38

View Full Product Details