Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 75143038-75143550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LHX8
Entrez Gene ID
431707
Gene Name
LIM homeobox 8
Gene Aliases
LHX7
UniGene
Hs.403934
Gene Chromosome Location
Chr.1: 75128434-75199454 on Build GRCh38
Transcripts
NM_001001933.1 NM_001256114.1 XM_017001316.1 XM_017001317.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5518172 c.555C>T p.L185L Chr.1: 75143283-75143283 on Build GRCh38
6064357 c.409T>G p.S137A Chr.1: 75143137-75143137 on Build GRCh38
7002851 c.608A>G p.N203S Chr.1: 75143336-75143336 on Build GRCh38
7089442 c.555C>A p.L185L Chr.1: 75143283-75143283 on Build GRCh38
3492326 c.468C>T p.V156V Chr.1: 75143196-75143196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031280507 Chr.1: 75143505-75143505 on Build GRCh38
rs12084309 Chr.1: 75143169-75143169 on Build GRCh38
rs762257171 Chr.1: 75143235-75143235 on Build GRCh38
rs762535495 Chr.1: 75143211-75143211 on Build GRCh38
rs367958622 Chr.1: 75143132-75143132 on Build GRCh38
rs376834338 Chr.1: 75143154-75143154 on Build GRCh38
rs1040705028 Chr.1: 75143442-75143442 on Build GRCh38
rs765335899 Chr.1: 75143301-75143301 on Build GRCh38
rs776278766 Chr.1: 75143503-75143503 on Build GRCh38
rs768239726 Chr.1: 75143350-75143350 on Build GRCh38
rs372526079 Chr.1: 75143373-75143373 on Build GRCh38
rs754503872 Chr.1: 75143334-75143335 on Build GRCh38
rs777629713 Chr.1: 75143165-75143165 on Build GRCh38
rs773244431 Chr.1: 75143208-75143208 on Build GRCh38
rs773828288 Chr.1: 75143370-75143370 on Build GRCh38
rs889040920 Chr.1: 75143096-75143096 on Build GRCh38
rs570713311 Chr.1: 75143522-75143522 on Build GRCh38
rs767774054 Chr.1: 75143123-75143123 on Build GRCh38
rs775346408 Chr.1: 75143194-75143194 on Build GRCh38
rs754073470 Chr.1: 75143256-75143256 on Build GRCh38
rs550720602 Chr.1: 75143426-75143426 on Build GRCh38
rs761169415 Chr.1: 75143242-75143242 on Build GRCh38
rs865888187 Chr.1: 75143447-75143447 on Build GRCh38
rs1006977699 Chr.1: 75143372-75143372 on Build GRCh38
rs754988725 Chr.1: 75143288-75143288 on Build GRCh38
rs767872147 Chr.1: 75143237-75143237 on Build GRCh38
rs762176278 Chr.1: 75143102-75143102 on Build GRCh38
rs982386836 Chr.1: 75143266-75143266 on Build GRCh38
rs997223743 Chr.1: 75143351-75143351 on Build GRCh38
rs12041465 Chr.1: 75143364-75143364 on Build GRCh38
rs558167920 Chr.1: 75143331-75143331 on Build GRCh38
rs779790278 Chr.1: 75143335-75143335 on Build GRCh38
rs772454795 Chr.1: 75143365-75143365 on Build GRCh38
rs148556375 Chr.1: 75143148-75143148 on Build GRCh38
rs750724069 Chr.1: 75143238-75143238 on Build GRCh38
rs944528166 Chr.1: 75143441-75143441 on Build GRCh38
rs962366298 Chr.1: 75143355-75143355 on Build GRCh38
rs745723075 Chr.1: 75143173-75143173 on Build GRCh38
rs191182882 Chr.1: 75143097-75143097 on Build GRCh38
rs766738323 Chr.1: 75143378-75143378 on Build GRCh38
rs756269952 Chr.1: 75143141-75143141 on Build GRCh38
rs775116820 Chr.1: 75143088-75143088 on Build GRCh38
rs752170062 Chr.1: 75143099-75143099 on Build GRCh38
rs759144207 Chr.1: 75143078-75143078 on Build GRCh38
rs755990929 Chr.1: 75143332-75143332 on Build GRCh38
rs774931115 Chr.1: 75143234-75143234 on Build GRCh38
rs769300858 Chr.1: 75143082-75143082 on Build GRCh38
rs769297194 Chr.1: 75143188-75143188 on Build GRCh38
rs750917640 Chr.1: 75143131-75143131 on Build GRCh38
rs753913303 Chr.1: 75143159-75143159 on Build GRCh38
rs768294649 Chr.1: 75143231-75143231 on Build GRCh38
rs757121845 Chr.1: 75143327-75143327 on Build GRCh38
rs762590554 Chr.1: 75143093-75143093 on Build GRCh38
rs766574879 Chr.1: 75143253-75143253 on Build GRCh38
rs746953778 Chr.1: 75143166-75143166 on Build GRCh38
rs930696983 Chr.1: 75143396-75143396 on Build GRCh38
rs374741343 Chr.1: 75143172-75143172 on Build GRCh38
rs372110957 Chr.1: 75143328-75143328 on Build GRCh38
rs371719492 Chr.1: 75143164-75143164 on Build GRCh38
rs751468028 Chr.1: 75143306-75143306 on Build GRCh38
rs980074032 Chr.1: 75143460-75143460 on Build GRCh38
rs1018410091 Chr.1: 75143375-75143375 on Build GRCh38
rs986172951 Chr.1: 75143422-75143422 on Build GRCh38
rs1011085947 Chr.1: 75143176-75143176 on Build GRCh38
rs934323573 Chr.1: 75143517-75143517 on Build GRCh38
rs748952412 Chr.1: 75143347-75143347 on Build GRCh38

View Full Product Details