Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3854682-3855220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAVS
Entrez Gene ID
57506
Gene Name
mitochondrial antiviral signaling protein
Gene Aliases
CARDIF, IPS-1, IPS1, VISA
UniGene
Hs.570362
Gene Chromosome Location
Chr.20: 3846799-3876123 on Build GRCh38
Transcripts
NM_001206491.1 NM_020746.4 NR_037921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6159744 c.102C>G p.L34L Chr.20: 3854726-3854726 on Build GRCh38
6017799 c.100C>T p.L34F Chr.20: 3854724-3854724 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs569921246 Chr.20: 3855119-3855119 on Build GRCh38
rs776208323 Chr.20: 3854762-3854762 on Build GRCh38
rs186659488 Chr.20: 3854923-3854923 on Build GRCh38
rs1037260354 Chr.20: 3855064-3855064 on Build GRCh38
rs559533874 Chr.20: 3854916-3854916 on Build GRCh38
rs988062754 Chr.20: 3854895-3854896 on Build GRCh38
rs992040030 Chr.20: 3854853-3854853 on Build GRCh38
rs780448955 Chr.20: 3854784-3854784 on Build GRCh38
rs1013438788 Chr.20: 3855131-3855131 on Build GRCh38
rs887071286 Chr.20: 3855121-3855121 on Build GRCh38
rs1032173039 Chr.20: 3854772-3854772 on Build GRCh38
rs995151804 Chr.20: 3855079-3855079 on Build GRCh38
rs969169651 Chr.20: 3855135-3855135 on Build GRCh38
rs200003136 Chr.20: 3854894-3854895 on Build GRCh38
rs374964470 Chr.20: 3854953-3854953 on Build GRCh38
rs570601968 Chr.20: 3854948-3854948 on Build GRCh38
rs373410618 Chr.20: 3854891-3854891 on Build GRCh38
rs910035947 Chr.20: 3855048-3855048 on Build GRCh38
rs772837486 Chr.20: 3854736-3854736 on Build GRCh38
rs61256246 Chr.20: 3854909-3854910 on Build GRCh38
rs971478579 Chr.20: 3854862-3854862 on Build GRCh38
rs575308326 Chr.20: 3854854-3854854 on Build GRCh38
rs1049080802 Chr.20: 3855038-3855038 on Build GRCh38
rs374763729 Chr.20: 3854753-3854753 on Build GRCh38
rs1034362011 Chr.20: 3855173-3855173 on Build GRCh38
rs528880433 Chr.20: 3855073-3855073 on Build GRCh38
rs951300194 Chr.20: 3854843-3854843 on Build GRCh38
rs370322140 Chr.20: 3854726-3854726 on Build GRCh38
rs760099301 Chr.20: 3854746-3854746 on Build GRCh38
rs967683667 Chr.20: 3855181-3855181 on Build GRCh38
rs147795132 Chr.20: 3854805-3854805 on Build GRCh38
rs201260999 Chr.20: 3854765-3854765 on Build GRCh38
rs11906417 Chr.20: 3855063-3855063 on Build GRCh38
rs189597273 Chr.20: 3854996-3854996 on Build GRCh38
rs890916750 Chr.20: 3854727-3854727 on Build GRCh38
rs569543732 Chr.20: 3855165-3855165 on Build GRCh38
rs750578843 Chr.20: 3854780-3854780 on Build GRCh38
rs763921462 Chr.20: 3854778-3854778 on Build GRCh38
rs990814876 Chr.20: 3854895-3854895 on Build GRCh38
rs898159852 Chr.20: 3855047-3855047 on Build GRCh38
rs550959130 Chr.20: 3855082-3855082 on Build GRCh38
rs980144014 Chr.20: 3854870-3854870 on Build GRCh38
rs891960914 Chr.20: 3854706-3854706 on Build GRCh38
rs932732341 Chr.20: 3855009-3855009 on Build GRCh38
rs927314456 Chr.20: 3854875-3854875 on Build GRCh38
rs112929008 Chr.20: 3855174-3855174 on Build GRCh38
rs990203779 Chr.20: 3855177-3855177 on Build GRCh38
rs528166167 Chr.20: 3854976-3854976 on Build GRCh38
rs752455730 Chr.20: 3854767-3854767 on Build GRCh38
rs1051254153 Chr.20: 3854719-3854719 on Build GRCh38
rs771147727 Chr.20: 3855171-3855171 on Build GRCh38
rs111465200 Chr.20: 3855179-3855179 on Build GRCh38
rs551615145 Chr.20: 3854930-3854930 on Build GRCh38
rs10154010 Chr.20: 3855065-3855065 on Build GRCh38
rs1024794525 Chr.20: 3855132-3855132 on Build GRCh38
rs775031370 Chr.20: 3854715-3854715 on Build GRCh38
rs377480589 Chr.20: 3854781-3854781 on Build GRCh38
rs758407620 Chr.20: 3854773-3854773 on Build GRCh38
rs182502861 Chr.20: 3854850-3854850 on Build GRCh38
rs904980962 Chr.20: 3855140-3855140 on Build GRCh38
rs913518072 Chr.20: 3854896-3854896 on Build GRCh38
rs374198441 Chr.20: 3854891-3854891 on Build GRCh38
rs376720980 Chr.20: 3854764-3854764 on Build GRCh38
rs940277322 Chr.20: 3855057-3855058 on Build GRCh38
rs748731938 Chr.20: 3854720-3854720 on Build GRCh38
rs541790504 Chr.20: 3854994-3854994 on Build GRCh38
rs1001943093 Chr.20: 3855141-3855141 on Build GRCh38

View Full Product Details