Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6050622-6051125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRN4
Entrez Gene ID
164312
Gene Name
leucine rich repeat neuronal 4
Gene Aliases
C20orf75, NLRR-4, NLRR4, dJ1056H1.1
UniGene
Hs.149133
Gene Chromosome Location
Chr.20: 6040546-6054080 on Build GRCh38
Transcripts
NM_152611.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3841424 c.658G>C p.E220Q Chr.20: 6050981-6050981 on Build GRCh38
6840001 c.781C>A p.Q261K Chr.20: 6050858-6050858 on Build GRCh38
6048533 c.739delG p.D247fs*5 Chr.20: 6050900-6050900 on Build GRCh38
4557644 c.736G>A p.G246R Chr.20: 6050903-6050903 on Build GRCh38
4099992 c.772C>T p.L258L Chr.20: 6050867-6050867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895151257 Chr.20: 6050679-6050679 on Build GRCh38
rs1043822100 Chr.20: 6050723-6050723 on Build GRCh38
rs1029927232 Chr.20: 6050805-6050805 on Build GRCh38
rs757663952 Chr.20: 6050778-6050780 on Build GRCh38
rs939619732 Chr.20: 6050659-6050659 on Build GRCh38
rs750612665 Chr.20: 6050870-6050870 on Build GRCh38
rs957150978 Chr.20: 6050821-6050821 on Build GRCh38
rs549833945 Chr.20: 6051024-6051024 on Build GRCh38
rs759496712 Chr.20: 6050849-6050849 on Build GRCh38
rs749825335 Chr.20: 6050786-6050786 on Build GRCh38
rs538518239 Chr.20: 6051057-6051057 on Build GRCh38
rs745636377 Chr.20: 6050921-6050921 on Build GRCh38
rs776446442 Chr.20: 6050920-6050920 on Build GRCh38
rs772728026 Chr.20: 6050830-6050830 on Build GRCh38
rs553554387 Chr.20: 6051094-6051094 on Build GRCh38
rs866859615 Chr.20: 6051045-6051045 on Build GRCh38
rs757844567 Chr.20: 6050900-6050900 on Build GRCh38
rs777237511 Chr.20: 6050845-6050845 on Build GRCh38
rs758884862 Chr.20: 6050955-6050955 on Build GRCh38
rs779542063 Chr.20: 6050888-6050888 on Build GRCh38
rs145845840 Chr.20: 6050931-6050931 on Build GRCh38
rs769236335 Chr.20: 6050793-6050793 on Build GRCh38
rs751594476 Chr.20: 6050755-6050755 on Build GRCh38
rs766871616 Chr.20: 6050873-6050873 on Build GRCh38
rs766399695 Chr.20: 6050726-6050726 on Build GRCh38
rs149005401 Chr.20: 6050913-6050913 on Build GRCh38
rs781244984 Chr.20: 6050760-6050760 on Build GRCh38
rs764176235 Chr.20: 6050744-6050744 on Build GRCh38
rs1032835682 Chr.20: 6051049-6051049 on Build GRCh38
rs776356227 Chr.20: 6050832-6050832 on Build GRCh38
rs775902316 Chr.20: 6051009-6051009 on Build GRCh38
rs762878328 Chr.20: 6050925-6050925 on Build GRCh38
rs781639147 Chr.20: 6050880-6050880 on Build GRCh38
rs755581748 Chr.20: 6050884-6050884 on Build GRCh38
rs746331127 Chr.20: 6050952-6050952 on Build GRCh38
rs369656470 Chr.20: 6050928-6050928 on Build GRCh38
rs894374837 Chr.20: 6050643-6050643 on Build GRCh38
rs778598801 Chr.20: 6050794-6050794 on Build GRCh38
rs750856108 Chr.20: 6050782-6050782 on Build GRCh38
rs1024239182 Chr.20: 6050692-6050692 on Build GRCh38
rs747630022 Chr.20: 6050801-6050801 on Build GRCh38
rs377635329 Chr.20: 6050727-6050727 on Build GRCh38
rs761782923 Chr.20: 6050859-6050859 on Build GRCh38
rs777354951 Chr.20: 6050743-6050775 on Build GRCh38
rs766678293 Chr.20: 6050937-6050937 on Build GRCh38
rs759404820 Chr.20: 6050738-6050738 on Build GRCh38
rs6053847 Chr.20: 6051085-6051085 on Build GRCh38
rs376601971 Chr.20: 6050987-6050987 on Build GRCh38
rs118117649 Chr.20: 6050803-6050803 on Build GRCh38
rs754214267 Chr.20: 6050880-6050880 on Build GRCh38
rs1028083417 Chr.20: 6051087-6051087 on Build GRCh38
rs903139203 Chr.20: 6051044-6051044 on Build GRCh38
rs780805998 Chr.20: 6051007-6051007 on Build GRCh38
rs957098455 Chr.20: 6051053-6051053 on Build GRCh38
rs1055022246 Chr.20: 6050664-6050664 on Build GRCh38
rs6085368 Chr.20: 6051048-6051048 on Build GRCh38
rs556070083 Chr.20: 6050924-6050924 on Build GRCh38
rs35182819 Chr.20: 6050952-6050952 on Build GRCh38
rs987428121 Chr.20: 6050868-6050868 on Build GRCh38
rs757507866 Chr.20: 6050759-6050759 on Build GRCh38
rs955178548 Chr.20: 6051100-6051100 on Build GRCh38
rs374374787 Chr.20: 6050983-6050983 on Build GRCh38
rs145098571 Chr.20: 6050863-6050863 on Build GRCh38
rs1055235057 Chr.20: 6050941-6050941 on Build GRCh38
rs368831179 Chr.20: 6050656-6050656 on Build GRCh38
rs751936362 Chr.20: 6050754-6050756 on Build GRCh38
rs199654275 Chr.20: 6050906-6050906 on Build GRCh38
rs373531592 Chr.20: 6050940-6050940 on Build GRCh38
rs770571352 Chr.20: 6050914-6050914 on Build GRCh38
rs145884853 Chr.20: 6050893-6050893 on Build GRCh38
rs34360701 Chr.20: 6050831-6050831 on Build GRCh38
rs79597363 Chr.20: 6051021-6051021 on Build GRCh38
rs563183540 Chr.20: 6050711-6050711 on Build GRCh38
rs765913123 Chr.20: 6050947-6050947 on Build GRCh38
rs1013596788 Chr.20: 6050981-6050981 on Build GRCh38
rs189191034 Chr.20: 6051025-6051025 on Build GRCh38
rs749203463 Chr.20: 6051031-6051031 on Build GRCh38
rs780396814 Chr.20: 6050785-6050785 on Build GRCh38
rs751684476 Chr.20: 6050852-6050852 on Build GRCh38
rs940183686 Chr.20: 6050903-6050903 on Build GRCh38
rs777174675 Chr.20: 6050956-6050956 on Build GRCh38

View Full Product Details