Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 9538127-9538636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAK5
Entrez Gene ID
57144
Gene Name
p21 (RAC1) activated kinase 5
Gene Aliases
PAK7
UniGene
Hs.32539
Gene Chromosome Location
Chr.20: 9537390-9839042 on Build GRCh38
Transcripts
NM_020341.3 NM_177990.2 XM_017027960.1 XM_017027961.1 XM_017027962.1 XM_017027963.1 XM_017027964.1 XM_017027965.1

SNP Detail

SNP ID Chromosome Location
rs765860910 Chr.20: 9538348-9538348 on Build GRCh38
rs1008039190 Chr.20: 9538347-9538347 on Build GRCh38
rs1052274510 Chr.20: 9538208-9538208 on Build GRCh38
rs868191829 Chr.20: 9538546-9538546 on Build GRCh38
rs915966735 Chr.20: 9538448-9538448 on Build GRCh38
rs919881476 Chr.20: 9538377-9538377 on Build GRCh38
rs558433363 Chr.20: 9538343-9538343 on Build GRCh38
rs77121410 Chr.20: 9538330-9538330 on Build GRCh38
rs952027888 Chr.20: 9538437-9538437 on Build GRCh38
rs577370753 Chr.20: 9538611-9538611 on Build GRCh38
rs963235918 Chr.20: 9538246-9538246 on Build GRCh38
rs984784130 Chr.20: 9538438-9538438 on Build GRCh38
rs914932039 Chr.20: 9538368-9538368 on Build GRCh38
rs961281938 Chr.20: 9538355-9538355 on Build GRCh38
rs919266641 Chr.20: 9538217-9538217 on Build GRCh38
rs899152583 Chr.20: 9538580-9538580 on Build GRCh38
rs758430629 Chr.20: 9538228-9538228 on Build GRCh38
rs969612945 Chr.20: 9538305-9538305 on Build GRCh38
rs1046332113 Chr.20: 9538299-9538299 on Build GRCh38
rs540369175 Chr.20: 9538543-9538543 on Build GRCh38
rs948902184 Chr.20: 9538296-9538296 on Build GRCh38
rs574421060 Chr.20: 9538466-9538466 on Build GRCh38
rs761793584 Chr.20: 9538170-9538170 on Build GRCh38
rs972048439 Chr.20: 9538202-9538202 on Build GRCh38
rs778066247 Chr.20: 9538591-9538591 on Build GRCh38
rs549684388 Chr.20: 9538166-9538166 on Build GRCh38
rs907412938 Chr.20: 9538311-9538311 on Build GRCh38
rs948834190 Chr.20: 9538463-9538463 on Build GRCh38
rs534583233 Chr.20: 9538285-9538285 on Build GRCh38
rs961611828 Chr.20: 9538194-9538194 on Build GRCh38
rs59858752 Chr.20: 9538548-9538548 on Build GRCh38
rs534309685 Chr.20: 9538369-9538369 on Build GRCh38
rs930677401 Chr.20: 9538226-9538226 on Build GRCh38
rs760224735 Chr.20: 9538331-9538331 on Build GRCh38
rs946390876 Chr.20: 9538381-9538382 on Build GRCh38
rs1049443460 Chr.20: 9538577-9538577 on Build GRCh38
rs994730330 Chr.20: 9538289-9538289 on Build GRCh38
rs893423580 Chr.20: 9538561-9538561 on Build GRCh38
rs889506037 Chr.20: 9538579-9538579 on Build GRCh38
rs1043318456 Chr.20: 9538406-9538406 on Build GRCh38
rs890584562 Chr.20: 9538216-9538216 on Build GRCh38
rs981999970 Chr.20: 9538307-9538308 on Build GRCh38
rs999522021 Chr.20: 9538314-9538314 on Build GRCh38
rs922244552 Chr.20: 9538424-9538425 on Build GRCh38
rs1049129813 Chr.20: 9538231-9538231 on Build GRCh38
rs1036296034 Chr.20: 9538567-9538567 on Build GRCh38
rs569849476 Chr.20: 9538175-9538175 on Build GRCh38
rs554178116 Chr.20: 9538373-9538373 on Build GRCh38
rs193116826 Chr.20: 9538587-9538587 on Build GRCh38
rs767466863 Chr.20: 9538230-9538230 on Build GRCh38
rs187997132 Chr.20: 9538346-9538346 on Build GRCh38
rs947008493 Chr.20: 9538565-9538565 on Build GRCh38
rs1045865730 Chr.20: 9538480-9538480 on Build GRCh38
rs577543861 Chr.20: 9538341-9538341 on Build GRCh38
rs750363870 Chr.20: 9538318-9538318 on Build GRCh38
rs1019390570 Chr.20: 9538607-9538607 on Build GRCh38
rs912453467 Chr.20: 9538176-9538176 on Build GRCh38
rs928788267 Chr.20: 9538530-9538530 on Build GRCh38
rs1051940860 Chr.20: 9538547-9538547 on Build GRCh38
rs953843495 Chr.20: 9538161-9538161 on Build GRCh38
rs1004801600 Chr.20: 9538223-9538223 on Build GRCh38

View Full Product Details