Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18488669-18489194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBBP9
Entrez Gene ID
10741
Gene Name
RB binding protein 9, serine hydrolase
Gene Aliases
BOG, RBBP10
UniGene
Hs.69330
Gene Chromosome Location
Chr.20: 18486540-18497250 on Build GRCh38
Transcripts
NM_006606.2 XM_005260652.2

SNP Detail

SNP ID Chromosome Location
rs533665209 Chr.20: 18489045-18489045 on Build GRCh38
rs977344494 Chr.20: 18488978-18488978 on Build GRCh38
rs898713795 Chr.20: 18488914-18488914 on Build GRCh38
rs940178178 Chr.20: 18489106-18489106 on Build GRCh38
rs79817023 Chr.20: 18488953-18488953 on Build GRCh38
rs1004505296 Chr.20: 18488842-18488842 on Build GRCh38
rs6136389 Chr.20: 18488932-18488932 on Build GRCh38
rs111576824 Chr.20: 18489102-18489102 on Build GRCh38
rs987539575 Chr.20: 18489065-18489065 on Build GRCh38
rs910421029 Chr.20: 18488758-18488758 on Build GRCh38
rs1045925539 Chr.20: 18488763-18488763 on Build GRCh38
rs148012285 Chr.20: 18488969-18488970 on Build GRCh38
rs925882679 Chr.20: 18488987-18488987 on Build GRCh38
rs879406044 Chr.20: 18488972-18488973 on Build GRCh38
rs933444594 Chr.20: 18488736-18488736 on Build GRCh38
rs1053678195 Chr.20: 18488799-18488799 on Build GRCh38
rs1045886872 Chr.20: 18489086-18489086 on Build GRCh38
rs967183836 Chr.20: 18488975-18488975 on Build GRCh38
rs906269392 Chr.20: 18488911-18488911 on Build GRCh38
rs544238903 Chr.20: 18489155-18489155 on Build GRCh38
rs866610637 Chr.20: 18488969-18488969 on Build GRCh38
rs137912009 Chr.20: 18488743-18488743 on Build GRCh38
rs569249964 Chr.20: 18488899-18488899 on Build GRCh38
rs1026354716 Chr.20: 18489063-18489063 on Build GRCh38
rs187894567 Chr.20: 18488947-18488947 on Build GRCh38
rs945010531 Chr.20: 18488900-18488900 on Build GRCh38
rs34425568 Chr.20: 18488986-18488987 on Build GRCh38
rs566252598 Chr.20: 18489050-18489050 on Build GRCh38
rs935879262 Chr.20: 18488990-18488990 on Build GRCh38
rs943206378 Chr.20: 18488783-18488783 on Build GRCh38
rs571560610 Chr.20: 18488722-18488722 on Build GRCh38
rs572574223 Chr.20: 18489042-18489042 on Build GRCh38
rs535171892 Chr.20: 18489132-18489133 on Build GRCh38
rs35822681 Chr.20: 18488971-18488972 on Build GRCh38
rs949851718 Chr.20: 18489064-18489064 on Build GRCh38
rs1017189050 Chr.20: 18488851-18488851 on Build GRCh38
rs774734488 Chr.20: 18488806-18488806 on Build GRCh38
rs373569133 Chr.20: 18488721-18488721 on Build GRCh38
rs73108942 Chr.20: 18488973-18488973 on Build GRCh38
rs142481781 Chr.20: 18489112-18489112 on Build GRCh38
rs550987840 Chr.20: 18488857-18488857 on Build GRCh38
rs554770474 Chr.20: 18488994-18488994 on Build GRCh38
rs150079252 Chr.20: 18489122-18489122 on Build GRCh38
rs200385198 Chr.20: 18488971-18488971 on Build GRCh38
rs993444131 Chr.20: 18489052-18489052 on Build GRCh38
rs74180903 Chr.20: 18488953-18488954 on Build GRCh38
rs1013058056 Chr.20: 18488993-18488993 on Build GRCh38
rs894602130 Chr.20: 18488991-18488991 on Build GRCh38
rs6111989 Chr.20: 18489160-18489160 on Build GRCh38
rs1042099330 Chr.20: 18488910-18488910 on Build GRCh38

View Full Product Details