Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 21397188-21397704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKX2-4
Entrez Gene ID
644524
Gene Name
NK2 homeobox 4
Gene Aliases
NKX2.4, NKX2D
UniGene
Hs.456662
Gene Chromosome Location
Chr.20: 21395367-21397409 on Build GRCh38
Transcripts
NM_033176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7290069 c.148G>A p.A50T Chr.20: 21397252-21397252 on Build GRCh38
5857494 c.82A>G p.S28G Chr.20: 21397318-21397318 on Build GRCh38
4097071 c.101C>T p.A34V Chr.20: 21397299-21397299 on Build GRCh38
5579089 c.167C>T p.S56F Chr.20: 21397233-21397233 on Build GRCh38
121313 c.96C>T p.D32D Chr.20: 21397304-21397304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931799632 Chr.20: 21397558-21397558 on Build GRCh38
rs375499093 Chr.20: 21397354-21397354 on Build GRCh38
rs771572212 Chr.20: 21397269-21397280 on Build GRCh38
rs767421319 Chr.20: 21397414-21397414 on Build GRCh38
rs750323884 Chr.20: 21397415-21397415 on Build GRCh38
rs1014618929 Chr.20: 21397649-21397649 on Build GRCh38
rs766827318 Chr.20: 21397420-21397421 on Build GRCh38
rs748894330 Chr.20: 21397276-21397276 on Build GRCh38
rs987301122 Chr.20: 21397673-21397673 on Build GRCh38
rs746291889 Chr.20: 21397441-21397441 on Build GRCh38
rs561552829 Chr.20: 21397541-21397541 on Build GRCh38
rs748885111 Chr.20: 21397375-21397375 on Build GRCh38
rs892026916 Chr.20: 21397495-21397495 on Build GRCh38
rs921766640 Chr.20: 21397550-21397550 on Build GRCh38
rs1037662264 Chr.20: 21397524-21397524 on Build GRCh38
rs769481693 Chr.20: 21397208-21397208 on Build GRCh38
rs1013354379 Chr.20: 21397647-21397647 on Build GRCh38
rs531397937 Chr.20: 21397529-21397529 on Build GRCh38
rs996741393 Chr.20: 21397419-21397419 on Build GRCh38
rs907719574 Chr.20: 21397518-21397518 on Build GRCh38
rs763099818 Chr.20: 21397339-21397339 on Build GRCh38
rs757117256 Chr.20: 21397352-21397352 on Build GRCh38
rs1011800884 Chr.20: 21397624-21397624 on Build GRCh38
rs368288200 Chr.20: 21397379-21397379 on Build GRCh38
rs760055744 Chr.20: 21397436-21397448 on Build GRCh38
rs1001710798 Chr.20: 21397642-21397642 on Build GRCh38
rs755614656 Chr.20: 21397368-21397368 on Build GRCh38
rs760948311 Chr.20: 21397386-21397386 on Build GRCh38
rs953370975 Chr.20: 21397671-21397671 on Build GRCh38
rs996887530 Chr.20: 21397280-21397280 on Build GRCh38
rs761167183 Chr.20: 21397294-21397294 on Build GRCh38
rs1041833986 Chr.20: 21397566-21397566 on Build GRCh38
rs769807708 Chr.20: 21397400-21397400 on Build GRCh38
rs781549683 Chr.20: 21397438-21397438 on Build GRCh38
rs932747867 Chr.20: 21397480-21397480 on Build GRCh38
rs779720469 Chr.20: 21397372-21397372 on Build GRCh38
rs527557171 Chr.20: 21397616-21397616 on Build GRCh38
rs772421083 Chr.20: 21397282-21397282 on Build GRCh38
rs764193950 Chr.20: 21397409-21397409 on Build GRCh38
rs907793136 Chr.20: 21397469-21397469 on Build GRCh38
rs890179114 Chr.20: 21397545-21397545 on Build GRCh38
rs970226871 Chr.20: 21397523-21397523 on Build GRCh38
rs777082238 Chr.20: 21397388-21397388 on Build GRCh38
rs202223387 Chr.20: 21397643-21397643 on Build GRCh38
rs958719152 Chr.20: 21397642-21397642 on Build GRCh38
rs545561549 Chr.20: 21397462-21397462 on Build GRCh38
rs771306250 Chr.20: 21397386-21397386 on Build GRCh38
rs942201665 Chr.20: 21397475-21397476 on Build GRCh38
rs765333698 Chr.20: 21397325-21397325 on Build GRCh38
rs778666048 Chr.20: 21397377-21397377 on Build GRCh38
rs750411767 Chr.20: 21397361-21397361 on Build GRCh38
rs747864895 Chr.20: 21397437-21397437 on Build GRCh38
rs1033256629 Chr.20: 21397635-21397635 on Build GRCh38
rs766763186 Chr.20: 21397298-21397298 on Build GRCh38
rs761470230 Chr.20: 21397411-21397411 on Build GRCh38
rs960015683 Chr.20: 21397526-21397526 on Build GRCh38
rs778381057 Chr.20: 21397434-21397434 on Build GRCh38
rs775421436 Chr.20: 21397404-21397404 on Build GRCh38
rs777170480 Chr.20: 21397303-21397303 on Build GRCh38
rs973323442 Chr.20: 21397657-21397657 on Build GRCh38
rs1008220612 Chr.20: 21397487-21397492 on Build GRCh38
rs982869315 Chr.20: 21397682-21397682 on Build GRCh38
rs773433132 Chr.20: 21397286-21397286 on Build GRCh38
rs941866515 Chr.20: 21397470-21397470 on Build GRCh38
rs1053032879 Chr.20: 21397582-21397583 on Build GRCh38
rs768185578 Chr.20: 21397376-21397376 on Build GRCh38
rs377225538 Chr.20: 21397424-21397424 on Build GRCh38
rs553457506 Chr.20: 21397275-21397275 on Build GRCh38
rs1031850070 Chr.20: 21397367-21397367 on Build GRCh38
rs928293233 Chr.20: 21397439-21397439 on Build GRCh38
rs200359338 Chr.20: 21397405-21397405 on Build GRCh38
rs1011727367 Chr.20: 21397501-21397501 on Build GRCh38
rs777150633 Chr.20: 21397434-21397457 on Build GRCh38
rs752850268 Chr.20: 21397335-21397335 on Build GRCh38
rs1031923815 Chr.20: 21397489-21397489 on Build GRCh38
rs903362939 Chr.20: 21397584-21397584 on Build GRCh38
rs921862284 Chr.20: 21397669-21397669 on Build GRCh38
rs764436514 Chr.20: 21397342-21397342 on Build GRCh38
rs938608930 Chr.20: 21397241-21397241 on Build GRCh38
rs1006052606 Chr.20: 21397261-21397261 on Build GRCh38
rs906061784 Chr.20: 21397641-21397642 on Build GRCh38
rs754563985 Chr.20: 21397433-21397433 on Build GRCh38
rs760042337 Chr.20: 21397394-21397394 on Build GRCh38
rs1047603118 Chr.20: 21397242-21397242 on Build GRCh38
rs938329316 Chr.20: 21397460-21397460 on Build GRCh38
rs971855675 Chr.20: 21397651-21397651 on Build GRCh38
rs886282707 Chr.20: 21397257-21397257 on Build GRCh38
rs541576256 Chr.20: 21397530-21397530 on Build GRCh38
rs780203277 Chr.20: 21397427-21397427 on Build GRCh38
rs983099951 Chr.20: 21397528-21397528 on Build GRCh38
rs759562308 Chr.20: 21397317-21397317 on Build GRCh38
rs944057932 Chr.20: 21397580-21397580 on Build GRCh38
rs1050164459 Chr.20: 21397544-21397544 on Build GRCh38
rs781256834 Chr.20: 21397360-21397360 on Build GRCh38
rs758347792 Chr.20: 21397608-21397608 on Build GRCh38
rs1037527050 Chr.20: 21397474-21397474 on Build GRCh38
rs1028756433 Chr.20: 21397351-21397351 on Build GRCh38
rs770282111 Chr.20: 21397444-21397444 on Build GRCh38
rs772961256 Chr.20: 21397268-21397269 on Build GRCh38
rs1024325777 Chr.20: 21397648-21397648 on Build GRCh38
rs1008660654 Chr.20: 21397562-21397562 on Build GRCh38
rs562281925 Chr.20: 21397483-21397483 on Build GRCh38
rs751310960 Chr.20: 21397350-21397350 on Build GRCh38
rs940551514 Chr.20: 21397523-21397523 on Build GRCh38
rs753254137 Chr.20: 21397432-21397432 on Build GRCh38

View Full Product Details