Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23603125-23603619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CST9
Entrez Gene ID
128822
Gene Name
cystatin 9
Gene Aliases
CLM, CTES7A
UniGene
Hs.698020
Gene Chromosome Location
Chr.20: 23602410-23605973 on Build GRCh38
Transcripts
NM_001008693.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5857508 c.388T>A p.F130I Chr.20: 23603602-23603602 on Build GRCh38
4097164 c.399C>T p.V133V Chr.20: 23603591-23603591 on Build GRCh38
77535 c.404G>C p.S135T Chr.20: 23603586-23603586 on Build GRCh38
3404981 c.439G>A p.G147R Chr.20: 23603551-23603551 on Build GRCh38
4547413 c.421G>A p.G141R Chr.20: 23603569-23603569 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993081579 Chr.20: 23603273-23603273 on Build GRCh38
rs726217 Chr.20: 23603479-23603479 on Build GRCh38
rs754060890 Chr.20: 23603243-23603243 on Build GRCh38
rs766984489 Chr.20: 23603570-23603570 on Build GRCh38
rs562843756 Chr.20: 23603483-23603483 on Build GRCh38
rs761342879 Chr.20: 23603496-23603496 on Build GRCh38
rs569028379 Chr.20: 23603186-23603186 on Build GRCh38
rs6114191 Chr.20: 23603350-23603350 on Build GRCh38
rs879369286 Chr.20: 23603436-23603436 on Build GRCh38
rs141512970 Chr.20: 23603591-23603591 on Build GRCh38
rs913277481 Chr.20: 23603542-23603542 on Build GRCh38
rs200348736 Chr.20: 23603564-23603564 on Build GRCh38
rs553536989 Chr.20: 23603407-23603407 on Build GRCh38
rs1005645980 Chr.20: 23603413-23603413 on Build GRCh38
rs765692596 Chr.20: 23603507-23603507 on Build GRCh38
rs1014048842 Chr.20: 23603456-23603456 on Build GRCh38
rs755831774 Chr.20: 23603502-23603502 on Build GRCh38
rs936948295 Chr.20: 23603298-23603298 on Build GRCh38
rs372303402 Chr.20: 23603567-23603567 on Build GRCh38
rs772662834 Chr.20: 23603572-23603572 on Build GRCh38
rs143494749 Chr.20: 23603528-23603528 on Build GRCh38
rs917134887 Chr.20: 23603274-23603274 on Build GRCh38
rs374039161 Chr.20: 23603494-23603494 on Build GRCh38
rs936070334 Chr.20: 23603292-23603292 on Build GRCh38
rs922135740 Chr.20: 23603587-23603587 on Build GRCh38
rs770063440 Chr.20: 23603563-23603563 on Build GRCh38
rs975875928 Chr.20: 23603200-23603200 on Build GRCh38
rs769514965 Chr.20: 23603475-23603475 on Build GRCh38
rs1027986371 Chr.20: 23603163-23603163 on Build GRCh38
rs200905994 Chr.20: 23603497-23603497 on Build GRCh38
rs1046513196 Chr.20: 23603427-23603427 on Build GRCh38
rs78410968 Chr.20: 23603365-23603365 on Build GRCh38
rs777334307 Chr.20: 23603546-23603546 on Build GRCh38
rs757305235 Chr.20: 23603541-23603541 on Build GRCh38
rs770914457 Chr.20: 23603535-23603535 on Build GRCh38
rs910411834 Chr.20: 23603174-23603174 on Build GRCh38
rs552349391 Chr.20: 23603569-23603569 on Build GRCh38
rs888133865 Chr.20: 23603408-23603408 on Build GRCh38
rs961424631 Chr.20: 23603446-23603446 on Build GRCh38
rs996431503 Chr.20: 23603162-23603162 on Build GRCh38
rs897265452 Chr.20: 23603469-23603469 on Build GRCh38
rs370765020 Chr.20: 23603529-23603529 on Build GRCh38
rs961210920 Chr.20: 23603261-23603261 on Build GRCh38
rs945078630 Chr.20: 23603258-23603258 on Build GRCh38
rs759692453 Chr.20: 23603461-23603461 on Build GRCh38
rs200359394 Chr.20: 23603545-23603545 on Build GRCh38
rs952459088 Chr.20: 23603180-23603180 on Build GRCh38
rs537666966 Chr.20: 23603187-23603187 on Build GRCh38
rs1053187545 Chr.20: 23603300-23603300 on Build GRCh38
rs924953606 Chr.20: 23603294-23603295 on Build GRCh38
rs148850942 Chr.20: 23603518-23603518 on Build GRCh38
rs1015312440 Chr.20: 23603425-23603425 on Build GRCh38
rs778185964 Chr.20: 23603225-23603225 on Build GRCh38
rs747635704 Chr.20: 23603484-23603485 on Build GRCh38
rs781140039 Chr.20: 23603549-23603549 on Build GRCh38
rs148360730 Chr.20: 23603552-23603552 on Build GRCh38
rs900772199 Chr.20: 23603160-23603160 on Build GRCh38
rs760390828 Chr.20: 23603574-23603574 on Build GRCh38
rs948620248 Chr.20: 23603279-23603279 on Build GRCh38
rs1040946627 Chr.20: 23603361-23603361 on Build GRCh38
rs554615802 Chr.20: 23603199-23603199 on Build GRCh38
rs726216 Chr.20: 23603594-23603594 on Build GRCh38
rs574717700 Chr.20: 23603145-23603145 on Build GRCh38
rs891888767 Chr.20: 23603319-23603319 on Build GRCh38
rs374258117 Chr.20: 23603580-23603580 on Build GRCh38
rs774293022 Chr.20: 23603568-23603568 on Build GRCh38
rs747494517 Chr.20: 23603534-23603534 on Build GRCh38
rs867966806 Chr.20: 23603237-23603237 on Build GRCh38
rs925976961 Chr.20: 23603290-23603290 on Build GRCh38
rs895355474 Chr.20: 23603318-23603318 on Build GRCh38
rs757420756 Chr.20: 23603280-23603280 on Build GRCh38
rs996420138 Chr.20: 23603382-23603382 on Build GRCh38
rs540250292 Chr.20: 23603422-23603422 on Build GRCh38
rs771736382 Chr.20: 23603495-23603496 on Build GRCh38
rs944859982 Chr.20: 23603354-23603354 on Build GRCh38
rs540205199 Chr.20: 23603380-23603380 on Build GRCh38
rs376269565 Chr.20: 23603477-23603477 on Build GRCh38
rs753577662 Chr.20: 23603590-23603590 on Build GRCh38
rs775248126 Chr.20: 23603565-23603565 on Build GRCh38
rs952737799 Chr.20: 23603480-23603480 on Build GRCh38
rs753169628 Chr.20: 23603524-23603524 on Build GRCh38
rs750031879 Chr.20: 23603501-23603501 on Build GRCh38

View Full Product Details