Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 24963677-24964184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APMAP
Entrez Gene ID
57136
Gene Name
adipocyte plasma membrane associated protein
Gene Aliases
BSCv, C20orf3
UniGene
Hs.472330
Gene Chromosome Location
Chr.20: 24962925-24992974 on Build GRCh38
Transcripts
NM_020531.2 XM_005260763.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7117736 c.1168G>A p.E390K Chr.20: 24963896-24963896 on Build GRCh38
5757880 c.1101C>T p.L367L Chr.20: 24963963-24963963 on Build GRCh38
5757881 c.1051C>T p.Q351* Chr.20: 24964013-24964013 on Build GRCh38
477925 c.1209C>G p.F403L Chr.20: 24963855-24963855 on Build GRCh38
5757878 c.1164C>T p.I388I Chr.20: 24963900-24963900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748965399 Chr.20: 24963840-24963840 on Build GRCh38
rs768353350 Chr.20: 24963906-24963906 on Build GRCh38
rs767562843 Chr.20: 24963982-24963983 on Build GRCh38
rs748875666 Chr.20: 24963902-24963902 on Build GRCh38
rs998152301 Chr.20: 24964134-24964134 on Build GRCh38
rs771316582 Chr.20: 24963908-24963908 on Build GRCh38
rs754004636 Chr.20: 24963782-24963782 on Build GRCh38
rs368628252 Chr.20: 24963932-24963932 on Build GRCh38
rs868774926 Chr.20: 24963926-24963926 on Build GRCh38
rs141543812 Chr.20: 24963960-24963960 on Build GRCh38
rs779734592 Chr.20: 24963837-24963837 on Build GRCh38
rs776538519 Chr.20: 24963868-24963868 on Build GRCh38
rs140902364 Chr.20: 24963818-24963818 on Build GRCh38
rs142067594 Chr.20: 24963875-24963875 on Build GRCh38
rs200956189 Chr.20: 24964034-24964034 on Build GRCh38
rs751430954 Chr.20: 24963881-24963881 on Build GRCh38
rs775710141 Chr.20: 24963985-24963985 on Build GRCh38
rs1025159947 Chr.20: 24963937-24963937 on Build GRCh38
rs540535894 Chr.20: 24964111-24964111 on Build GRCh38
rs368949633 Chr.20: 24963812-24963812 on Build GRCh38
rs767242659 Chr.20: 24963953-24963953 on Build GRCh38
rs901279357 Chr.20: 24964146-24964146 on Build GRCh38
rs747436800 Chr.20: 24963907-24963907 on Build GRCh38
rs758218188 Chr.20: 24963795-24963795 on Build GRCh38
rs777147178 Chr.20: 24963921-24963921 on Build GRCh38
rs1045772 Chr.20: 24963811-24963811 on Build GRCh38
rs769406450 Chr.20: 24963832-24963832 on Build GRCh38
rs28364787 Chr.20: 24963774-24963774 on Build GRCh38
rs764683989 Chr.20: 24964022-24964022 on Build GRCh38
rs202155184 Chr.20: 24963923-24963923 on Build GRCh38
rs767278761 Chr.20: 24963992-24963992 on Build GRCh38
rs375790305 Chr.20: 24963882-24963882 on Build GRCh38
rs893954897 Chr.20: 24964087-24964087 on Build GRCh38
rs745451833 Chr.20: 24963831-24963831 on Build GRCh38
rs147898766 Chr.20: 24964072-24964073 on Build GRCh38
rs759669997 Chr.20: 24963786-24963786 on Build GRCh38
rs544710471 Chr.20: 24963724-24963724 on Build GRCh38
rs757715908 Chr.20: 24963809-24963810 on Build GRCh38
rs373605988 Chr.20: 24963979-24963979 on Build GRCh38
rs756689798 Chr.20: 24963793-24963793 on Build GRCh38
rs755575099 Chr.20: 24963888-24963888 on Build GRCh38
rs371057635 Chr.20: 24963978-24963978 on Build GRCh38
rs754433059 Chr.20: 24964020-24964020 on Build GRCh38
rs554356321 Chr.20: 24964023-24964023 on Build GRCh38
rs147024967 Chr.20: 24963988-24963988 on Build GRCh38
rs975669909 Chr.20: 24963789-24963789 on Build GRCh38
rs537070868 Chr.20: 24963954-24963954 on Build GRCh38
rs370222954 Chr.20: 24963897-24963897 on Build GRCh38
rs781606078 Chr.20: 24964035-24964035 on Build GRCh38
rs148619824 Chr.20: 24963870-24963870 on Build GRCh38
rs113742785 Chr.20: 24964028-24964028 on Build GRCh38
rs28364786 Chr.20: 24963944-24963944 on Build GRCh38
rs771531812 Chr.20: 24963854-24963854 on Build GRCh38
rs781230950 Chr.20: 24963884-24963884 on Build GRCh38
rs375375077 Chr.20: 24963924-24963924 on Build GRCh38
rs553194798 Chr.20: 24963986-24963986 on Build GRCh38
rs778867719 Chr.20: 24964060-24964060 on Build GRCh38
rs749835503 Chr.20: 24964051-24964051 on Build GRCh38
rs370420110 Chr.20: 24964054-24964054 on Build GRCh38
rs201276168 Chr.20: 24963787-24963787 on Build GRCh38
rs536436450 Chr.20: 24963983-24963983 on Build GRCh38
rs529991245 Chr.20: 24964068-24964068 on Build GRCh38
rs746303223 Chr.20: 24964036-24964036 on Build GRCh38
rs926690441 Chr.20: 24963715-24963715 on Build GRCh38
rs777741358 Chr.20: 24963796-24963796 on Build GRCh38
rs746317654 Chr.20: 24963981-24963981 on Build GRCh38
rs145319394 Chr.20: 24963824-24963824 on Build GRCh38
rs933156466 Chr.20: 24963980-24963980 on Build GRCh38
rs774586855 Chr.20: 24963987-24963987 on Build GRCh38
rs989545848 Chr.20: 24963959-24963959 on Build GRCh38
rs977962909 Chr.20: 24963750-24963750 on Build GRCh38
rs532361283 Chr.20: 24964147-24964147 on Build GRCh38
rs1048742750 Chr.20: 24963814-24963814 on Build GRCh38
rs1021026277 Chr.20: 24964119-24964119 on Build GRCh38
rs145342112 Chr.20: 24963975-24963975 on Build GRCh38
rs754589371 Chr.20: 24963969-24963969 on Build GRCh38
rs986434251 Chr.20: 24964162-24964162 on Build GRCh38
rs376982662 Chr.20: 24964038-24964038 on Build GRCh38
rs564662290 Chr.20: 24964006-24964006 on Build GRCh38
rs11550622 Chr.20: 24964005-24964005 on Build GRCh38
rs760598487 Chr.20: 24963998-24963998 on Build GRCh38
rs958289101 Chr.20: 24963723-24963723 on Build GRCh38
rs981285341 Chr.20: 24963708-24963708 on Build GRCh38
rs761717180 Chr.20: 24963948-24963948 on Build GRCh38
rs114304241 Chr.20: 24964037-24964037 on Build GRCh38
rs200397867 Chr.20: 24963952-24963952 on Build GRCh38
rs752517715 Chr.20: 24963788-24963788 on Build GRCh38
rs151131612 Chr.20: 24963887-24963887 on Build GRCh38
rs763255949 Chr.20: 24963938-24963938 on Build GRCh38
rs1055635977 Chr.20: 24963880-24963880 on Build GRCh38
rs551876491 Chr.20: 24964148-24964148 on Build GRCh38
rs112413522 Chr.20: 24963761-24963761 on Build GRCh38
rs999296095 Chr.20: 24964098-24964098 on Build GRCh38
rs748347548 Chr.20: 24964066-24964066 on Build GRCh38
rs947342825 Chr.20: 24964055-24964055 on Build GRCh38
rs762767449 Chr.20: 24964077-24964077 on Build GRCh38
rs367779402 Chr.20: 24963996-24963996 on Build GRCh38
rs764444977 Chr.20: 24963943-24963943 on Build GRCh38
rs777146172 Chr.20: 24963873-24963873 on Build GRCh38
rs772388741 Chr.20: 24963846-24963846 on Build GRCh38
rs764299774 Chr.20: 24964112-24964112 on Build GRCh38
rs111757368 Chr.20: 24963847-24963847 on Build GRCh38
rs966671331 Chr.20: 24963962-24963962 on Build GRCh38
rs1011007676 Chr.20: 24964088-24964088 on Build GRCh38
rs765859623 Chr.20: 24963955-24963955 on Build GRCh38
rs564159918 Chr.20: 24964120-24964120 on Build GRCh38

View Full Product Details